The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001941.4 | NP_001932.2 | 1 - 2 | 224 | 115 |
NM_024423.3 | NP_077741.2 | 1 - 2 | 224 | 115 | |
GenBank mRNA | AK291748.1 | - | 1 - 2 | 150 | 115 |
D17427.1 | - | 1 - 2 | 136 | 115 | |
DA860471.1 | - | 1 - 2 | 224 | 115 | |
X83929.1 | - | 1 - 3 | 90 | 115 |