The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007046.3 | NP_008977.1 | 1 - 2 | 673 | 118 |
GenBank mRNA | AF088916.1 | - | 1 - 2 | 174 | 118 |
AK075517.1 | - | 1 - 2 | 598 | 118 | |
AK307539.1 | - | 1 - 2 | 602 | 118 | |
AL050138.1 | - | 1 - 2 | 104 | 118 | |
AL552691.3 | - | 1 - 2 | 679 | 118 | |
BC007530.1 | - | 1 - 2 | 345 | 118 | |
BC136279.1 | - | 1 - 2 | 273 | 118 | |
BC142638.1 | - | 1 - 2 | 606 | 118 | |
BC142688.1 | - | 1 - 2 | 555 | 118 | |
CN350368.1 | - | 1 - 2 | 614 | 118 |