Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79833682-79834171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX4
Entrez Gene ID
8535
Gene Name
chromobox 4
Gene Aliases
NBP16, PC2
UniGene
Hs.405046
Gene Chromosome Location
Chr.17: 79833156-79839414 on Build GRCh38
Transcripts
NM_003655.2 XM_011525399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387552 c.1638C>T p.T546T Chr.17: 79834004-79834004 on Build GRCh38
6854883 c.1539G>A p.T513T Chr.17: 79834103-79834103 on Build GRCh38
6454349 c.1677G>A p.T559T Chr.17: 79833965-79833965 on Build GRCh38
7113764 c.1662C>A p.F554L Chr.17: 79833980-79833980 on Build GRCh38
4467607 c.1499C>T p.S500L Chr.17: 79834143-79834143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771101934 Chr.17: 79834091-79834091 on Build GRCh38
rs924884978 Chr.17: 79834139-79834144 on Build GRCh38
rs948770975 Chr.17: 79833899-79833899 on Build GRCh38
rs756917393 Chr.17: 79833998-79833998 on Build GRCh38
rs767323954 Chr.17: 79833915-79833915 on Build GRCh38
rs760477690 Chr.17: 79833970-79833970 on Build GRCh38
rs756548734 Chr.17: 79834075-79834075 on Build GRCh38
rs992599983 Chr.17: 79834037-79834037 on Build GRCh38
rs376832453 Chr.17: 79834131-79834131 on Build GRCh38
rs963828967 Chr.17: 79833821-79833821 on Build GRCh38
rs764759904 Chr.17: 79834122-79834123 on Build GRCh38
rs200238347 Chr.17: 79833917-79833917 on Build GRCh38
rs757906326 Chr.17: 79834134-79834135 on Build GRCh38
rs564210946 Chr.17: 79834072-79834072 on Build GRCh38
rs781587836 Chr.17: 79833928-79833928 on Build GRCh38
rs200965379 Chr.17: 79834128-79834128 on Build GRCh38
rs759489445 Chr.17: 79833950-79833950 on Build GRCh38
rs377592612 Chr.17: 79833909-79833909 on Build GRCh38
rs761409501 Chr.17: 79834123-79834128 on Build GRCh38
rs775410592 Chr.17: 79834104-79834104 on Build GRCh38
rs549712900 Chr.17: 79834100-79834100 on Build GRCh38
rs764780851 Chr.17: 79834004-79834004 on Build GRCh38
rs969000696 Chr.17: 79833947-79833947 on Build GRCh38
rs778352766 Chr.17: 79833922-79833922 on Build GRCh38
rs761494208 Chr.17: 79834124-79834124 on Build GRCh38
rs745827861 Chr.17: 79834111-79834112 on Build GRCh38
rs747536556 Chr.17: 79833923-79833923 on Build GRCh38
rs146523753 Chr.17: 79834058-79834058 on Build GRCh38
rs754126884 Chr.17: 79834118-79834119 on Build GRCh38
rs975573572 Chr.17: 79833824-79833824 on Build GRCh38
rs771147949 Chr.17: 79833917-79833917 on Build GRCh38
rs763670648 Chr.17: 79833992-79833992 on Build GRCh38
rs774520158 Chr.17: 79833923-79833924 on Build GRCh38
rs1007896340 Chr.17: 79834077-79834077 on Build GRCh38
rs914713991 Chr.17: 79833826-79833826 on Build GRCh38
rs757454510 Chr.17: 79834145-79834145 on Build GRCh38
rs1012031571 Chr.17: 79833712-79833712 on Build GRCh38
rs1040796552 Chr.17: 79833889-79833889 on Build GRCh38
rs180834650 Chr.17: 79833756-79833756 on Build GRCh38
rs770089482 Chr.17: 79833936-79833936 on Build GRCh38
rs776282365 Chr.17: 79834120-79834134 on Build GRCh38
rs753484290 Chr.17: 79833996-79833996 on Build GRCh38
rs753247490 Chr.17: 79834136-79834136 on Build GRCh38
rs996569274 Chr.17: 79833910-79833910 on Build GRCh38
rs529192897 Chr.17: 79834137-79834137 on Build GRCh38
rs771033233 Chr.17: 79833949-79833949 on Build GRCh38
rs924885077 Chr.17: 79833860-79833860 on Build GRCh38
rs547706029 Chr.17: 79834148-79834148 on Build GRCh38
rs752531500 Chr.17: 79833916-79833916 on Build GRCh38
rs377289234 Chr.17: 79833966-79833966 on Build GRCh38
rs867027777 Chr.17: 79833887-79833887 on Build GRCh38
rs764827212 Chr.17: 79834125-79834125 on Build GRCh38
rs773577839 Chr.17: 79833937-79833937 on Build GRCh38
rs748551572 Chr.17: 79833935-79833935 on Build GRCh38
rs968611252 Chr.17: 79833705-79833705 on Build GRCh38
rs749940879 Chr.17: 79834049-79834049 on Build GRCh38
rs978588232 Chr.17: 79833715-79833715 on Build GRCh38
rs555880348 Chr.17: 79833820-79833820 on Build GRCh38
rs900524567 Chr.17: 79833907-79833907 on Build GRCh38
rs749454180 Chr.17: 79833916-79833917 on Build GRCh38
rs916897286 Chr.17: 79833750-79833750 on Build GRCh38
rs749975542 Chr.17: 79834127-79834128 on Build GRCh38
rs774452384 Chr.17: 79834098-79834098 on Build GRCh38
rs1012468134 Chr.17: 79833725-79833725 on Build GRCh38
rs749648445 Chr.17: 79834142-79834142 on Build GRCh38
rs749310461 Chr.17: 79833945-79833945 on Build GRCh38
rs541674947 Chr.17: 79833918-79833918 on Build GRCh38
rs779018829 Chr.17: 79834146-79834146 on Build GRCh38
rs749503302 Chr.17: 79834090-79834090 on Build GRCh38
rs201780947 Chr.17: 79834140-79834140 on Build GRCh38
rs760673997 Chr.17: 79834110-79834110 on Build GRCh38
rs763862805 Chr.17: 79834115-79834115 on Build GRCh38
rs934364653 Chr.17: 79833726-79833726 on Build GRCh38
rs769503411 Chr.17: 79833757-79833757 on Build GRCh38
rs781769596 Chr.17: 79834070-79834070 on Build GRCh38
rs1019358082 Chr.17: 79833811-79833811 on Build GRCh38
rs568219945 Chr.17: 79834127-79834127 on Build GRCh38
rs776459002 Chr.17: 79834118-79834118 on Build GRCh38
rs144757374 Chr.17: 79833921-79833921 on Build GRCh38
rs1047848880 Chr.17: 79833834-79833834 on Build GRCh38
rs977671141 Chr.17: 79833858-79833859 on Build GRCh38
rs867588853 Chr.17: 79834081-79834081 on Build GRCh38
rs1021803587 Chr.17: 79834123-79834134 on Build GRCh38
rs1045184900 Chr.17: 79833735-79833735 on Build GRCh38
rs545813747 Chr.17: 79833926-79833926 on Build GRCh38
rs767389624 Chr.17: 79833965-79833965 on Build GRCh38
rs1044679739 Chr.17: 79833758-79833758 on Build GRCh38
rs886535448 Chr.17: 79833873-79833873 on Build GRCh38
rs908027065 Chr.17: 79834139-79834139 on Build GRCh38
rs916970832 Chr.17: 79834048-79834048 on Build GRCh38
rs992506882 Chr.17: 79833739-79833739 on Build GRCh38
rs771989725 Chr.17: 79834103-79834103 on Build GRCh38
rs531394505 Chr.17: 79834095-79834095 on Build GRCh38
rs778343571 Chr.17: 79834076-79834076 on Build GRCh38
rs543616093 Chr.17: 79833791-79833791 on Build GRCh38
rs899402661 Chr.17: 79833772-79833772 on Build GRCh38
rs866315866 Chr.17: 79833829-79833829 on Build GRCh38

View Full Product Details