Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79833954-79834441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX4
Entrez Gene ID
8535
Gene Name
chromobox 4
Gene Aliases
NBP16, PC2
UniGene
Hs.405046
Gene Chromosome Location
Chr.17: 79833156-79839414 on Build GRCh38
Transcripts
NM_003655.2 XM_011525399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6435101 c.1325G>T p.S442I Chr.17: 79834317-79834317 on Build GRCh38
3388267 c.1439C>T p.T480M Chr.17: 79834203-79834203 on Build GRCh38
4400793 c.1421A>G p.D474G Chr.17: 79834221-79834221 on Build GRCh38
4462952 c.1269C>G p.P423P Chr.17: 79834373-79834373 on Build GRCh38
5473962 c.1262G>A p.R421H Chr.17: 79834380-79834380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771101934 Chr.17: 79834091-79834091 on Build GRCh38
rs924884978 Chr.17: 79834139-79834144 on Build GRCh38
rs762730314 Chr.17: 79834178-79834178 on Build GRCh38
rs756917393 Chr.17: 79833998-79833998 on Build GRCh38
rs569815846 Chr.17: 79834314-79834314 on Build GRCh38
rs745582907 Chr.17: 79834329-79834329 on Build GRCh38
rs780077146 Chr.17: 79834241-79834241 on Build GRCh38
rs772135793 Chr.17: 79834152-79834152 on Build GRCh38
rs758579083 Chr.17: 79834295-79834295 on Build GRCh38
rs756548734 Chr.17: 79834075-79834075 on Build GRCh38
rs992599983 Chr.17: 79834037-79834037 on Build GRCh38
rs376832453 Chr.17: 79834131-79834131 on Build GRCh38
rs764759904 Chr.17: 79834122-79834123 on Build GRCh38
rs757906326 Chr.17: 79834134-79834135 on Build GRCh38
rs762486081 Chr.17: 79834279-79834279 on Build GRCh38
rs564210946 Chr.17: 79834072-79834072 on Build GRCh38
rs768529966 Chr.17: 79834258-79834258 on Build GRCh38
rs200965379 Chr.17: 79834128-79834128 on Build GRCh38
rs201414154 Chr.17: 79834157-79834157 on Build GRCh38
rs761409501 Chr.17: 79834123-79834128 on Build GRCh38
rs549712900 Chr.17: 79834100-79834100 on Build GRCh38
rs775410592 Chr.17: 79834104-79834104 on Build GRCh38
rs996192935 Chr.17: 79834252-79834252 on Build GRCh38
rs764780851 Chr.17: 79834004-79834004 on Build GRCh38
rs201298776 Chr.17: 79834387-79834387 on Build GRCh38
rs761494208 Chr.17: 79834124-79834124 on Build GRCh38
rs761227308 Chr.17: 79834193-79834193 on Build GRCh38
rs999023886 Chr.17: 79834341-79834341 on Build GRCh38
rs765968311 Chr.17: 79834187-79834187 on Build GRCh38
rs745827861 Chr.17: 79834111-79834112 on Build GRCh38
rs146523753 Chr.17: 79834058-79834058 on Build GRCh38
rs780165123 Chr.17: 79834156-79834156 on Build GRCh38
rs754126884 Chr.17: 79834118-79834119 on Build GRCh38
rs769467167 Chr.17: 79834277-79834277 on Build GRCh38
rs537118948 Chr.17: 79834320-79834320 on Build GRCh38
rs763670648 Chr.17: 79833992-79833992 on Build GRCh38
rs757114531 Chr.17: 79834401-79834401 on Build GRCh38
rs1007896340 Chr.17: 79834077-79834077 on Build GRCh38
rs757454510 Chr.17: 79834145-79834145 on Build GRCh38
rs769427551 Chr.17: 79834165-79834165 on Build GRCh38
rs565830961 Chr.17: 79834161-79834161 on Build GRCh38
rs12949594 Chr.17: 79834180-79834180 on Build GRCh38
rs1006682227 Chr.17: 79834339-79834339 on Build GRCh38
rs375182651 Chr.17: 79834332-79834332 on Build GRCh38
rs776282365 Chr.17: 79834120-79834134 on Build GRCh38
rs753484290 Chr.17: 79833996-79833996 on Build GRCh38
rs753247490 Chr.17: 79834136-79834136 on Build GRCh38
rs557687848 Chr.17: 79834303-79834303 on Build GRCh38
rs62075209 Chr.17: 79834334-79834334 on Build GRCh38
rs1024096639 Chr.17: 79834349-79834349 on Build GRCh38
rs781397691 Chr.17: 79834306-79834306 on Build GRCh38
rs773952129 Chr.17: 79834283-79834283 on Build GRCh38
rs762442225 Chr.17: 79834291-79834291 on Build GRCh38
rs578248026 Chr.17: 79834394-79834394 on Build GRCh38
rs529192897 Chr.17: 79834137-79834137 on Build GRCh38
rs777393766 Chr.17: 79834310-79834310 on Build GRCh38
rs915873206 Chr.17: 79834159-79834159 on Build GRCh38
rs547706029 Chr.17: 79834148-79834148 on Build GRCh38
rs773114344 Chr.17: 79834278-79834278 on Build GRCh38
rs754297150 Chr.17: 79834289-79834289 on Build GRCh38
rs764827212 Chr.17: 79834125-79834125 on Build GRCh38
rs770743303 Chr.17: 79834281-79834281 on Build GRCh38
rs749940879 Chr.17: 79834049-79834049 on Build GRCh38
rs757568848 Chr.17: 79834200-79834200 on Build GRCh38
rs545823788 Chr.17: 79834415-79834415 on Build GRCh38
rs778909756 Chr.17: 79834406-79834406 on Build GRCh38
rs749975542 Chr.17: 79834127-79834128 on Build GRCh38
rs756120158 Chr.17: 79834309-79834309 on Build GRCh38
rs774452384 Chr.17: 79834098-79834098 on Build GRCh38
rs751772343 Chr.17: 79834379-79834379 on Build GRCh38
rs749648445 Chr.17: 79834142-79834142 on Build GRCh38
rs372900825 Chr.17: 79834385-79834385 on Build GRCh38
rs1032221588 Chr.17: 79834368-79834368 on Build GRCh38
rs775116136 Chr.17: 79834333-79834333 on Build GRCh38
rs751160083 Chr.17: 79834189-79834189 on Build GRCh38
rs952318533 Chr.17: 79834391-79834391 on Build GRCh38
rs779018829 Chr.17: 79834146-79834146 on Build GRCh38
rs749503302 Chr.17: 79834090-79834090 on Build GRCh38
rs201780947 Chr.17: 79834140-79834140 on Build GRCh38
rs970173980 Chr.17: 79834372-79834372 on Build GRCh38
rs199722528 Chr.17: 79834169-79834169 on Build GRCh38
rs763320095 Chr.17: 79834371-79834371 on Build GRCh38
rs1040492308 Chr.17: 79834171-79834171 on Build GRCh38
rs760673997 Chr.17: 79834110-79834110 on Build GRCh38
rs773400384 Chr.17: 79834363-79834363 on Build GRCh38
rs781003050 Chr.17: 79834263-79834263 on Build GRCh38
rs780276613 Chr.17: 79834298-79834298 on Build GRCh38
rs763862805 Chr.17: 79834115-79834115 on Build GRCh38
rs779400926 Chr.17: 79834205-79834205 on Build GRCh38
rs759134406 Chr.17: 79834285-79834285 on Build GRCh38
rs755248434 Chr.17: 79834384-79834384 on Build GRCh38
rs900577226 Chr.17: 79834191-79834191 on Build GRCh38
rs1014494992 Chr.17: 79834347-79834347 on Build GRCh38
rs781769596 Chr.17: 79834070-79834070 on Build GRCh38
rs764722519 Chr.17: 79834195-79834195 on Build GRCh38
rs568219945 Chr.17: 79834127-79834127 on Build GRCh38
rs776459002 Chr.17: 79834118-79834118 on Build GRCh38
rs748019695 Chr.17: 79834308-79834308 on Build GRCh38
rs765495146 Chr.17: 79834292-79834292 on Build GRCh38
rs867588853 Chr.17: 79834081-79834081 on Build GRCh38
rs1021803587 Chr.17: 79834123-79834134 on Build GRCh38
rs766517517 Chr.17: 79834374-79834374 on Build GRCh38
rs747121317 Chr.17: 79834256-79834256 on Build GRCh38
rs908122277 Chr.17: 79834412-79834412 on Build GRCh38
rs931144078 Chr.17: 79834260-79834260 on Build GRCh38
rs747907856 Chr.17: 79834269-79834269 on Build GRCh38
rs751578106 Chr.17: 79834299-79834299 on Build GRCh38
rs374900659 Chr.17: 79834388-79834388 on Build GRCh38
rs765803327 Chr.17: 79834345-79834345 on Build GRCh38
rs908027065 Chr.17: 79834139-79834139 on Build GRCh38
rs758667165 Chr.17: 79834216-79834216 on Build GRCh38
rs916970832 Chr.17: 79834048-79834048 on Build GRCh38
rs770653490 Chr.17: 79834319-79834319 on Build GRCh38
rs372077305 Chr.17: 79834196-79834196 on Build GRCh38
rs764630148 Chr.17: 79834287-79834287 on Build GRCh38
rs771989725 Chr.17: 79834103-79834103 on Build GRCh38
rs531394505 Chr.17: 79834095-79834095 on Build GRCh38
rs778343571 Chr.17: 79834076-79834076 on Build GRCh38
rs750598235 Chr.17: 79834211-79834211 on Build GRCh38
rs750837988 Chr.17: 79834294-79834294 on Build GRCh38

View Full Product Details