Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79834236-79834732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX4
Entrez Gene ID
8535
Gene Name
chromobox 4
Gene Aliases
NBP16, PC2
UniGene
Hs.405046
Gene Chromosome Location
Chr.17: 79833156-79839414 on Build GRCh38
Transcripts
NM_003655.2 XM_011525399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6435101 c.1325G>T p.S442I Chr.17: 79834317-79834317 on Build GRCh38
309883 c.1008C>G p.S336R Chr.17: 79834634-79834634 on Build GRCh38
5350420 c.972G>A p.P324P Chr.17: 79834670-79834670 on Build GRCh38
6437706 c.1030G>T p.G344C Chr.17: 79834612-79834612 on Build GRCh38
3523240 c.1371C>T p.L457L Chr.17: 79834271-79834271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746909019 Chr.17: 79834682-79834684 on Build GRCh38
rs569815846 Chr.17: 79834314-79834314 on Build GRCh38
rs755619971 Chr.17: 79834682-79834682 on Build GRCh38
rs758870576 Chr.17: 79834691-79834691 on Build GRCh38
rs745582907 Chr.17: 79834329-79834329 on Build GRCh38
rs766713791 Chr.17: 79834483-79834483 on Build GRCh38
rs758579083 Chr.17: 79834295-79834295 on Build GRCh38
rs561614557 Chr.17: 79834618-79834618 on Build GRCh38
rs1047818084 Chr.17: 79834442-79834456 on Build GRCh38
rs754632700 Chr.17: 79834611-79834611 on Build GRCh38
rs762486081 Chr.17: 79834279-79834279 on Build GRCh38
rs768529966 Chr.17: 79834258-79834258 on Build GRCh38
rs749542196 Chr.17: 79834442-79834453 on Build GRCh38
rs746437560 Chr.17: 79834542-79834542 on Build GRCh38
rs143178915 Chr.17: 79834534-79834534 on Build GRCh38
rs768939986 Chr.17: 79834698-79834698 on Build GRCh38
rs751349665 Chr.17: 79834670-79834670 on Build GRCh38
rs767968687 Chr.17: 79834440-79834441 on Build GRCh38
rs201298776 Chr.17: 79834387-79834387 on Build GRCh38
rs1001670894 Chr.17: 79834613-79834613 on Build GRCh38
rs999023886 Chr.17: 79834341-79834341 on Build GRCh38
rs757314416 Chr.17: 79834523-79834523 on Build GRCh38
rs750369340 Chr.17: 79834645-79834645 on Build GRCh38
rs771371633 Chr.17: 79834629-79834629 on Build GRCh38
rs762852218 Chr.17: 79834655-79834655 on Build GRCh38
rs374344512 Chr.17: 79834639-79834639 on Build GRCh38
rs769467167 Chr.17: 79834277-79834277 on Build GRCh38
rs537118948 Chr.17: 79834320-79834320 on Build GRCh38
rs753814271 Chr.17: 79834512-79834512 on Build GRCh38
rs757114531 Chr.17: 79834401-79834401 on Build GRCh38
rs763513931 Chr.17: 79834477-79834477 on Build GRCh38
rs150413647 Chr.17: 79834671-79834671 on Build GRCh38
rs749771267 Chr.17: 79834564-79834564 on Build GRCh38
rs772175815 Chr.17: 79834479-79834484 on Build GRCh38
rs773710155 Chr.17: 79834457-79834457 on Build GRCh38
rs767806014 Chr.17: 79834487-79834487 on Build GRCh38
rs1006682227 Chr.17: 79834339-79834339 on Build GRCh38
rs754013674 Chr.17: 79834593-79834593 on Build GRCh38
rs375182651 Chr.17: 79834332-79834332 on Build GRCh38
rs138355116 Chr.17: 79834672-79834672 on Build GRCh38
rs780734474 Chr.17: 79834555-79834555 on Build GRCh38
rs759961240 Chr.17: 79834580-79834580 on Build GRCh38
rs557687848 Chr.17: 79834303-79834303 on Build GRCh38
rs62075209 Chr.17: 79834334-79834334 on Build GRCh38
rs372584226 Chr.17: 79834541-79834541 on Build GRCh38
rs1024096639 Chr.17: 79834349-79834349 on Build GRCh38
rs781397691 Chr.17: 79834306-79834306 on Build GRCh38
rs773952129 Chr.17: 79834283-79834283 on Build GRCh38
rs762442225 Chr.17: 79834291-79834291 on Build GRCh38
rs578248026 Chr.17: 79834394-79834394 on Build GRCh38
rs976289571 Chr.17: 79834669-79834669 on Build GRCh38
rs774820335 Chr.17: 79834442-79834442 on Build GRCh38
rs768363069 Chr.17: 79834688-79834690 on Build GRCh38
rs777393766 Chr.17: 79834310-79834310 on Build GRCh38
rs772123871 Chr.17: 79834582-79834582 on Build GRCh38
rs201691344 Chr.17: 79834696-79834696 on Build GRCh38
rs1015171091 Chr.17: 79834652-79834652 on Build GRCh38
rs773114344 Chr.17: 79834278-79834278 on Build GRCh38
rs754297150 Chr.17: 79834289-79834289 on Build GRCh38
rs770743303 Chr.17: 79834281-79834281 on Build GRCh38
rs140935310 Chr.17: 79834701-79834701 on Build GRCh38
rs758217068 Chr.17: 79834595-79834595 on Build GRCh38
rs776802623 Chr.17: 79834643-79834643 on Build GRCh38
rs545823788 Chr.17: 79834415-79834415 on Build GRCh38
rs767173541 Chr.17: 79834679-79834679 on Build GRCh38
rs778909756 Chr.17: 79834406-79834406 on Build GRCh38
rs756120158 Chr.17: 79834309-79834309 on Build GRCh38
rs751772343 Chr.17: 79834379-79834379 on Build GRCh38
rs908338836 Chr.17: 79834445-79834445 on Build GRCh38
rs760864836 Chr.17: 79834590-79834590 on Build GRCh38
rs372900825 Chr.17: 79834385-79834385 on Build GRCh38
rs764247948 Chr.17: 79834592-79834592 on Build GRCh38
rs770141347 Chr.17: 79834451-79834451 on Build GRCh38
rs148095227 Chr.17: 79834664-79834664 on Build GRCh38
rs1032221588 Chr.17: 79834368-79834368 on Build GRCh38
rs751206406 Chr.17: 79834601-79834601 on Build GRCh38
rs775116136 Chr.17: 79834333-79834333 on Build GRCh38
rs943862891 Chr.17: 79834450-79834450 on Build GRCh38
rs952318533 Chr.17: 79834391-79834391 on Build GRCh38
rs746551157 Chr.17: 79834448-79834448 on Build GRCh38
rs970173980 Chr.17: 79834372-79834372 on Build GRCh38
rs763320095 Chr.17: 79834371-79834371 on Build GRCh38
rs779401391 Chr.17: 79834630-79834630 on Build GRCh38
rs760922762 Chr.17: 79834642-79834642 on Build GRCh38
rs564025198 Chr.17: 79834484-79834484 on Build GRCh38
rs773400384 Chr.17: 79834363-79834363 on Build GRCh38
rs781003050 Chr.17: 79834263-79834263 on Build GRCh38
rs765286538 Chr.17: 79834594-79834594 on Build GRCh38
rs780276613 Chr.17: 79834298-79834298 on Build GRCh38
rs756437476 Chr.17: 79834442-79834444 on Build GRCh38
rs779242736 Chr.17: 79834442-79834447 on Build GRCh38
rs778821541 Chr.17: 79834526-79834526 on Build GRCh38
rs759134406 Chr.17: 79834285-79834285 on Build GRCh38
rs775780673 Chr.17: 79834583-79834583 on Build GRCh38
rs755248434 Chr.17: 79834384-79834384 on Build GRCh38
rs1014494992 Chr.17: 79834347-79834347 on Build GRCh38
rs200178625 Chr.17: 79834565-79834565 on Build GRCh38
rs748019695 Chr.17: 79834308-79834308 on Build GRCh38
rs765495146 Chr.17: 79834292-79834292 on Build GRCh38
rs774653368 Chr.17: 79834578-79834578 on Build GRCh38
rs775297086 Chr.17: 79834483-79834503 on Build GRCh38
rs553690928 Chr.17: 79834690-79834690 on Build GRCh38
rs200817261 Chr.17: 79834491-79834491 on Build GRCh38
rs766517517 Chr.17: 79834374-79834374 on Build GRCh38
rs150230495 Chr.17: 79834702-79834702 on Build GRCh38
rs377573080 Chr.17: 79834596-79834596 on Build GRCh38
rs905628116 Chr.17: 79834610-79834610 on Build GRCh38
rs3833850 Chr.17: 79834442-79834450 on Build GRCh38
rs970019924 Chr.17: 79834431-79834431 on Build GRCh38
rs562956582 Chr.17: 79834441-79834442 on Build GRCh38
rs776374574 Chr.17: 79834699-79834701 on Build GRCh38
rs908122277 Chr.17: 79834412-79834412 on Build GRCh38
rs931144078 Chr.17: 79834260-79834260 on Build GRCh38
rs747907856 Chr.17: 79834269-79834269 on Build GRCh38
rs374900659 Chr.17: 79834388-79834388 on Build GRCh38
rs768205139 Chr.17: 79834543-79834543 on Build GRCh38
rs751578106 Chr.17: 79834299-79834299 on Build GRCh38
rs745991606 Chr.17: 79834478-79834479 on Build GRCh38
rs765803327 Chr.17: 79834345-79834345 on Build GRCh38
rs368921956 Chr.17: 79834674-79834674 on Build GRCh38
rs770653490 Chr.17: 79834319-79834319 on Build GRCh38
rs771797783 Chr.17: 79834436-79834436 on Build GRCh38
rs764147049 Chr.17: 79834508-79834508 on Build GRCh38
rs764630148 Chr.17: 79834287-79834287 on Build GRCh38
rs1043751560 Chr.17: 79834511-79834511 on Build GRCh38
rs1024526002 Chr.17: 79834608-79834608 on Build GRCh38
rs757936894 Chr.17: 79834538-79834538 on Build GRCh38
rs752665414 Chr.17: 79834489-79834489 on Build GRCh38
rs777325836 Chr.17: 79834688-79834688 on Build GRCh38
rs1014405016 Chr.17: 79834604-79834604 on Build GRCh38
rs750837988 Chr.17: 79834294-79834294 on Build GRCh38
rs371126696 Chr.17: 79834634-79834634 on Build GRCh38
rs752138935 Chr.17: 79834681-79834681 on Build GRCh38
rs944911073 Chr.17: 79834438-79834438 on Build GRCh38
rs377665529 Chr.17: 79834640-79834640 on Build GRCh38

View Full Product Details