The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002191.3 | NP_002182.1 | 1 - 2 | 414 | 64 |
GenBank mRNA | BC006391.2 | - | 1 - 2 | 351 | 64 |
BT006954.1 | - | 1 - 2 | 271 | 64 | |
M13144.1 | - | 1 - 2 | 228 | 64 | |
M13981.1 | - | 1 - 2 | 415 | 64 |