The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005522.4 | NP_005513.1 | 1 - 2 | 747 | 80 |
NM_153620.2 | NP_705873.2 | 2 - 3 | 544 | 80 | |
GenBank mRNA | AK308067.1 | - | 2 - 3 | 492 | 80 |
AK313514.1 | - | 1 - 2 | 738 | 80 | |
BC032547.2 | - | 1 - 2 | 715 | 80 | |
S79871.1 | - | 2 - 3 | 444 | 80 | |
S79910.1 | - | 1 - 2 | 647 | 80 | |
U10421.1 | - | 1 - 2 | 653 | 80 |