Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81899164-81899660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC11
Entrez Gene ID
51529
Gene Name
anaphase promoting complex subunit 11
Gene Aliases
APC11, Apc11p, HSPC214
UniGene
Hs.534456
Gene Chromosome Location
Chr.17: 81890790-81900533 on Build GRCh38
Transcripts
NM_001002244.2 NM_001002245.2 NM_001002246.2 NM_001002247.2 NM_001002248.2 NM_001002249.2 NM_001289414.1 NM_001289415.1 NM_001289416.1 NM_001289417.1 NM_001289418.1 NM_001289419.1 NM_001289420.1 NM_016476.11

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6379736 c.175G>A p.G59R Chr.17: 81899307-81899307 on Build GRCh38
3370968 c.204C>T p.I68I Chr.17: 81899336-81899336 on Build GRCh38
2805661 c.293C>T p.A98V Chr.17: 81899425-81899425 on Build GRCh38
5972516 c.238G>C p.G80R Chr.17: 81899370-81899370 on Build GRCh38
2805660 c.283A>G p.T95A Chr.17: 81899415-81899415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039805780 Chr.17: 81899388-81899388 on Build GRCh38
rs376590482 Chr.17: 81899304-81899304 on Build GRCh38
rs752176567 Chr.17: 81899447-81899447 on Build GRCh38
rs752189744 Chr.17: 81899506-81899506 on Build GRCh38
rs967205113 Chr.17: 81899299-81899299 on Build GRCh38
rs773005506 Chr.17: 81899235-81899235 on Build GRCh38
rs776800690 Chr.17: 81899266-81899266 on Build GRCh38
rs754466058 Chr.17: 81899412-81899412 on Build GRCh38
rs997784302 Chr.17: 81899421-81899421 on Build GRCh38
rs552502166 Chr.17: 81899395-81899395 on Build GRCh38
rs779862510 Chr.17: 81899535-81899535 on Build GRCh38
rs192673888 Chr.17: 81899528-81899528 on Build GRCh38
rs781172834 Chr.17: 81899527-81899527 on Build GRCh38
rs199541249 Chr.17: 81899461-81899461 on Build GRCh38
rs373711038 Chr.17: 81899190-81899190 on Build GRCh38
rs755752455 Chr.17: 81899587-81899588 on Build GRCh38
rs973909052 Chr.17: 81899217-81899217 on Build GRCh38
rs142854740 Chr.17: 81899263-81899263 on Build GRCh38
rs141853377 Chr.17: 81899354-81899354 on Build GRCh38
rs751313643 Chr.17: 81899516-81899516 on Build GRCh38
rs202012453 Chr.17: 81899560-81899560 on Build GRCh38
rs113423282 Chr.17: 81899585-81899585 on Build GRCh38
rs997879105 Chr.17: 81899280-81899280 on Build GRCh38
rs189044396 Chr.17: 81899491-81899491 on Build GRCh38
rs74006126 Chr.17: 81899579-81899579 on Build GRCh38
rs763194577 Chr.17: 81899382-81899382 on Build GRCh38
rs962512335 Chr.17: 81899214-81899214 on Build GRCh38
rs150198509 Chr.17: 81899325-81899325 on Build GRCh38
rs749341789 Chr.17: 81899468-81899468 on Build GRCh38
rs918477932 Chr.17: 81899233-81899233 on Build GRCh38
rs774505492 Chr.17: 81899474-81899474 on Build GRCh38
rs909329491 Chr.17: 81899204-81899204 on Build GRCh38
rs768857475 Chr.17: 81899473-81899473 on Build GRCh38
rs750990094 Chr.17: 81899399-81899399 on Build GRCh38
rs777301557 Chr.17: 81899453-81899453 on Build GRCh38
rs1036062336 Chr.17: 81899459-81899459 on Build GRCh38
rs771963363 Chr.17: 81899220-81899220 on Build GRCh38
rs763796694 Chr.17: 81899512-81899512 on Build GRCh38
rs765487120 Chr.17: 81899288-81899288 on Build GRCh38
rs763214101 Chr.17: 81899300-81899300 on Build GRCh38
rs781674317 Chr.17: 81899315-81899315 on Build GRCh38
rs746749274 Chr.17: 81899456-81899456 on Build GRCh38
rs756021661 Chr.17: 81899531-81899531 on Build GRCh38
rs778526122 Chr.17: 81899332-81899332 on Build GRCh38
rs550213920 Chr.17: 81899229-81899230 on Build GRCh38
rs769963517 Chr.17: 81899464-81899464 on Build GRCh38
rs771737681 Chr.17: 81899342-81899342 on Build GRCh38
rs201159530 Chr.17: 81899452-81899452 on Build GRCh38
rs538053280 Chr.17: 81899467-81899467 on Build GRCh38
rs991593683 Chr.17: 81899198-81899198 on Build GRCh38
rs1026407388 Chr.17: 81899572-81899572 on Build GRCh38
rs576296396 Chr.17: 81899606-81899606 on Build GRCh38
rs376416519 Chr.17: 81899210-81899210 on Build GRCh38
rs746799218 Chr.17: 81899348-81899348 on Build GRCh38
rs753279624 Chr.17: 81899321-81899321 on Build GRCh38
rs985397758 Chr.17: 81899607-81899607 on Build GRCh38
rs759683269 Chr.17: 81899369-81899369 on Build GRCh38
rs142049212 Chr.17: 81899353-81899353 on Build GRCh38
rs146931437 Chr.17: 81899511-81899511 on Build GRCh38
rs753029345 Chr.17: 81899291-81899291 on Build GRCh38
rs74006125 Chr.17: 81899261-81899261 on Build GRCh38
rs61751632 Chr.17: 81899345-81899345 on Build GRCh38
rs981410809 Chr.17: 81899528-81899529 on Build GRCh38
rs766813330 Chr.17: 81899497-81899497 on Build GRCh38
rs778529238 Chr.17: 81899429-81899429 on Build GRCh38
rs539001186 Chr.17: 81899571-81899571 on Build GRCh38
rs942367381 Chr.17: 81899594-81899594 on Build GRCh38
rs761061419 Chr.17: 81899361-81899361 on Build GRCh38
rs371131210 Chr.17: 81899381-81899381 on Build GRCh38
rs773681686 Chr.17: 81899488-81899488 on Build GRCh38
rs747894485 Chr.17: 81899219-81899219 on Build GRCh38
rs151096381 Chr.17: 81899311-81899311 on Build GRCh38
rs751900854 Chr.17: 81899306-81899306 on Build GRCh38
rs573674995 Chr.17: 81899493-81899493 on Build GRCh38
rs367604570 Chr.17: 81899569-81899569 on Build GRCh38
rs770056495 Chr.17: 81899370-81899370 on Build GRCh38
rs773999857 Chr.17: 81899208-81899208 on Build GRCh38
rs757095904 Chr.17: 81899524-81899524 on Build GRCh38
rs543571216 Chr.17: 81899287-81899287 on Build GRCh38
rs973846974 Chr.17: 81899502-81899502 on Build GRCh38
rs767922019 Chr.17: 81899481-81899481 on Build GRCh38
rs553577795 Chr.17: 81899555-81899555 on Build GRCh38
rs374400220 Chr.17: 81899392-81899392 on Build GRCh38
rs768678101 Chr.17: 81899550-81899550 on Build GRCh38
rs957548917 Chr.17: 81899484-81899484 on Build GRCh38
rs370976498 Chr.17: 81899462-81899462 on Build GRCh38
rs764245237 Chr.17: 81899386-81899386 on Build GRCh38
rs749383774 Chr.17: 81899538-81899538 on Build GRCh38
rs762154505 Chr.17: 81899476-81899476 on Build GRCh38
rs761037944 Chr.17: 81899577-81899577 on Build GRCh38
rs747887439 Chr.17: 81899338-81899338 on Build GRCh38

View Full Product Details