Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RXRA
Entrez Gene ID
6256
Gene Name
retinoid X receptor alpha
Gene Aliases
NR2B1
Chromosome Location
Chr.9: 134326463 - 134440586 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291920.1 NP_001278849.1 5 - 6 1620 74
NM_001291921.1 NP_001278850.1 4 - 5 1411 74
NM_002957.5 NP_002948.1 5 - 6 948 74
GenBank mRNA AB307705.1 - 5 - 6 779 74
AK131081.1 - 1 - 2 65 74
AK131192.1 - 4 - 5 1411 74
AK131513.1 - 5 - 6 1620 74
BC063827.1 - 5 - 6 941 74
BC096246.2 - 5 - 6 74
BC110998.1 - 5 - 6 910 74
HQ692843.1 - 5 - 6 779 74
X52773.1 - 5 - 6 854 74

View Full Product Details