The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005962.4 | NP_005953.4 | 2 - 3 | 414 | 83 |
NM_130439.3 | NP_569157.2 | 2 - 3 | 612 | 83 | |
GenBank mRNA | AK307090.1 | - | 2 - 3 | 153 | 83 |
AK309718.1 | - | 2 - 3 | 227 | 83 | |
BC035128.1 | - | 2 - 3 | 618 | 83 | |
BG700036.1 | - | 2 - 3 | 398 | 83 | |
BM927870.1 | - | 2 - 3 | 397 | 83 | |
BX417051.2 | - | 2 - 3 | 410 | 83 | |
CF594363.1 | - | - | 446 | 83 | |
CR536576.1 | - | 2 - 3 | 207 | 83 | |
D63940.1 | - | 1 - 2 | 99 | 83 | |
KU178208.1 | - | 1 - 2 | 99 | 83 | |
KU178209.1 | - | 1 - 2 | 99 | 83 |