Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 6870223-6870762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP28
Entrez Gene ID
79822
Gene Name
Rho GTPase activating protein 28
Gene Aliases
-
UniGene
Hs.183114
Gene Chromosome Location
Chr.18: 6729718-6915716 on Build GRCh38
Transcripts
NM_001010000.2 XM_005258144.1 XM_005258145.1 XM_005258146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6482974 c.443A>T p.K148I Chr.18: 6870698-6870698 on Build GRCh38
7356205 c.408G>C p.M136I Chr.18: 6870663-6870663 on Build GRCh38
4662833 c.949T>G p.L317V Chr.18: 6870727-6870727 on Build GRCh38
6422186 c.383C>T p.S128L Chr.18: 6870638-6870638 on Build GRCh38
3821701 c.446C>T p.S149L Chr.18: 6870701-6870701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867413651 Chr.18: 6870656-6870656 on Build GRCh38
rs145725626 Chr.18: 6870672-6870672 on Build GRCh38
rs991526826 Chr.18: 6870380-6870380 on Build GRCh38
rs573320683 Chr.18: 6870484-6870484 on Build GRCh38
rs1005316407 Chr.18: 6870497-6870497 on Build GRCh38
rs932344098 Chr.18: 6870479-6870479 on Build GRCh38
rs140498442 Chr.18: 6870669-6870669 on Build GRCh38
rs151056858 Chr.18: 6870720-6870720 on Build GRCh38
rs767768875 Chr.18: 6870331-6870331 on Build GRCh38
rs766408917 Chr.18: 6870657-6870657 on Build GRCh38
rs181017908 Chr.18: 6870251-6870251 on Build GRCh38
rs752754418 Chr.18: 6870334-6870334 on Build GRCh38
rs1030747037 Chr.18: 6870441-6870441 on Build GRCh38
rs754260607 Chr.18: 6870543-6870543 on Build GRCh38
rs755478637 Chr.18: 6870554-6870554 on Build GRCh38
rs141139106 Chr.18: 6870579-6870579 on Build GRCh38
rs369043297 Chr.18: 6870632-6870632 on Build GRCh38
rs537846222 Chr.18: 6870397-6870397 on Build GRCh38
rs567296702 Chr.18: 6870690-6870690 on Build GRCh38
rs761762410 Chr.18: 6870442-6870442 on Build GRCh38
rs62082833 Chr.18: 6870525-6870525 on Build GRCh38
rs377260177 Chr.18: 6870718-6870718 on Build GRCh38
rs774085134 Chr.18: 6870616-6870616 on Build GRCh38
rs749235830 Chr.18: 6870588-6870588 on Build GRCh38
rs779490362 Chr.18: 6870695-6870695 on Build GRCh38
rs978087719 Chr.18: 6870465-6870465 on Build GRCh38
rs542753830 Chr.18: 6870464-6870464 on Build GRCh38
rs957952036 Chr.18: 6870470-6870470 on Build GRCh38
rs144126596 Chr.18: 6870625-6870625 on Build GRCh38
rs902613941 Chr.18: 6870692-6870692 on Build GRCh38
rs374848022 Chr.18: 6870714-6870714 on Build GRCh38
rs374389761 Chr.18: 6870540-6870540 on Build GRCh38
rs916440323 Chr.18: 6870483-6870483 on Build GRCh38
rs574045659 Chr.18: 6870375-6870375 on Build GRCh38
rs768630189 Chr.18: 6870592-6870592 on Build GRCh38
rs779195127 Chr.18: 6870557-6870557 on Build GRCh38
rs750524531 Chr.18: 6870513-6870513 on Build GRCh38
rs760019406 Chr.18: 6870663-6870663 on Build GRCh38
rs374298701 Chr.18: 6870670-6870670 on Build GRCh38
rs528222972 Chr.18: 6870491-6870491 on Build GRCh38
rs947226743 Chr.18: 6870429-6870429 on Build GRCh38
rs900573870 Chr.18: 6870392-6870392 on Build GRCh38
rs374485208 Chr.18: 6870457-6870457 on Build GRCh38
rs772436955 Chr.18: 6870648-6870648 on Build GRCh38
rs773520808 Chr.18: 6870652-6870652 on Build GRCh38
rs754114978 Chr.18: 6870362-6870365 on Build GRCh38
rs991477712 Chr.18: 6870622-6870622 on Build GRCh38
rs113399281 Chr.18: 6870468-6870468 on Build GRCh38
rs559429150 Chr.18: 6870490-6870490 on Build GRCh38
rs372462113 Chr.18: 6870654-6870654 on Build GRCh38
rs929620858 Chr.18: 6870552-6870552 on Build GRCh38
rs369914308 Chr.18: 6870573-6870573 on Build GRCh38
rs1241988 Chr.18: 6870586-6870586 on Build GRCh38
rs888032314 Chr.18: 6870332-6870332 on Build GRCh38
rs377267124 Chr.18: 6870564-6870564 on Build GRCh38
rs113019683 Chr.18: 6870364-6870367 on Build GRCh38
rs761025338 Chr.18: 6870721-6870721 on Build GRCh38
rs753930092 Chr.18: 6870662-6870662 on Build GRCh38
rs934533209 Chr.18: 6870452-6870452 on Build GRCh38
rs756966879 Chr.18: 6870568-6870568 on Build GRCh38
rs747979354 Chr.18: 6870719-6870719 on Build GRCh38
rs997636144 Chr.18: 6870428-6870428 on Build GRCh38
rs929541168 Chr.18: 6870267-6870267 on Build GRCh38
rs146800572 Chr.18: 6870642-6870642 on Build GRCh38
rs896856960 Chr.18: 6870262-6870262 on Build GRCh38
rs750034646 Chr.18: 6870682-6870682 on Build GRCh38
rs990639175 Chr.18: 6870471-6870471 on Build GRCh38
rs1047933438 Chr.18: 6870282-6870282 on Build GRCh38
rs765768815 Chr.18: 6870668-6870668 on Build GRCh38
rs371159621 Chr.18: 6870737-6870737 on Build GRCh38
rs756415928 Chr.18: 6870466-6870466 on Build GRCh38

View Full Product Details