Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9182302-9182817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
UniGene
Hs.464585
Gene Chromosome Location
Chr.18: 9136753-9285985 on Build GRCh38
Transcripts
NM_001083625.2 NM_001204056.1 NM_015208.4 XM_005258092.3 XM_005258093.3 XM_005258095.2 XM_005258096.2 XM_011525638.2 XM_011525639.2 XM_017025661.1 XM_017025662.1 XM_017025663.1 XM_017025664.1 XM_017025665.1 XM_017025666.1 XM_017025667.1 XM_017025668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5555799 c.15G>T p.G5G Chr.18: 9182447-9182447 on Build GRCh38
3147964 c.33G>A p.Q11Q Chr.18: 9182465-9182465 on Build GRCh38
7418791 c.44C>A p.S15Y Chr.18: 9182476-9182476 on Build GRCh38
352711 c.33G>T p.Q11H Chr.18: 9182465-9182465 on Build GRCh38
4936822 c.61A>G p.M21V Chr.18: 9182493-9182493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563193485 Chr.18: 9182633-9182633 on Build GRCh38
rs954037931 Chr.18: 9182657-9182657 on Build GRCh38
rs775617692 Chr.18: 9182436-9182436 on Build GRCh38
rs373440163 Chr.18: 9182647-9182647 on Build GRCh38
rs895068862 Chr.18: 9182414-9182414 on Build GRCh38
rs747450028 Chr.18: 9182559-9182559 on Build GRCh38
rs112983081 Chr.18: 9182622-9182622 on Build GRCh38
rs747537965 Chr.18: 9182392-9182392 on Build GRCh38
rs373772396 Chr.18: 9182409-9182409 on Build GRCh38
rs114129498 Chr.18: 9182447-9182447 on Build GRCh38
rs1039954650 Chr.18: 9182606-9182606 on Build GRCh38
rs753374916 Chr.18: 9182529-9182529 on Build GRCh38
rs767324591 Chr.18: 9182460-9182460 on Build GRCh38
rs1017585334 Chr.18: 9182711-9182711 on Build GRCh38
rs949261537 Chr.18: 9182680-9182680 on Build GRCh38
rs879379827 Chr.18: 9182577-9182580 on Build GRCh38
rs201026074 Chr.18: 9182332-9182332 on Build GRCh38
rs771460140 Chr.18: 9182561-9182561 on Build GRCh38
rs1046620584 Chr.18: 9182683-9182683 on Build GRCh38
rs985704069 Chr.18: 9182674-9182674 on Build GRCh38
rs376731033 Chr.18: 9182411-9182411 on Build GRCh38
rs545646505 Chr.18: 9182773-9182773 on Build GRCh38
rs770039664 Chr.18: 9182426-9182426 on Build GRCh38
rs376717827 Chr.18: 9182418-9182418 on Build GRCh38
rs1044029733 Chr.18: 9182482-9182482 on Build GRCh38
rs781603040 Chr.18: 9182564-9182564 on Build GRCh38
rs539263592 Chr.18: 9182337-9182337 on Build GRCh38
rs905110737 Chr.18: 9182684-9182684 on Build GRCh38
rs1031767831 Chr.18: 9182611-9182611 on Build GRCh38
rs747742303 Chr.18: 9182619-9182619 on Build GRCh38
rs138905967 Chr.18: 9182599-9182599 on Build GRCh38
rs149819427 Chr.18: 9182331-9182332 on Build GRCh38
rs1020398056 Chr.18: 9182788-9182788 on Build GRCh38
rs557800615 Chr.18: 9182357-9182357 on Build GRCh38
rs750304913 Chr.18: 9182490-9182490 on Build GRCh38
rs760610825 Chr.18: 9182515-9182515 on Build GRCh38
rs59801599 Chr.18: 9182333-9182334 on Build GRCh38
rs376838528 Chr.18: 9182365-9182365 on Build GRCh38
rs1034724085 Chr.18: 9182329-9182329 on Build GRCh38
rs112685892 Chr.18: 9182334-9182334 on Build GRCh38
rs758784044 Chr.18: 9182550-9182550 on Build GRCh38
rs745931862 Chr.18: 9182567-9182567 on Build GRCh38
rs768605911 Chr.18: 9182541-9182541 on Build GRCh38
rs947993702 Chr.18: 9182472-9182472 on Build GRCh38
rs746173657 Chr.18: 9182424-9182424 on Build GRCh38
rs772603944 Chr.18: 9182387-9182387 on Build GRCh38
rs778292198 Chr.18: 9182553-9182553 on Build GRCh38
rs369978995 Chr.18: 9182646-9182646 on Build GRCh38
rs754495565 Chr.18: 9182530-9182530 on Build GRCh38
rs778382080 Chr.18: 9182388-9182388 on Build GRCh38
rs779243884 Chr.18: 9182403-9182405 on Build GRCh38
rs60039834 Chr.18: 9182334-9182335 on Build GRCh38
rs73392371 Chr.18: 9182364-9182364 on Build GRCh38
rs796083575 Chr.18: 9182777-9182779 on Build GRCh38
rs973283627 Chr.18: 9182743-9182743 on Build GRCh38
rs199642070 Chr.18: 9182333-9182333 on Build GRCh38
rs962929962 Chr.18: 9182712-9182712 on Build GRCh38
rs188663237 Chr.18: 9182598-9182598 on Build GRCh38
rs1011563403 Chr.18: 9182780-9182780 on Build GRCh38
rs377025467 Chr.18: 9182330-9182330 on Build GRCh38
rs766294727 Chr.18: 9182522-9182522 on Build GRCh38
rs759596305 Chr.18: 9182422-9182422 on Build GRCh38

View Full Product Details