Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9195550-9196053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
UniGene
Hs.464585
Gene Chromosome Location
Chr.18: 9136753-9285985 on Build GRCh38
Transcripts
NM_001083625.2 NM_001204056.1 NM_015208.4 XM_005258092.3 XM_005258093.3 XM_005258095.2 XM_005258096.2 XM_011525638.2 XM_011525639.2 XM_017025661.1 XM_017025662.1 XM_017025663.1 XM_017025664.1 XM_017025665.1 XM_017025666.1 XM_017025667.1 XM_017025668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246820 c.153G>C p.E51D Chr.18: 9195616-9195616 on Build GRCh38
3959444 c.153G>T p.E51D Chr.18: 9195616-9195616 on Build GRCh38
1245005 c.133C>T p.R45* Chr.18: 9195596-9195596 on Build GRCh38
438422 c.230A>G p.D77G Chr.18: 9195693-9195693 on Build GRCh38
1245006 c.187C>T p.P63S Chr.18: 9195650-9195650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964688077 Chr.18: 9195891-9195891 on Build GRCh38
rs776581917 Chr.18: 9195642-9195642 on Build GRCh38
rs941443815 Chr.18: 9195855-9195855 on Build GRCh38
rs879318333 Chr.18: 9195659-9195659 on Build GRCh38
rs780114226 Chr.18: 9195597-9195597 on Build GRCh38
rs767072042 Chr.18: 9195718-9195718 on Build GRCh38
rs1046376870 Chr.18: 9195627-9195627 on Build GRCh38
rs897326960 Chr.18: 9195890-9195890 on Build GRCh38
rs775131460 Chr.18: 9195665-9195665 on Build GRCh38
rs746386764 Chr.18: 9195590-9195590 on Build GRCh38
rs780597549 Chr.18: 9195720-9195720 on Build GRCh38
rs1023249627 Chr.18: 9195726-9195726 on Build GRCh38
rs138574671 Chr.18: 9195609-9195609 on Build GRCh38
rs923142158 Chr.18: 9195783-9195783 on Build GRCh38
rs757607440 Chr.18: 9195578-9195578 on Build GRCh38
rs1027508263 Chr.18: 9195922-9195922 on Build GRCh38
rs34208641 Chr.18: 9195600-9195600 on Build GRCh38
rs1008601392 Chr.18: 9195869-9195869 on Build GRCh38
rs902187468 Chr.18: 9195858-9195858 on Build GRCh38
rs906822480 Chr.18: 9195658-9195658 on Build GRCh38
rs967799525 Chr.18: 9195740-9195740 on Build GRCh38
rs893935305 Chr.18: 9195750-9195750 on Build GRCh38
rs759402316 Chr.18: 9195650-9195650 on Build GRCh38
rs557821112 Chr.18: 9195927-9195927 on Build GRCh38
rs375705287 Chr.18: 9195684-9195684 on Build GRCh38
rs751953590 Chr.18: 9195577-9195577 on Build GRCh38
rs755088847 Chr.18: 9195733-9195733 on Build GRCh38
rs751291055 Chr.18: 9195679-9195679 on Build GRCh38
rs960403570 Chr.18: 9195717-9195717 on Build GRCh38
rs776290792 Chr.18: 9195745-9195745 on Build GRCh38
rs113325027 Chr.18: 9195853-9195853 on Build GRCh38
rs182180534 Chr.18: 9195976-9195976 on Build GRCh38
rs770660759 Chr.18: 9195634-9195634 on Build GRCh38
rs763810808 Chr.18: 9195672-9195672 on Build GRCh38
rs866906032 Chr.18: 9195650-9195651 on Build GRCh38
rs750876573 Chr.18: 9195706-9195706 on Build GRCh38
rs770698045 Chr.18: 9195744-9195744 on Build GRCh38
rs571786906 Chr.18: 9195886-9195886 on Build GRCh38
rs779067463 Chr.18: 9195739-9195739 on Build GRCh38
rs956717390 Chr.18: 9195767-9195767 on Build GRCh38
rs976033864 Chr.18: 9195774-9195774 on Build GRCh38
rs908669045 Chr.18: 9196009-9196009 on Build GRCh38
rs185371592 Chr.18: 9195594-9195594 on Build GRCh38
rs767788199 Chr.18: 9195705-9195705 on Build GRCh38
rs923817596 Chr.18: 9195763-9195763 on Build GRCh38
rs975037119 Chr.18: 9195908-9195908 on Build GRCh38
rs772286903 Chr.18: 9195628-9195630 on Build GRCh38
rs371617857 Chr.18: 9195625-9195625 on Build GRCh38
rs911214059 Chr.18: 9195854-9195854 on Build GRCh38
rs542499477 Chr.18: 9195722-9195722 on Build GRCh38
rs951934425 Chr.18: 9195961-9195961 on Build GRCh38
rs1012439122 Chr.18: 9196000-9196000 on Build GRCh38
rs764769223 Chr.18: 9195656-9195656 on Build GRCh38
rs781696862 Chr.18: 9195579-9195579 on Build GRCh38
rs746788208 Chr.18: 9195617-9195617 on Build GRCh38
rs908554694 Chr.18: 9195592-9195592 on Build GRCh38
rs146036312 Chr.18: 9195602-9195602 on Build GRCh38
rs931886080 Chr.18: 9195841-9195841 on Build GRCh38
rs189976015 Chr.18: 9195836-9195836 on Build GRCh38
rs896764494 Chr.18: 9195968-9195968 on Build GRCh38
rs1010678634 Chr.18: 9195760-9195760 on Build GRCh38
rs940108548 Chr.18: 9196022-9196022 on Build GRCh38
rs538893084 Chr.18: 9195888-9195888 on Build GRCh38

View Full Product Details