Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10468385-10468895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4 XM_011525603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148281 c.192G>A p.R64R Chr.18: 10468602-10468602 on Build GRCh38
4945625 c.235T>C p.S79P Chr.18: 10468645-10468645 on Build GRCh38
6082415 c.121A>C p.K41Q Chr.18: 10468531-10468531 on Build GRCh38
4921054 c.81C>T p.L27L Chr.18: 10468491-10468491 on Build GRCh38
1245180 c.79C>G p.L27V Chr.18: 10468489-10468489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375027558 Chr.18: 10468573-10468573 on Build GRCh38
rs747007917 Chr.18: 10468665-10468665 on Build GRCh38
rs753501769 Chr.18: 10468602-10468602 on Build GRCh38
rs989802858 Chr.18: 10468752-10468752 on Build GRCh38
rs777419740 Chr.18: 10468769-10468769 on Build GRCh38
rs982512077 Chr.18: 10468566-10468566 on Build GRCh38
rs937599530 Chr.18: 10468629-10468629 on Build GRCh38
rs779788793 Chr.18: 10468478-10468478 on Build GRCh38
rs35079028 Chr.18: 10468599-10468599 on Build GRCh38
rs747716206 Chr.18: 10468680-10468680 on Build GRCh38
rs117742712 Chr.18: 10468699-10468699 on Build GRCh38
rs996786485 Chr.18: 10468740-10468740 on Build GRCh38
rs763261435 Chr.18: 10468500-10468500 on Build GRCh38
rs372155709 Chr.18: 10468648-10468648 on Build GRCh38
rs764070191 Chr.18: 10468428-10468428 on Build GRCh38
rs763949577 Chr.18: 10468568-10468568 on Build GRCh38
rs918189905 Chr.18: 10468434-10468434 on Build GRCh38
rs556931254 Chr.18: 10468552-10468552 on Build GRCh38
rs1026832366 Chr.18: 10468744-10468744 on Build GRCh38
rs533674242 Chr.18: 10468855-10468855 on Build GRCh38
rs912389918 Chr.18: 10468864-10468864 on Build GRCh38
rs138626055 Chr.18: 10468520-10468520 on Build GRCh38
rs768987522 Chr.18: 10468489-10468489 on Build GRCh38
rs1056415673 Chr.18: 10468630-10468630 on Build GRCh38
rs76695028 Chr.18: 10468871-10468871 on Build GRCh38
rs540295256 Chr.18: 10468719-10468719 on Build GRCh38
rs776614172 Chr.18: 10468675-10468675 on Build GRCh38
rs887170930 Chr.18: 10468683-10468683 on Build GRCh38
rs1008201130 Chr.18: 10468442-10468442 on Build GRCh38
rs149564774 Chr.18: 10468530-10468530 on Build GRCh38
rs760810531 Chr.18: 10468536-10468536 on Build GRCh38
rs755673638 Chr.18: 10468592-10468592 on Build GRCh38
rs750005652 Chr.18: 10468469-10468469 on Build GRCh38
rs766634852 Chr.18: 10468418-10468418 on Build GRCh38
rs200114605 Chr.18: 10468660-10468660 on Build GRCh38
rs772865987 Chr.18: 10468623-10468623 on Build GRCh38
rs372105770 Chr.18: 10468580-10468580 on Build GRCh38
rs539412 Chr.18: 10468456-10468456 on Build GRCh38
rs748654713 Chr.18: 10468523-10468523 on Build GRCh38
rs771018610 Chr.18: 10468669-10468669 on Build GRCh38
rs770333212 Chr.18: 10468433-10468433 on Build GRCh38
rs1046045873 Chr.18: 10468615-10468615 on Build GRCh38
rs73388589 Chr.18: 10468682-10468682 on Build GRCh38
rs749861468 Chr.18: 10468589-10468589 on Build GRCh38
rs751520081 Chr.18: 10468444-10468444 on Build GRCh38
rs748528385 Chr.18: 10468634-10468634 on Build GRCh38
rs765909861 Chr.18: 10468848-10468848 on Build GRCh38
rs771016289 Chr.18: 10468546-10468546 on Build GRCh38
rs945759916 Chr.18: 10468817-10468817 on Build GRCh38
rs375478769 Chr.18: 10468701-10468701 on Build GRCh38
rs780843389 Chr.18: 10468465-10468465 on Build GRCh38
rs762834923 Chr.18: 10468425-10468425 on Build GRCh38
rs767277027 Chr.18: 10468731-10468732 on Build GRCh38
rs367556237 Chr.18: 10468578-10468578 on Build GRCh38
rs945875943 Chr.18: 10468545-10468545 on Build GRCh38
rs368936130 Chr.18: 10468424-10468424 on Build GRCh38
rs942631161 Chr.18: 10468865-10468865 on Build GRCh38
rs544205809 Chr.18: 10468778-10468778 on Build GRCh38
rs372550648 Chr.18: 10468697-10468697 on Build GRCh38
rs778373480 Chr.18: 10468659-10468659 on Build GRCh38
rs746098930 Chr.18: 10468679-10468679 on Build GRCh38
rs140181954 Chr.18: 10468750-10468750 on Build GRCh38
rs749566230 Chr.18: 10468479-10468479 on Build GRCh38
rs147889681 Chr.18: 10468720-10468720 on Build GRCh38
rs371495185 Chr.18: 10468790-10468790 on Build GRCh38
rs529185489 Chr.18: 10468470-10468470 on Build GRCh38
rs774229764 Chr.18: 10468686-10468686 on Build GRCh38
rs764386254 Chr.18: 10468560-10468560 on Build GRCh38
rs963977619 Chr.18: 10468475-10468475 on Build GRCh38
rs755657503 Chr.18: 10468473-10468473 on Build GRCh38
rs779237976 Chr.18: 10468627-10468627 on Build GRCh38
rs982564545 Chr.18: 10468768-10468768 on Build GRCh38
rs114256170 Chr.18: 10468638-10468638 on Build GRCh38
rs114418217 Chr.18: 10468872-10468872 on Build GRCh38
rs754697340 Chr.18: 10468605-10468605 on Build GRCh38
rs969646990 Chr.18: 10468718-10468718 on Build GRCh38
rs970500163 Chr.18: 10468783-10468783 on Build GRCh38
rs376806313 Chr.18: 10468738-10468738 on Build GRCh38

View Full Product Details