Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10487455-10487949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4071253 c.1185C>A p.S395S Chr.18: 10487678-10487678 on Build GRCh38
4071252 c.1148A>G p.N383S Chr.18: 10487641-10487641 on Build GRCh38
708459 c.1317C>T p.N439N Chr.18: 10487810-10487810 on Build GRCh38
708458 c.1429G>T p.A477S Chr.18: 10487922-10487922 on Build GRCh38
7242121 c.1282G>A p.E428K Chr.18: 10487775-10487775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781563529 Chr.18: 10487909-10487909 on Build GRCh38
rs527431894 Chr.18: 10487863-10487863 on Build GRCh38
rs772842810 Chr.18: 10487774-10487774 on Build GRCh38
rs759687921 Chr.18: 10487677-10487677 on Build GRCh38
rs142674398 Chr.18: 10487903-10487903 on Build GRCh38
rs778685004 Chr.18: 10487651-10487651 on Build GRCh38
rs116485256 Chr.18: 10487609-10487609 on Build GRCh38
rs139112764 Chr.18: 10487723-10487723 on Build GRCh38
rs372294613 Chr.18: 10487642-10487642 on Build GRCh38
rs143654273 Chr.18: 10487548-10487548 on Build GRCh38
rs772612325 Chr.18: 10487546-10487546 on Build GRCh38
rs756921124 Chr.18: 10487907-10487907 on Build GRCh38
rs562518496 Chr.18: 10487788-10487788 on Build GRCh38
rs374253239 Chr.18: 10487794-10487794 on Build GRCh38
rs763105125 Chr.18: 10487760-10487760 on Build GRCh38
rs888645053 Chr.18: 10487523-10487523 on Build GRCh38
rs374423135 Chr.18: 10487828-10487828 on Build GRCh38
rs773153017 Chr.18: 10487597-10487597 on Build GRCh38
rs368201557 Chr.18: 10487853-10487853 on Build GRCh38
rs199677361 Chr.18: 10487750-10487750 on Build GRCh38
rs757964573 Chr.18: 10487897-10487897 on Build GRCh38
rs551321118 Chr.18: 10487822-10487822 on Build GRCh38
rs750246771 Chr.18: 10487708-10487708 on Build GRCh38
rs368882967 Chr.18: 10487700-10487700 on Build GRCh38
rs578125813 Chr.18: 10487565-10487565 on Build GRCh38
rs373303747 Chr.18: 10487792-10487792 on Build GRCh38
rs148807402 Chr.18: 10487620-10487620 on Build GRCh38
rs780062599 Chr.18: 10487721-10487721 on Build GRCh38
rs778212807 Chr.18: 10487741-10487741 on Build GRCh38
rs116143459 Chr.18: 10487787-10487787 on Build GRCh38
rs748836130 Chr.18: 10487627-10487627 on Build GRCh38
rs779375002 Chr.18: 10487626-10487626 on Build GRCh38
rs374248487 Chr.18: 10487611-10487611 on Build GRCh38
rs758081145 Chr.18: 10487793-10487793 on Build GRCh38
rs765354197 Chr.18: 10487684-10487684 on Build GRCh38
rs773767949 Chr.18: 10487658-10487658 on Build GRCh38
rs148965715 Chr.18: 10487529-10487529 on Build GRCh38
rs746237007 Chr.18: 10487916-10487916 on Build GRCh38
rs567615098 Chr.18: 10487547-10487547 on Build GRCh38
rs766352109 Chr.18: 10487552-10487552 on Build GRCh38
rs766126994 Chr.18: 10487784-10487784 on Build GRCh38
rs560171977 Chr.18: 10487846-10487846 on Build GRCh38
rs766181159 Chr.18: 10487855-10487855 on Build GRCh38
rs749401498 Chr.18: 10487830-10487830 on Build GRCh38
rs751310085 Chr.18: 10487599-10487599 on Build GRCh38
rs148910277 Chr.18: 10487902-10487902 on Build GRCh38
rs762290378 Chr.18: 10487539-10487539 on Build GRCh38
rs556439300 Chr.18: 10487549-10487549 on Build GRCh38
rs372175332 Chr.18: 10487703-10487703 on Build GRCh38
rs776528613 Chr.18: 10487669-10487669 on Build GRCh38
rs114154601 Chr.18: 10487752-10487752 on Build GRCh38
rs776301011 Chr.18: 10487856-10487856 on Build GRCh38
rs375861639 Chr.18: 10487652-10487652 on Build GRCh38
rs754478741 Chr.18: 10487724-10487724 on Build GRCh38
rs771861487 Chr.18: 10487739-10487739 on Build GRCh38
rs768144488 Chr.18: 10487641-10487641 on Build GRCh38
rs114473881 Chr.18: 10487621-10487621 on Build GRCh38
rs566310523 Chr.18: 10487508-10487508 on Build GRCh38
rs573503346 Chr.18: 10487511-10487511 on Build GRCh38
rs377642958 Chr.18: 10487770-10487770 on Build GRCh38
rs752888866 Chr.18: 10487688-10487688 on Build GRCh38
rs775482025 Chr.18: 10487829-10487829 on Build GRCh38
rs765531330 Chr.18: 10487590-10487590 on Build GRCh38
rs752200924 Chr.18: 10487871-10487871 on Build GRCh38
rs143975400 Chr.18: 10487736-10487736 on Build GRCh38
rs200082076 Chr.18: 10487825-10487825 on Build GRCh38
rs775062125 Chr.18: 10487753-10487753 on Build GRCh38
rs150728546 Chr.18: 10487663-10487663 on Build GRCh38
rs1012530675 Chr.18: 10487873-10487873 on Build GRCh38
rs374468665 Chr.18: 10487864-10487864 on Build GRCh38
rs759606550 Chr.18: 10487578-10487578 on Build GRCh38
rs529163507 Chr.18: 10487759-10487759 on Build GRCh38
rs376074678 Chr.18: 10487811-10487811 on Build GRCh38
rs533529179 Chr.18: 10487527-10487527 on Build GRCh38
rs145563866 Chr.18: 10487885-10487885 on Build GRCh38
rs755938219 Chr.18: 10487716-10487716 on Build GRCh38
rs892064810 Chr.18: 10487820-10487820 on Build GRCh38
rs1055285662 Chr.18: 10487834-10487834 on Build GRCh38
rs778631558 Chr.18: 10487734-10487734 on Build GRCh38
rs771470436 Chr.18: 10487664-10487664 on Build GRCh38
rs958343302 Chr.18: 10487592-10487592 on Build GRCh38
rs139844939 Chr.18: 10487755-10487755 on Build GRCh38
rs760496693 Chr.18: 10487777-10487777 on Build GRCh38
rs772558905 Chr.18: 10487657-10487657 on Build GRCh38
rs774600867 Chr.18: 10487849-10487849 on Build GRCh38
rs3185480 Chr.18: 10487921-10487921 on Build GRCh38

View Full Product Details