Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GP1BA
Entrez Gene ID
2811
Gene Name
glycoprotein Ib platelet alpha subunit
Gene Aliases
BDPLT1, BDPLT3, BSS, CD42B, CD42b-alpha, DBPLT3, GP1B, GPIbA, GPIbalpha, VWDP
Chromosome Location
Chr.17: 4932275 - 4935030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000173.6 NP_000164.5 1 - 2 96 64
GenBank mRNA BC027955.1 - 1 - 2 74 64
J02940.1 - 1 - 2 41 64
KC120774.1 - 1 - 2 74 64
KC120775.1 - 1 - 2 74 64
KC120776.1 - 1 - 2 74 64

View Full Product Details