Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22413516-22414026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE1
Entrez Gene ID
64693
Gene Name
cutaneous T-cell lymphoma-associated antigen 1
Gene Aliases
CT21.1, CT21.2, CTAGE, CTAGE-1, CTAGE-2
UniGene
Hs.406709
Gene Chromosome Location
Chr.18: 22413601-22417915 on Build GRCh38
Transcripts
NM_172241.2

SNP Detail

SNP ID Chromosome Location
rs894944562 Chr.18: 22413575-22413575 on Build GRCh38
rs775059587 Chr.18: 22413635-22413635 on Build GRCh38
rs1018656007 Chr.18: 22413857-22413857 on Build GRCh38
rs990385083 Chr.18: 22413691-22413691 on Build GRCh38
rs549822305 Chr.18: 22413698-22413698 on Build GRCh38
rs983487380 Chr.18: 22413644-22413644 on Build GRCh38
rs765680057 Chr.18: 22413671-22413671 on Build GRCh38
rs377266326 Chr.18: 22413958-22413958 on Build GRCh38
rs1003677600 Chr.18: 22413540-22413540 on Build GRCh38
rs1052499802 Chr.18: 22413990-22413990 on Build GRCh38
rs1052549922 Chr.18: 22413683-22413683 on Build GRCh38
rs915788821 Chr.18: 22413948-22413948 on Build GRCh38
rs777376824 Chr.18: 22413718-22413718 on Build GRCh38
rs369824941 Chr.18: 22413697-22413697 on Build GRCh38
rs772213203 Chr.18: 22413693-22413693 on Build GRCh38
rs17669793 Chr.18: 22413957-22413957 on Build GRCh38
rs1012076588 Chr.18: 22413871-22413871 on Build GRCh38
rs568546928 Chr.18: 22413946-22413946 on Build GRCh38
rs1015316077 Chr.18: 22413654-22413654 on Build GRCh38
rs920469074 Chr.18: 22413568-22413568 on Build GRCh38
rs1027462197 Chr.18: 22413889-22413889 on Build GRCh38
rs976176275 Chr.18: 22413981-22413981 on Build GRCh38
rs948055016 Chr.18: 22413623-22413623 on Build GRCh38
rs141125292 Chr.18: 22413898-22413898 on Build GRCh38
rs935399085 Chr.18: 22413960-22413960 on Build GRCh38
rs146472718 Chr.18: 22413619-22413619 on Build GRCh38
rs1042071012 Chr.18: 22413717-22413717 on Build GRCh38
rs752232959 Chr.18: 22413749-22413749 on Build GRCh38
rs966434221 Chr.18: 22413978-22413978 on Build GRCh38
rs1027555484 Chr.18: 22413617-22413617 on Build GRCh38
rs9965155 Chr.18: 22413951-22413951 on Build GRCh38
rs932148522 Chr.18: 22413690-22413690 on Build GRCh38
rs895005118 Chr.18: 22413838-22413838 on Build GRCh38
rs953575134 Chr.18: 22413991-22413991 on Build GRCh38
rs925392409 Chr.18: 22413725-22413725 on Build GRCh38
rs576740557 Chr.18: 22413745-22413745 on Build GRCh38
rs113073293 Chr.18: 22413555-22413555 on Build GRCh38
rs369086149 Chr.18: 22413864-22413864 on Build GRCh38
rs930530048 Chr.18: 22413597-22413597 on Build GRCh38
rs1012212341 Chr.18: 22413586-22413586 on Build GRCh38
rs915828461 Chr.18: 22413620-22413620 on Build GRCh38
rs565709785 Chr.18: 22413699-22413699 on Build GRCh38
rs920414942 Chr.18: 22413875-22413875 on Build GRCh38
rs549885956 Chr.18: 22413598-22413598 on Build GRCh38
rs1056225356 Chr.18: 22413757-22413757 on Build GRCh38
rs187294777 Chr.18: 22413638-22413638 on Build GRCh38
rs1046831822 Chr.18: 22413667-22413667 on Build GRCh38
rs961798422 Chr.18: 22413546-22413546 on Build GRCh38
rs760760454 Chr.18: 22413873-22413873 on Build GRCh38
rs757839608 Chr.18: 22413858-22413858 on Build GRCh38
rs191684009 Chr.18: 22413997-22413997 on Build GRCh38
rs555534376 Chr.18: 22413785-22413785 on Build GRCh38
rs1008529796 Chr.18: 22413794-22413794 on Build GRCh38
rs552154596 Chr.18: 22413670-22413670 on Build GRCh38

View Full Product Details