The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000944.4 | NP_000935.1 | 10 - 11 | 1833 | 88 |
NM_001130691.1 | NP_001124163.1 | 10 - 11 | 1833 | 88 | |
NM_001130692.1 | NP_001124164.1 | 9 - 10 | 1707 | 88 | |
XM_017008365.1 | XP_016863854.1 | 10 - 11 | 1137 | 88 | |
GenBank mRNA | AB451338.1 | - | 10 - 11 | 1158 | 88 |
AB451487.1 | - | 10 - 11 | 1158 | 88 | |
AK290532.1 | - | 10 - 11 | 1695 | 88 | |
AL353950.1 | - | 10 - 11 | 1564 | 88 | |
AY904364.1 | - | 9 - 10 | 1003 | 88 | |
BC025714.1 | - | 10 - 11 | 1267 | 88 | |
EU192652.1 | - | 9 - 10 | 1707 | 88 | |
EU192653.1 | - | 4 - 5 | 1150 | 88 | |
J05480.1 | - | 4 - 5 | 354 | 88 | |
KU178269.1 | - | 10 - 11 | 1158 | 88 | |
KU178270.1 | - | 10 - 11 | 1158 | 88 | |
L14778.1 | - | 10 - 11 | 1305 | 88 |