The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024844.1 | NP_001020015.1 | 4 - 5 | 388 | 84 |
NM_002231.3 | NP_002222.1 | 4 - 5 | 388 | 84 | |
XM_006718223.2 | XP_006718286.1 | 5 - 6 | 427 | 84 | |
GenBank mRNA | AK225410.1 | - | 4 - 5 | 314 | 84 |
AK225708.1 | - | 4 - 5 | 342 | 84 | |
AK312677.1 | - | 4 - 5 | 314 | 84 | |
AY303776.1 | - | 2 - 3 | 140 | 84 | |
BC000726.2 | - | 4 - 5 | 286 | 84 | |
BC001821.2 | - | 4 - 5 | 286 | 84 | |
BT019761.1 | - | 2 - 3 | 140 | 84 | |
CB127804.1 | - | 4 - 5 | 404 | 84 | |
CR407650.1 | - | 2 - 3 | 140 | 84 | |
CR542255.1 | - | 2 - 3 | 140 | 84 | |
S48196.1 | - | 4 - 5 | 306 | 84 | |
U20770.1 | - | 4 - 5 | 305 | 84 |