The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128148.2 | NP_001121620.1 | 13 - 14 | 1611 | 79 |
NM_001313965.1 | NP_001300894.1 | 12 - 13 | 1409 | 79 | |
NM_001313966.1 | NP_001300895.1 | 11 - 12 | 1213 | 79 | |
NM_003234.3 | NP_003225.2 | 13 - 14 | 1752 | 79 | |
GenBank mRNA | AB209254.1 | - | 13 - 14 | 1515 | 79 |
AK291723.1 | - | 13 - 14 | 1752 | 79 | |
AK294758.1 | - | 11 - 12 | 1143 | 79 | |
BC001188.1 | - | 13 - 14 | 1525 | 79 | |
BX537966.1 | - | 12 - 13 | 1339 | 79 | |
M11507.1 | - | 13 - 14 | 1564 | 79 | |
X01060.1 | - | 13 - 14 | 1732 | 79 |