Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CORT
Entrez Gene ID
1325
Gene Name
cortistatin
Gene Aliases
CST-14, CST-17, CST-29
Chromosome Location
Chr.1: 10449719 - 10452003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302.4 NP_001293.3 1 - 2 608 135
GenBank mRNA AF013252.1 - 1 - 2 188 135
AF086433.1 - 1 - 2 413 135
AY358561.1 - 1 - 2 608 135
BC040034.1 - 1 - 2 156 135
BC119724.1 - 1 - 2 288 135
BC119725.1 - 1 - 2 288 135

View Full Product Details