The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007117.4 | NP_009048.1 | 1 - 2 | 432 | 76 |
GenBank mRNA | AK313473.1 | - | 1 - 2 | 158 | 76 |
BC110515.2 | - | 1 - 2 | 74 | 76 | |
BC110516.2 | - | 1 - 2 | 74 | 76 | |
DA526260.1 | - | 1 - 2 | 521 | 76 |