Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAPBP
Entrez Gene ID
6892
Gene Name
TAP binding protein (tapasin)
Gene Aliases
NGS17, TAPA, TPN, TPSN
Chromosome Location
Chr.6: 33299694 - 33314387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003190.4 NP_003181.3 7 - 8 1680 65
NM_172209.2 NP_757346.2 6 - 7 1419 65
GenBank mRNA AB010639.1 - 7 - 8 1347 65
AF009510.1 - 7 - 8 1350 65
AF029750.1 - 7 - 8 1372 65
BC064966.1 - 6 - 7 801 65
Y13582.1 - 7 - 8 1463 65

View Full Product Details