Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIM2
Entrez Gene ID
9447
Gene Name
absent in melanoma 2
Gene Aliases
PYHIN4
Chromosome Location
Chr.1: 159059226 - 159132351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004833.1 NP_004824.1 1 - 2 232 97
GenBank mRNA AF024714.1 - 1 - 2 232 97
AK292042.1 - 1 - 2 141 97
BC010940.1 - 1 - 2 234 97

View Full Product Details