Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74139913-74140429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO15
Entrez Gene ID
201456
Gene Name
F-box protein 15
Gene Aliases
FBX15
UniGene
Hs.664011
Gene Chromosome Location
Chr.18: 74073343-74147865 on Build GRCh38
Transcripts
NM_001142958.1 NM_152676.2 XM_011525856.1 XM_011525858.2 XM_011525859.1 XM_011525860.1 XM_017025603.1 XM_017025604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311408 c.170C>A p.A57D Chr.18: 74140259-74140259 on Build GRCh38
5581058 c.134A>G p.K45R Chr.18: 74140295-74140295 on Build GRCh38
1206796 c.208T>C p.C70R Chr.18: 74140221-74140221 on Build GRCh38
1153267 c.168T>C p.H56H Chr.18: 74140261-74140261 on Build GRCh38
438373 c.221T>C p.L74P Chr.18: 74140208-74140208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028883200 Chr.18: 74140342-74140342 on Build GRCh38
rs896617952 Chr.18: 74140249-74140249 on Build GRCh38
rs974649937 Chr.18: 74140363-74140363 on Build GRCh38
rs756878788 Chr.18: 74140173-74140173 on Build GRCh38
rs569903000 Chr.18: 74140325-74140325 on Build GRCh38
rs17088833 Chr.18: 74140379-74140379 on Build GRCh38
rs768907377 Chr.18: 74140169-74140169 on Build GRCh38
rs776482101 Chr.18: 74139940-74139940 on Build GRCh38
rs750173319 Chr.18: 74140257-74140257 on Build GRCh38
rs61741369 Chr.18: 74140213-74140213 on Build GRCh38
rs765884442 Chr.18: 74140258-74140258 on Build GRCh38
rs570464813 Chr.18: 74139949-74139949 on Build GRCh38
rs9949460 Chr.18: 74139956-74139956 on Build GRCh38
rs966703084 Chr.18: 74140280-74140280 on Build GRCh38
rs761318319 Chr.18: 74140013-74140013 on Build GRCh38
rs964540706 Chr.18: 74140078-74140078 on Build GRCh38
rs951820877 Chr.18: 74139955-74139955 on Build GRCh38
rs974996503 Chr.18: 74140100-74140100 on Build GRCh38
rs975954712 Chr.18: 74140030-74140030 on Build GRCh38
rs895316523 Chr.18: 74140199-74140199 on Build GRCh38
rs564100560 Chr.18: 74140300-74140300 on Build GRCh38
rs774424800 Chr.18: 74140177-74140177 on Build GRCh38
rs560411409 Chr.18: 74140370-74140370 on Build GRCh38
rs191633511 Chr.18: 74140130-74140130 on Build GRCh38
rs762160136 Chr.18: 74140187-74140187 on Build GRCh38
rs961093505 Chr.18: 74140003-74140003 on Build GRCh38
rs1002432233 Chr.18: 74140181-74140181 on Build GRCh38
rs965666077 Chr.18: 74140275-74140275 on Build GRCh38
rs573379305 Chr.18: 74140060-74140060 on Build GRCh38
rs780063793 Chr.18: 74140303-74140303 on Build GRCh38
rs898736633 Chr.18: 74140173-74140173 on Build GRCh38
rs752154397 Chr.18: 74140348-74140348 on Build GRCh38
rs750007218 Chr.18: 74140104-74140104 on Build GRCh38
rs183168282 Chr.18: 74140343-74140343 on Build GRCh38
rs753812889 Chr.18: 74140285-74140285 on Build GRCh38
rs939608644 Chr.18: 74140132-74140132 on Build GRCh38
rs112580986 Chr.18: 74140035-74140035 on Build GRCh38
rs1000807329 Chr.18: 74140293-74140293 on Build GRCh38
rs760094454 Chr.18: 74140105-74140105 on Build GRCh38
rs997443666 Chr.18: 74140327-74140327 on Build GRCh38
rs887523309 Chr.18: 74140219-74140219 on Build GRCh38
rs575812028 Chr.18: 74140150-74140150 on Build GRCh38
rs867239429 Chr.18: 74140283-74140283 on Build GRCh38
rs776172296 Chr.18: 74140153-74140153 on Build GRCh38
rs907567475 Chr.18: 74140131-74140131 on Build GRCh38
rs1014596166 Chr.18: 74140260-74140260 on Build GRCh38
rs113568258 Chr.18: 74140004-74140004 on Build GRCh38
rs868327947 Chr.18: 74140071-74140071 on Build GRCh38
rs966671984 Chr.18: 74140070-74140070 on Build GRCh38
rs1012391953 Chr.18: 74140202-74140202 on Build GRCh38
rs142562723 Chr.18: 74140075-74140075 on Build GRCh38
rs372589654 Chr.18: 74140319-74140319 on Build GRCh38
rs766259583 Chr.18: 74140267-74140267 on Build GRCh38

View Full Product Details