Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1043509-1043909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA7
Entrez Gene ID
10347
Gene Name
ATP binding cassette subfamily A member 7
Gene Aliases
ABCA-SSN, ABCX, AD9
UniGene
Hs.134514
Gene Chromosome Location
Chr.19: 1040103-1065572 on Build GRCh38
Transcripts
NM_019112.3 XM_006722616.1 XM_006722617.2 XM_011527628.2 XM_011527629.1 XM_011527630.1 XM_011527631.1 XM_011527632.1 XM_011527633.2 XM_011527634.1 XM_011527635.1 XM_017026142.1 XM_017026143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4415225 c.955A>G p.T319A Chr.19: 1043749-1043749 on Build GRCh38
474104 c.997C>G p.P333A Chr.19: 1043791-1043791 on Build GRCh38
6447965 c.946G>A p.E316K Chr.19: 1043740-1043740 on Build GRCh38
6589846 c.1001G>A p.R334Q Chr.19: 1043795-1043795 on Build GRCh38
300136 c.938G>A p.R313Q Chr.19: 1043732-1043732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557177777 Chr.19: 1043569-1043569 on Build GRCh38
rs775053639 Chr.19: 1043888-1043888 on Build GRCh38
rs892537110 Chr.19: 1043622-1043622 on Build GRCh38
rs3752231 Chr.19: 1043639-1043639 on Build GRCh38
rs767853315 Chr.19: 1043735-1043735 on Build GRCh38
rs762403032 Chr.19: 1043889-1043889 on Build GRCh38
rs780664148 Chr.19: 1043880-1043880 on Build GRCh38
rs746671968 Chr.19: 1043789-1043789 on Build GRCh38
rs768401967 Chr.19: 1043689-1043689 on Build GRCh38
rs758916987 Chr.19: 1043800-1043800 on Build GRCh38
rs149023827 Chr.19: 1043748-1043748 on Build GRCh38
rs540229809 Chr.19: 1043626-1043626 on Build GRCh38
rs766417476 Chr.19: 1043848-1043848 on Build GRCh38
rs749233588 Chr.19: 1043686-1043686 on Build GRCh38
rs3752230 Chr.19: 1043576-1043576 on Build GRCh38
rs148427350 Chr.19: 1043740-1043740 on Build GRCh38
rs3752232 Chr.19: 1043749-1043749 on Build GRCh38
rs1016461161 Chr.19: 1043736-1043736 on Build GRCh38
rs35385365 Chr.19: 1043676-1043677 on Build GRCh38
rs377652801 Chr.19: 1043590-1043590 on Build GRCh38
rs1042997911 Chr.19: 1043596-1043596 on Build GRCh38
rs979256348 Chr.19: 1043717-1043717 on Build GRCh38
rs752619389 Chr.19: 1043866-1043866 on Build GRCh38
rs141983399 Chr.19: 1043767-1043767 on Build GRCh38
rs137894448 Chr.19: 1043690-1043690 on Build GRCh38
rs954965783 Chr.19: 1043614-1043614 on Build GRCh38
rs753224215 Chr.19: 1043551-1043551 on Build GRCh38
rs571699570 Chr.19: 1043830-1043830 on Build GRCh38
rs578025097 Chr.19: 1043529-1043529 on Build GRCh38
rs746693184 Chr.19: 1043879-1043879 on Build GRCh38
rs147846250 Chr.19: 1043795-1043795 on Build GRCh38
rs201091850 Chr.19: 1043754-1043754 on Build GRCh38
rs747889847 Chr.19: 1043692-1043692 on Build GRCh38
rs760193677 Chr.19: 1043700-1043700 on Build GRCh38
rs541638574 Chr.19: 1043712-1043716 on Build GRCh38
rs774929060 Chr.19: 1043814-1043814 on Build GRCh38
rs369738612 Chr.19: 1043751-1043751 on Build GRCh38
rs949345932 Chr.19: 1043839-1043839 on Build GRCh38
rs757652134 Chr.19: 1043731-1043731 on Build GRCh38
rs750738976 Chr.19: 1043844-1043844 on Build GRCh38
rs978625933 Chr.19: 1043805-1043805 on Build GRCh38
rs771743560 Chr.19: 1043695-1043695 on Build GRCh38
rs756204320 Chr.19: 1043746-1043746 on Build GRCh38
rs764418887 Chr.19: 1043712-1043712 on Build GRCh38
rs919012985 Chr.19: 1043554-1043554 on Build GRCh38
rs775025290 Chr.19: 1043791-1043791 on Build GRCh38
rs199966168 Chr.19: 1043699-1043699 on Build GRCh38
rs545443027 Chr.19: 1043559-1043559 on Build GRCh38
rs528822740 Chr.19: 1043621-1043621 on Build GRCh38
rs4622634 Chr.19: 1043865-1043865 on Build GRCh38
rs779866979 Chr.19: 1043677-1043677 on Build GRCh38
rs561550240 Chr.19: 1043597-1043597 on Build GRCh38
rs745384559 Chr.19: 1043884-1043884 on Build GRCh38
rs545550435 Chr.19: 1043885-1043885 on Build GRCh38
rs751990303 Chr.19: 1043727-1043727 on Build GRCh38
rs374123781 Chr.19: 1043847-1043847 on Build GRCh38
rs907555253 Chr.19: 1043852-1043852 on Build GRCh38
rs778216019 Chr.19: 1043552-1043552 on Build GRCh38
rs868213967 Chr.19: 1043732-1043732 on Build GRCh38
rs1037875348 Chr.19: 1043883-1043883 on Build GRCh38
rs753983404 Chr.19: 1043857-1043857 on Build GRCh38
rs763488450 Chr.19: 1043703-1043703 on Build GRCh38
rs1014099833 Chr.19: 1043638-1043638 on Build GRCh38
rs764689222 Chr.19: 1043803-1043803 on Build GRCh38
rs916221115 Chr.19: 1043544-1043544 on Build GRCh38
rs111762673 Chr.19: 1043598-1043598 on Build GRCh38
rs772909200 Chr.19: 1043772-1043772 on Build GRCh38
rs767978009 Chr.19: 1043825-1043825 on Build GRCh38
rs576688236 Chr.19: 1043542-1043543 on Build GRCh38
rs758258750 Chr.19: 1043874-1043874 on Build GRCh38
rs1037840176 Chr.19: 1043573-1043573 on Build GRCh38
rs771798639 Chr.19: 1043768-1043768 on Build GRCh38
rs936652217 Chr.19: 1043599-1043599 on Build GRCh38
rs551118463 Chr.19: 1043802-1043803 on Build GRCh38
rs777515407 Chr.19: 1043876-1043876 on Build GRCh38
rs61050214 Chr.19: 1043799-1043799 on Build GRCh38
rs765901781 Chr.19: 1043701-1043701 on Build GRCh38
rs933091199 Chr.19: 1043558-1043558 on Build GRCh38
rs146086314 Chr.19: 1043794-1043794 on Build GRCh38

View Full Product Details