Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP2K7
Entrez Gene ID
5609
Gene Name
mitogen-activated protein kinase kinase 7
Gene Aliases
JNKK2, MAPKK7, MEK, MEK 7, MKK7, PRKMK7, SAPKK-4, SAPKK4
Chromosome Location
Chr.19: 7903780 - 7914483 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297556.1 NP_001284485.1 1 - 2 288 80
NM_145185.3 NP_660186.1 1 - 2 288 80
GenBank mRNA AF003199.1 - 1 - 2 184 80
AF006689.1 - 1 - 2 198 80
AF013588.1 - 1 - 2 190 80
AF013589.1 - 1 - 2 190 80
AF014401.1 - 1 - 2 123 80
AF022805.1 - 1 - 2 183 80
AK313899.1 - 1 - 2 189 80
BC038295.2 - 1 - 2 227 80
HY063492.1 - 1 - 2 239 80

View Full Product Details