Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2249030-2249535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMH
Entrez Gene ID
268
Gene Name
anti-Mullerian hormone
Gene Aliases
MIF, MIS
UniGene
Hs.112432
Gene Chromosome Location
Chr.19: 2249114-2252073 on Build GRCh38
Transcripts
NM_000479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5139698 c.202_203insC p.L70fs*11 Chr.19: 2249534-2249535 on Build GRCh38
6084811 c.117C>T p.F39F Chr.19: 2249449-2249449 on Build GRCh38
4131575 c.146G>T p.S49I Chr.19: 2249478-2249478 on Build GRCh38
6286653 c.131A>G p.D44G Chr.19: 2249463-2249463 on Build GRCh38
4076353 c.193G>A p.G65S Chr.19: 2249525-2249525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032283677 Chr.19: 2249254-2249254 on Build GRCh38
rs923092050 Chr.19: 2249084-2249085 on Build GRCh38
rs372862051 Chr.19: 2249219-2249219 on Build GRCh38
rs976128683 Chr.19: 2249228-2249228 on Build GRCh38
rs769424434 Chr.19: 2249377-2249377 on Build GRCh38
rs765683297 Chr.19: 2249419-2249419 on Build GRCh38
rs773283377 Chr.19: 2249331-2249331 on Build GRCh38
rs138350437 Chr.19: 2249512-2249512 on Build GRCh38
rs960479195 Chr.19: 2249421-2249421 on Build GRCh38
rs765705359 Chr.19: 2249464-2249464 on Build GRCh38
rs1056287437 Chr.19: 2249319-2249319 on Build GRCh38
rs777482098 Chr.19: 2249378-2249378 on Build GRCh38
rs759273427 Chr.19: 2249396-2249396 on Build GRCh38
rs139215834 Chr.19: 2249324-2249324 on Build GRCh38
rs775579158 Chr.19: 2249402-2249402 on Build GRCh38
rs745784090 Chr.19: 2249494-2249494 on Build GRCh38
rs747624096 Chr.19: 2249372-2249372 on Build GRCh38
rs777832730 Chr.19: 2249273-2249273 on Build GRCh38
rs914940354 Chr.19: 2249159-2249159 on Build GRCh38
rs747631258 Chr.19: 2249327-2249327 on Build GRCh38
rs993753037 Chr.19: 2249202-2249202 on Build GRCh38
rs576266262 Chr.19: 2249451-2249451 on Build GRCh38
rs778553291 Chr.19: 2249487-2249487 on Build GRCh38
rs760757922 Chr.19: 2249344-2249344 on Build GRCh38
rs142456399 Chr.19: 2249408-2249408 on Build GRCh38
rs749214406 Chr.19: 2249384-2249384 on Build GRCh38
rs149953700 Chr.19: 2249352-2249352 on Build GRCh38
rs201699789 Chr.19: 2249441-2249441 on Build GRCh38
rs538374149 Chr.19: 2249420-2249420 on Build GRCh38
rs778276367 Chr.19: 2249292-2249292 on Build GRCh38
rs367560015 Chr.19: 2249462-2249462 on Build GRCh38
rs200057041 Chr.19: 2249361-2249361 on Build GRCh38
rs752761392 Chr.19: 2249289-2249289 on Build GRCh38
rs758344648 Chr.19: 2249229-2249229 on Build GRCh38
rs764523289 Chr.19: 2249346-2249346 on Build GRCh38
rs769793732 Chr.19: 2249329-2249329 on Build GRCh38
rs769745728 Chr.19: 2249513-2249513 on Build GRCh38
rs770835311 Chr.19: 2249334-2249334 on Build GRCh38
rs934962092 Chr.19: 2249135-2249135 on Build GRCh38
rs955841317 Chr.19: 2249235-2249235 on Build GRCh38
rs777384484 Chr.19: 2249443-2249443 on Build GRCh38
rs757212843 Chr.19: 2249450-2249450 on Build GRCh38
rs768244611 Chr.19: 2249322-2249322 on Build GRCh38
rs756729865 Chr.19: 2249171-2249171 on Build GRCh38
rs1007558765 Chr.19: 2249239-2249239 on Build GRCh38
rs768909669 Chr.19: 2249460-2249460 on Build GRCh38
rs1011797489 Chr.19: 2249375-2249375 on Build GRCh38
rs762279290 Chr.19: 2249415-2249415 on Build GRCh38
rs896653648 Chr.19: 2249191-2249191 on Build GRCh38
rs543637123 Chr.19: 2249240-2249240 on Build GRCh38
rs939211612 Chr.19: 2249178-2249178 on Build GRCh38
rs914452795 Chr.19: 2249245-2249245 on Build GRCh38
rs1056759671 Chr.19: 2249207-2249207 on Build GRCh38
rs905635054 Chr.19: 2249175-2249175 on Build GRCh38
rs61736578 Chr.19: 2249385-2249385 on Build GRCh38
rs759038762 Chr.19: 2249456-2249461 on Build GRCh38
rs746807694 Chr.19: 2249264-2249264 on Build GRCh38
rs150275464 Chr.19: 2249316-2249316 on Build GRCh38
rs754144894 Chr.19: 2249300-2249300 on Build GRCh38
rs763050307 Chr.19: 2249332-2249332 on Build GRCh38
rs149082963 Chr.19: 2249367-2249367 on Build GRCh38
rs61736571 Chr.19: 2249461-2249461 on Build GRCh38
rs10406324 Chr.19: 2249113-2249113 on Build GRCh38
rs186422293 Chr.19: 2249216-2249216 on Build GRCh38
rs780718920 Chr.19: 2249371-2249371 on Build GRCh38
rs147681174 Chr.19: 2249401-2249401 on Build GRCh38
rs1044163796 Chr.19: 2249153-2249153 on Build GRCh38
rs868839560 Chr.19: 2249212-2249212 on Build GRCh38
rs747213264 Chr.19: 2249459-2249459 on Build GRCh38
rs964589114 Chr.19: 2249067-2249067 on Build GRCh38
rs904986119 Chr.19: 2249173-2249173 on Build GRCh38
rs988683306 Chr.19: 2249112-2249112 on Build GRCh38
rs998039965 Chr.19: 2249365-2249365 on Build GRCh38
rs993404831 Chr.19: 2249215-2249215 on Build GRCh38
rs148201251 Chr.19: 2249167-2249167 on Build GRCh38
rs768805891 Chr.19: 2249511-2249511 on Build GRCh38
rs1056681820 Chr.19: 2249190-2249190 on Build GRCh38
rs746840034 Chr.19: 2249509-2249509 on Build GRCh38
rs967709344 Chr.19: 2249277-2249277 on Build GRCh38
rs766829297 Chr.19: 2249423-2249423 on Build GRCh38
rs779229624 Chr.19: 2249363-2249363 on Build GRCh38
rs752774158 Chr.19: 2249340-2249340 on Build GRCh38
rs976704341 Chr.19: 2249086-2249086 on Build GRCh38
rs773582762 Chr.19: 2249466-2249466 on Build GRCh38
rs10407022 Chr.19: 2249478-2249478 on Build GRCh38
rs374948337 Chr.19: 2249330-2249330 on Build GRCh38
rs530118328 Chr.19: 2249482-2249482 on Build GRCh38
rs150481705 Chr.19: 2249498-2249498 on Build GRCh38
rs764793288 Chr.19: 2249214-2249214 on Build GRCh38
rs573689958 Chr.19: 2249506-2249506 on Build GRCh38
rs975918270 Chr.19: 2249081-2249081 on Build GRCh38
rs1056864066 Chr.19: 2249287-2249287 on Build GRCh38
rs755786762 Chr.19: 2249476-2249476 on Build GRCh38
rs767665662 Chr.19: 2249337-2249337 on Build GRCh38
rs774158206 Chr.19: 2249387-2249387 on Build GRCh38
rs748840055 Chr.19: 2249446-2249446 on Build GRCh38
rs774060964 Chr.19: 2249336-2249336 on Build GRCh38
rs113205656 Chr.19: 2249294-2249294 on Build GRCh38
rs997714648 Chr.19: 2249253-2249253 on Build GRCh38
rs753970896 Chr.19: 2249414-2249414 on Build GRCh38
rs746663444 Chr.19: 2249317-2249317 on Build GRCh38
rs764149385 Chr.19: 2249409-2249409 on Build GRCh38
rs145122767 Chr.19: 2249355-2249355 on Build GRCh38
rs756604785 Chr.19: 2249290-2249290 on Build GRCh38
rs746313611 Chr.19: 2249439-2249439 on Build GRCh38
rs756306232 Chr.19: 2249267-2249267 on Build GRCh38
rs966583896 Chr.19: 2249241-2249241 on Build GRCh38
rs551507390 Chr.19: 2249280-2249280 on Build GRCh38
rs987789210 Chr.19: 2249142-2249142 on Build GRCh38
rs148294311 Chr.19: 2249468-2249468 on Build GRCh38
rs766630178 Chr.19: 2249469-2249469 on Build GRCh38
rs141090669 Chr.19: 2249259-2249259 on Build GRCh38
rs757627048 Chr.19: 2249307-2249307 on Build GRCh38
rs1018083481 Chr.19: 2249057-2249057 on Build GRCh38
rs754607106 Chr.19: 2249369-2249369 on Build GRCh38
rs752278804 Chr.19: 2249426-2249426 on Build GRCh38
rs753342766 Chr.19: 2249479-2249479 on Build GRCh38

View Full Product Details