Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3751311-3751815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA3
Entrez Gene ID
9546
Gene Name
amyloid beta precursor protein binding family A member 3
Gene Aliases
MGC:15815, X11L2, mint3
UniGene
Hs.25527
Gene Chromosome Location
Chr.19: 3750773-3761697 on Build GRCh38
Transcripts
NM_004886.3 XM_006722950.3 XM_006722951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466793 c.1452C>T p.T484T Chr.19: 3751497-3751497 on Build GRCh38
1612131 c.1401G>T p.T467T Chr.19: 3751548-3751548 on Build GRCh38
4627667 c.1450A>T p.T484S Chr.19: 3751499-3751499 on Build GRCh38
5679571 c.1508_1511delACGG p.D503fs*>72 Chr.19: 3751438-3751441 on Build GRCh38
3938053 c.1447G>A p.V483I Chr.19: 3751502-3751502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200440373 Chr.19: 3751405-3751405 on Build GRCh38
rs770662959 Chr.19: 3751339-3751339 on Build GRCh38
rs1003551358 Chr.19: 3751407-3751407 on Build GRCh38
rs761727298 Chr.19: 3751439-3751439 on Build GRCh38
rs959214084 Chr.19: 3751427-3751427 on Build GRCh38
rs980205888 Chr.19: 3751687-3751687 on Build GRCh38
rs773016634 Chr.19: 3751501-3751501 on Build GRCh38
rs1019358667 Chr.19: 3751773-3751773 on Build GRCh38
rs766623861 Chr.19: 3751503-3751503 on Build GRCh38
rs527783566 Chr.19: 3751434-3751434 on Build GRCh38
rs752192985 Chr.19: 3751593-3751593 on Build GRCh38
rs990545302 Chr.19: 3751342-3751343 on Build GRCh38
rs754612089 Chr.19: 3751590-3751590 on Build GRCh38
rs775452454 Chr.19: 3751564-3751564 on Build GRCh38
rs953793081 Chr.19: 3751437-3751437 on Build GRCh38
rs759680039 Chr.19: 3751562-3751562 on Build GRCh38
rs141313948 Chr.19: 3751466-3751466 on Build GRCh38
rs1030844063 Chr.19: 3751788-3751788 on Build GRCh38
rs928653426 Chr.19: 3751704-3751705 on Build GRCh38
rs200671047 Chr.19: 3751433-3751433 on Build GRCh38
rs373010038 Chr.19: 3751515-3751515 on Build GRCh38
rs764738525 Chr.19: 3751570-3751570 on Build GRCh38
rs76014551 Chr.19: 3751448-3751448 on Build GRCh38
rs369996777 Chr.19: 3751449-3751449 on Build GRCh38
rs558934427 Chr.19: 3751343-3751343 on Build GRCh38
rs749144277 Chr.19: 3751551-3751551 on Build GRCh38
rs761482690 Chr.19: 3751379-3751379 on Build GRCh38
rs573242051 Chr.19: 3751472-3751472 on Build GRCh38
rs775080469 Chr.19: 3751349-3751349 on Build GRCh38
rs948678387 Chr.19: 3751651-3751651 on Build GRCh38
rs989364231 Chr.19: 3751599-3751599 on Build GRCh38
rs565415590 Chr.19: 3751394-3751394 on Build GRCh38
rs780600772 Chr.19: 3751542-3751542 on Build GRCh38
rs112086597 Chr.19: 3751533-3751533 on Build GRCh38
rs754383819 Chr.19: 3751571-3751571 on Build GRCh38
rs934230663 Chr.19: 3751450-3751450 on Build GRCh38
rs373965806 Chr.19: 3751473-3751473 on Build GRCh38
rs368843220 Chr.19: 3751378-3751378 on Build GRCh38
rs946377259 Chr.19: 3751366-3751366 on Build GRCh38
rs781525398 Chr.19: 3751402-3751402 on Build GRCh38
rs774134080 Chr.19: 3751558-3751558 on Build GRCh38
rs1012571653 Chr.19: 3751751-3751751 on Build GRCh38
rs764957695 Chr.19: 3751446-3751446 on Build GRCh38
rs1045488547 Chr.19: 3751742-3751742 on Build GRCh38
rs528885772 Chr.19: 3751540-3751540 on Build GRCh38
rs765702851 Chr.19: 3751574-3751574 on Build GRCh38
rs777742054 Chr.19: 3751605-3751605 on Build GRCh38
rs565806902 Chr.19: 3751603-3751603 on Build GRCh38
rs1006492054 Chr.19: 3751765-3751765 on Build GRCh38
rs889465578 Chr.19: 3751752-3751752 on Build GRCh38
rs1043567628 Chr.19: 3751769-3751769 on Build GRCh38
rs868124631 Chr.19: 3751481-3751481 on Build GRCh38
rs200327952 Chr.19: 3751406-3751406 on Build GRCh38
rs202068228 Chr.19: 3751410-3751410 on Build GRCh38
rs995618777 Chr.19: 3751518-3751518 on Build GRCh38
rs912518538 Chr.19: 3751369-3751369 on Build GRCh38
rs148310075 Chr.19: 3751505-3751505 on Build GRCh38
rs762717709 Chr.19: 3751502-3751502 on Build GRCh38
rs545628345 Chr.19: 3751480-3751480 on Build GRCh38
rs886393025 Chr.19: 3751413-3751413 on Build GRCh38
rs758559722 Chr.19: 3751389-3751389 on Build GRCh38
rs548638861 Chr.19: 3751393-3751393 on Build GRCh38
rs938846299 Chr.19: 3751507-3751507 on Build GRCh38
rs754492976 Chr.19: 3751344-3751345 on Build GRCh38
rs771781792 Chr.19: 3751414-3751414 on Build GRCh38
rs914590344 Chr.19: 3751640-3751640 on Build GRCh38
rs368548425 Chr.19: 3751396-3751396 on Build GRCh38
rs769775292 Chr.19: 3751548-3751548 on Build GRCh38
rs985250940 Chr.19: 3751357-3751357 on Build GRCh38
rs889414556 Chr.19: 3751532-3751532 on Build GRCh38
rs759716589 Chr.19: 3751513-3751513 on Build GRCh38
rs775632915 Chr.19: 3751563-3751563 on Build GRCh38
rs754594706 Chr.19: 3751537-3751537 on Build GRCh38
rs760814326 Chr.19: 3751569-3751569 on Build GRCh38
rs746596194 Chr.19: 3751486-3751486 on Build GRCh38
rs183105670 Chr.19: 3751595-3751595 on Build GRCh38
rs191357185 Chr.19: 3751469-3751469 on Build GRCh38
rs911501038 Chr.19: 3751622-3751622 on Build GRCh38
rs930065455 Chr.19: 3751682-3751682 on Build GRCh38
rs938727365 Chr.19: 3751712-3751712 on Build GRCh38
rs376088061 Chr.19: 3751517-3751517 on Build GRCh38
rs1048596783 Chr.19: 3751724-3751724 on Build GRCh38
rs1003382018 Chr.19: 3751415-3751415 on Build GRCh38
rs974302165 Chr.19: 3751649-3751649 on Build GRCh38
rs760692286 Chr.19: 3751351-3751351 on Build GRCh38
rs770235603 Chr.19: 3751573-3751573 on Build GRCh38
rs934653402 Chr.19: 3751382-3751382 on Build GRCh38
rs143231370 Chr.19: 3751468-3751468 on Build GRCh38
rs756584046 Chr.19: 3751478-3751478 on Build GRCh38
rs912402149 Chr.19: 3751441-3751441 on Build GRCh38
rs373707571 Chr.19: 3751353-3751353 on Build GRCh38
rs750633331 Chr.19: 3751381-3751381 on Build GRCh38
rs770695052 Chr.19: 3751552-3751552 on Build GRCh38
rs752753735 Chr.19: 3751516-3751516 on Build GRCh38
rs776428950 Chr.19: 3751438-3751438 on Build GRCh38
rs974248627 Chr.19: 3751336-3751336 on Build GRCh38
rs560491204 Chr.19: 3751696-3751696 on Build GRCh38
rs762031466 Chr.19: 3751789-3751789 on Build GRCh38
rs551557891 Chr.19: 3751782-3751782 on Build GRCh38
rs367954076 Chr.19: 3751423-3751423 on Build GRCh38
rs569266210 Chr.19: 3751343-3751343 on Build GRCh38
rs759181500 Chr.19: 3751344-3751344 on Build GRCh38
rs368852560 Chr.19: 3751380-3751380 on Build GRCh38
rs746656418 Chr.19: 3751536-3751536 on Build GRCh38
rs758146503 Chr.19: 3751484-3751484 on Build GRCh38
rs900920296 Chr.19: 3751781-3751781 on Build GRCh38
rs375775594 Chr.19: 3751485-3751485 on Build GRCh38
rs774725531 Chr.19: 3751608-3751608 on Build GRCh38
rs897275007 Chr.19: 3751716-3751716 on Build GRCh38
rs760040806 Chr.19: 3751491-3751493 on Build GRCh38
rs954995342 Chr.19: 3751611-3751611 on Build GRCh38
rs768038344 Chr.19: 3751493-3751493 on Build GRCh38
rs771392976 Chr.19: 3751398-3751399 on Build GRCh38
rs548806609 Chr.19: 3751665-3751665 on Build GRCh38
rs1019296459 Chr.19: 3751534-3751534 on Build GRCh38
rs577323467 Chr.19: 3751399-3751399 on Build GRCh38
rs895105873 Chr.19: 3751730-3751730 on Build GRCh38
rs369792298 Chr.19: 3751494-3751494 on Build GRCh38
rs893220750 Chr.19: 3751395-3751395 on Build GRCh38
rs115055172 Chr.19: 3751411-3751411 on Build GRCh38
rs141575653 Chr.19: 3751527-3751527 on Build GRCh38
rs776872539 Chr.19: 3751411-3751417 on Build GRCh38
rs903662404 Chr.19: 3751778-3751778 on Build GRCh38
rs183053013 Chr.19: 3751335-3751335 on Build GRCh38
rs748042156 Chr.19: 3751497-3751497 on Build GRCh38
rs747648736 Chr.19: 3751543-3751543 on Build GRCh38
rs986857837 Chr.19: 3751613-3751613 on Build GRCh38
rs557401703 Chr.19: 3751417-3751417 on Build GRCh38
rs773106862 Chr.19: 3751549-3751549 on Build GRCh38
rs773822465 Chr.19: 3751580-3751581 on Build GRCh38
rs368571496 Chr.19: 3751405-3751405 on Build GRCh38
rs750762177 Chr.19: 3751589-3751589 on Build GRCh38
rs774064406 Chr.19: 3751470-3751470 on Build GRCh38
rs931588845 Chr.19: 3751719-3751719 on Build GRCh38
rs535187846 Chr.19: 3751352-3751352 on Build GRCh38
rs938881835 Chr.19: 3751700-3751700 on Build GRCh38
rs780116039 Chr.19: 3751392-3751392 on Build GRCh38
rs749628849 Chr.19: 3751482-3751482 on Build GRCh38
rs1037199727 Chr.19: 3751709-3751709 on Build GRCh38
rs769681561 Chr.19: 3751500-3751500 on Build GRCh38

View Full Product Details