Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4202584-4202993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
UniGene
Hs.197872
Gene Chromosome Location
Chr.19: 4182618-4224814 on Build GRCh38
Transcripts
NM_133475.1 XM_011527756.2 XM_011527757.2 XM_011527758.2 XM_011527761.1 XM_011527764.2 XM_011527765.2 XM_017026438.1 XM_017026439.1 XM_017026440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413901 c.114C>A p.S38R Chr.19: 4202898-4202898 on Build GRCh38
5892357 c.460C>T p.H154Y Chr.19: 4202920-4202920 on Build GRCh38
6600483 c.433G>T p.D145Y Chr.19: 4202893-4202893 on Build GRCh38
6600484 c.703G>T p.D235Y Chr.19: 4202893-4202893 on Build GRCh38
5892358 c.730C>T p.H244Y Chr.19: 4202920-4202920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554658401 Chr.19: 4202957-4202957 on Build GRCh38
rs759324121 Chr.19: 4202889-4202889 on Build GRCh38
rs776679160 Chr.19: 4202877-4202877 on Build GRCh38
rs352513 Chr.19: 4202686-4202686 on Build GRCh38
rs370472657 Chr.19: 4202856-4202856 on Build GRCh38
rs753395717 Chr.19: 4202944-4202944 on Build GRCh38
rs753451399 Chr.19: 4202846-4202846 on Build GRCh38
rs199870593 Chr.19: 4202855-4202855 on Build GRCh38
rs966292885 Chr.19: 4202685-4202685 on Build GRCh38
rs750761174 Chr.19: 4202755-4202755 on Build GRCh38
rs372973074 Chr.19: 4202814-4202814 on Build GRCh38
rs908932821 Chr.19: 4202791-4202791 on Build GRCh38
rs953670493 Chr.19: 4202760-4202760 on Build GRCh38
rs754730716 Chr.19: 4202828-4202828 on Build GRCh38
rs893230891 Chr.19: 4202610-4202610 on Build GRCh38
rs769200053 Chr.19: 4202864-4202864 on Build GRCh38
rs574474370 Chr.19: 4202968-4202968 on Build GRCh38
rs536272236 Chr.19: 4202800-4202800 on Build GRCh38
rs1040532209 Chr.19: 4202881-4202881 on Build GRCh38
rs769988640 Chr.19: 4202878-4202878 on Build GRCh38
rs767250492 Chr.19: 4202901-4202901 on Build GRCh38
rs373345352 Chr.19: 4202887-4202887 on Build GRCh38
rs981216414 Chr.19: 4202693-4202693 on Build GRCh38
rs917559381 Chr.19: 4202861-4202861 on Build GRCh38
rs7260407 Chr.19: 4202796-4202796 on Build GRCh38
rs752705824 Chr.19: 4202832-4202832 on Build GRCh38
rs1020318031 Chr.19: 4202632-4202632 on Build GRCh38
rs117709875 Chr.19: 4202630-4202630 on Build GRCh38
rs377567694 Chr.19: 4202842-4202842 on Build GRCh38
rs570931108 Chr.19: 4202777-4202777 on Build GRCh38
rs374613901 Chr.19: 4202902-4202902 on Build GRCh38
rs756922146 Chr.19: 4202851-4202851 on Build GRCh38
rs1029271120 Chr.19: 4202689-4202689 on Build GRCh38
rs767428132 Chr.19: 4202829-4202829 on Build GRCh38
rs201914800 Chr.19: 4202956-4202956 on Build GRCh38
rs908385087 Chr.19: 4202746-4202746 on Build GRCh38
rs566801844 Chr.19: 4202841-4202841 on Build GRCh38
rs553289586 Chr.19: 4202804-4202804 on Build GRCh38
rs764184826 Chr.19: 4202931-4202931 on Build GRCh38
rs780497315 Chr.19: 4202863-4202863 on Build GRCh38
rs1023035068 Chr.19: 4202631-4202631 on Build GRCh38
rs773316878 Chr.19: 4202886-4202886 on Build GRCh38
rs747514631 Chr.19: 4202871-4202871 on Build GRCh38
rs772338854 Chr.19: 4202858-4202858 on Build GRCh38
rs760671197 Chr.19: 4202907-4202907 on Build GRCh38

View Full Product Details