Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4207677-4208184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
UniGene
Hs.197872
Gene Chromosome Location
Chr.19: 4182618-4224814 on Build GRCh38
Transcripts
NM_133475.1 XM_011527756.2 XM_011527757.2 XM_011527758.2 XM_011527761.1 XM_011527764.2 XM_011527765.2 XM_017026438.1 XM_017026439.1 XM_017026440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4749003 c.425C>T p.A142V Chr.19: 4207885-4207885 on Build GRCh38
4792916 c.507C>T p.S169S Chr.19: 4207967-4207967 on Build GRCh38
3823189 c.358G>A p.E120K Chr.19: 4207818-4207818 on Build GRCh38
6345123 c.735G>T p.A245A Chr.19: 4207871-4207871 on Build GRCh38
5732598 c.491G>A p.R164H Chr.19: 4207951-4207951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180985811 Chr.19: 4208060-4208060 on Build GRCh38
rs751638416 Chr.19: 4207747-4207747 on Build GRCh38
rs2240670 Chr.19: 4208085-4208085 on Build GRCh38
rs769617334 Chr.19: 4207891-4207891 on Build GRCh38
rs755217002 Chr.19: 4207767-4207767 on Build GRCh38
rs766449144 Chr.19: 4207743-4207743 on Build GRCh38
rs1045886446 Chr.19: 4207894-4207894 on Build GRCh38
rs746465385 Chr.19: 4207884-4207884 on Build GRCh38
rs373534420 Chr.19: 4207838-4207838 on Build GRCh38
rs779858519 Chr.19: 4207883-4207883 on Build GRCh38
rs369150437 Chr.19: 4207817-4207817 on Build GRCh38
rs561235943 Chr.19: 4207907-4207907 on Build GRCh38
rs61742314 Chr.19: 4207872-4207872 on Build GRCh38
rs1012714011 Chr.19: 4208063-4208063 on Build GRCh38
rs770312302 Chr.19: 4207851-4207851 on Build GRCh38
rs751478860 Chr.19: 4207953-4207953 on Build GRCh38
rs185427256 Chr.19: 4208068-4208068 on Build GRCh38
rs555033453 Chr.19: 4208124-4208124 on Build GRCh38
rs139056414 Chr.19: 4207739-4207739 on Build GRCh38
rs566627945 Chr.19: 4208136-4208136 on Build GRCh38
rs545144384 Chr.19: 4208071-4208071 on Build GRCh38
rs1032789940 Chr.19: 4207854-4207854 on Build GRCh38
rs61742016 Chr.19: 4207967-4207967 on Build GRCh38
rs201999944 Chr.19: 4207950-4207950 on Build GRCh38
rs752311142 Chr.19: 4207968-4207968 on Build GRCh38
rs762667393 Chr.19: 4207951-4207951 on Build GRCh38
rs766287233 Chr.19: 4207952-4207952 on Build GRCh38
rs776433149 Chr.19: 4207844-4207844 on Build GRCh38
rs779596209 Chr.19: 4207820-4207820 on Build GRCh38
rs115845796 Chr.19: 4208143-4208143 on Build GRCh38
rs961058088 Chr.19: 4208139-4208139 on Build GRCh38
rs867331343 Chr.19: 4207954-4207954 on Build GRCh38
rs2240669 Chr.19: 4207943-4207943 on Build GRCh38
rs1010790842 Chr.19: 4207935-4207935 on Build GRCh38
rs545457141 Chr.19: 4207731-4207731 on Build GRCh38
rs753562794 Chr.19: 4207790-4207790 on Build GRCh38
rs746378055 Chr.19: 4207754-4207756 on Build GRCh38
rs777297305 Chr.19: 4207991-4207991 on Build GRCh38
rs773751643 Chr.19: 4207853-4207853 on Build GRCh38
rs749610511 Chr.19: 4207992-4207992 on Build GRCh38
rs190977424 Chr.19: 4208098-4208098 on Build GRCh38
rs775592071 Chr.19: 4207938-4207938 on Build GRCh38
rs564857105 Chr.19: 4208066-4208066 on Build GRCh38
rs1054442019 Chr.19: 4208050-4208050 on Build GRCh38
rs7251041 Chr.19: 4207855-4207855 on Build GRCh38
rs893406327 Chr.19: 4207917-4207918 on Build GRCh38
rs769374619 Chr.19: 4207906-4207906 on Build GRCh38
rs930768643 Chr.19: 4207928-4207928 on Build GRCh38
rs767795062 Chr.19: 4207774-4207774 on Build GRCh38
rs774486322 Chr.19: 4207857-4207857 on Build GRCh38
rs747559851 Chr.19: 4207880-4207880 on Build GRCh38
rs1050726940 Chr.19: 4207803-4207803 on Build GRCh38
rs747772173 Chr.19: 4207886-4207886 on Build GRCh38
rs920745545 Chr.19: 4207920-4207920 on Build GRCh38
rs765488561 Chr.19: 4207870-4207870 on Build GRCh38
rs759541180 Chr.19: 4207864-4207864 on Build GRCh38
rs899367523 Chr.19: 4207776-4207776 on Build GRCh38
rs560706247 Chr.19: 4207955-4207955 on Build GRCh38
rs768093620 Chr.19: 4207867-4207868 on Build GRCh38
rs757009734 Chr.19: 4207866-4207866 on Build GRCh38
rs779436558 Chr.19: 4208016-4208016 on Build GRCh38
rs758310661 Chr.19: 4207818-4207818 on Build GRCh38
rs374354798 Chr.19: 4207885-4207885 on Build GRCh38
rs953015568 Chr.19: 4207974-4207974 on Build GRCh38
rs544854127 Chr.19: 4207929-4207929 on Build GRCh38
rs778504405 Chr.19: 4207928-4207928 on Build GRCh38
rs746564435 Chr.19: 4207834-4207834 on Build GRCh38
rs574271131 Chr.19: 4207898-4207898 on Build GRCh38
rs1008875144 Chr.19: 4207810-4207810 on Build GRCh38
rs760866449 Chr.19: 4207940-4207940 on Build GRCh38
rs534848864 Chr.19: 4207865-4207865 on Build GRCh38
rs998882908 Chr.19: 4208045-4208045 on Build GRCh38
rs778777281 Chr.19: 4207816-4207816 on Build GRCh38
rs773878199 Chr.19: 4207735-4207735 on Build GRCh38
rs766852052 Chr.19: 4207856-4207856 on Build GRCh38
rs200294304 Chr.19: 4207960-4207960 on Build GRCh38
rs771222702 Chr.19: 4208005-4208005 on Build GRCh38
rs748471740 Chr.19: 4207845-4207845 on Build GRCh38
rs551438176 Chr.19: 4208043-4208043 on Build GRCh38
rs747712572 Chr.19: 4207966-4207967 on Build GRCh38
rs568351372 Chr.19: 4207899-4207899 on Build GRCh38
rs370963079 Chr.19: 4207871-4207871 on Build GRCh38
rs930862039 Chr.19: 4207791-4207791 on Build GRCh38
rs757058728 Chr.19: 4207809-4207809 on Build GRCh38
rs755823479 Chr.19: 4207983-4207983 on Build GRCh38

View Full Product Details