The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024937.3 | NP_001020108.1 | 2 - 3 | 369 | 62 |
NM_001321236.1 | NP_001308165.1 | 3 - 4 | 485 | 62 | |
NM_015716.4 | NP_056531.1 | 2 - 3 | 369 | 62 | |
NM_153827.4 | NP_722549.2 | 2 - 3 | 369 | 62 | |
NM_170663.4 | NP_733763.1 | 2 - 3 | 369 | 62 | |
XM_006721531.2 | XP_006721594.1 | 2 - 3 | 369 | 62 | |
XM_006721532.2 | XP_006721595.1 | 2 - 3 | 369 | 62 | |
XM_006721536.2 | XP_006721599.1 | 2 - 3 | 369 | 62 | |
XM_011523906.1 | XP_011522208.1 | 2 - 3 | 369 | 62 | |
XM_017024704.1 | XP_016880193.1 | 2 - 3 | 369 | 62 | |
XM_017024705.1 | XP_016880194.1 | 2 - 3 | 369 | 62 | |
XM_017024706.1 | XP_016880195.1 | 2 - 3 | 369 | 62 | |
XM_017024707.1 | XP_016880196.1 | 2 - 3 | 170 | 62 | |
GenBank mRNA | AB035698.1 | - | 2 - 3 | 125 | 62 |
AB041926.1 | - | 2 - 3 | 309 | 62 | |
AB209450.1 | - | 2 - 3 | 310 | 62 | |
AY775058.1 | - | 2 - 3 | 125 | 62 | |
BC034673.1 | - | 2 - 3 | 321 | 62 | |
CA392419.1 | - | 2 - 3 | 342 | 62 | |
DA653105.1 | - | 2 - 3 | 369 | 62 |