Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5866516-5867041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT5
Entrez Gene ID
2527
Gene Name
fucosyltransferase 5
Gene Aliases
FUC-TV
UniGene
Hs.631843
Gene Chromosome Location
Chr.19: 5865826-5870540 on Build GRCh38
Transcripts
NM_002034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3748076 c.727C>G p.P243A Chr.19: 5866999-5866999 on Build GRCh38
1641261 c.749C>T p.T250M Chr.19: 5866977-5866977 on Build GRCh38
1641262 c.740T>C p.M247T Chr.19: 5866986-5866986 on Build GRCh38
1396046 c.1013C>T p.T338M Chr.19: 5866713-5866713 on Build GRCh38
6319193 c.1053C>A p.C351* Chr.19: 5866673-5866673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758815585 Chr.19: 5866999-5866999 on Build GRCh38
rs768406993 Chr.19: 5866758-5866758 on Build GRCh38
rs773631667 Chr.19: 5866726-5866726 on Build GRCh38
rs891573142 Chr.19: 5866949-5866949 on Build GRCh38
rs61730511 Chr.19: 5866658-5866658 on Build GRCh38
rs746865173 Chr.19: 5866751-5866751 on Build GRCh38
rs200448911 Chr.19: 5866617-5866617 on Build GRCh38
rs577735251 Chr.19: 5866623-5866623 on Build GRCh38
rs201842894 Chr.19: 5866927-5866927 on Build GRCh38
rs757206885 Chr.19: 5866701-5866701 on Build GRCh38
rs762311380 Chr.19: 5866657-5866657 on Build GRCh38
rs201483736 Chr.19: 5866719-5866719 on Build GRCh38
rs142202309 Chr.19: 5866682-5866682 on Build GRCh38
rs772800545 Chr.19: 5866786-5866786 on Build GRCh38
rs939886425 Chr.19: 5866544-5866544 on Build GRCh38
rs765697026 Chr.19: 5866928-5866928 on Build GRCh38
rs200537067 Chr.19: 5867017-5867017 on Build GRCh38
rs753443559 Chr.19: 5866987-5866987 on Build GRCh38
rs774686203 Chr.19: 5866806-5866806 on Build GRCh38
rs751501795 Chr.19: 5866889-5866889 on Build GRCh38
rs758402932 Chr.19: 5866826-5866826 on Build GRCh38
rs141679775 Chr.19: 5866825-5866825 on Build GRCh38
rs766591948 Chr.19: 5866686-5866686 on Build GRCh38
rs766383156 Chr.19: 5866888-5866888 on Build GRCh38
rs771406213 Chr.19: 5866865-5866865 on Build GRCh38
rs772576224 Chr.19: 5866929-5866929 on Build GRCh38
rs571467519 Chr.19: 5866874-5866874 on Build GRCh38
rs374028 Chr.19: 5866538-5866538 on Build GRCh38
rs760744218 Chr.19: 5866879-5866879 on Build GRCh38
rs1010414065 Chr.19: 5866614-5866614 on Build GRCh38
rs745747279 Chr.19: 5866747-5866747 on Build GRCh38
rs774032270 Chr.19: 5866603-5866603 on Build GRCh38
rs779178800 Chr.19: 5866576-5866576 on Build GRCh38
rs779278026 Chr.19: 5866621-5866621 on Build GRCh38
rs755190918 Chr.19: 5866737-5866737 on Build GRCh38
rs372756609 Chr.19: 5866958-5866958 on Build GRCh38
rs779925260 Chr.19: 5866750-5866750 on Build GRCh38
rs148377456 Chr.19: 5867020-5867020 on Build GRCh38
rs749168808 Chr.19: 5866637-5866637 on Build GRCh38
rs200790634 Chr.19: 5866648-5866648 on Build GRCh38
rs749649222 Chr.19: 5866572-5866572 on Build GRCh38
rs407788 Chr.19: 5866571-5866571 on Build GRCh38
rs376720049 Chr.19: 5866928-5866928 on Build GRCh38
rs747300407 Chr.19: 5867013-5867013 on Build GRCh38
rs772952496 Chr.19: 5866575-5866575 on Build GRCh38
rs770142679 Chr.19: 5867021-5867021 on Build GRCh38
rs756192247 Chr.19: 5866746-5866746 on Build GRCh38
rs764440838 Chr.19: 5866880-5866880 on Build GRCh38
rs745452458 Chr.19: 5866998-5866998 on Build GRCh38
rs549863528 Chr.19: 5866768-5866768 on Build GRCh38
rs769020990 Chr.19: 5867016-5867016 on Build GRCh38
rs139123543 Chr.19: 5866707-5866707 on Build GRCh38
rs753410108 Chr.19: 5866655-5866655 on Build GRCh38
rs200114630 Chr.19: 5866549-5866549 on Build GRCh38
rs939855361 Chr.19: 5866679-5866679 on Build GRCh38
rs764025766 Chr.19: 5866823-5866823 on Build GRCh38
rs752413777 Chr.19: 5866976-5866976 on Build GRCh38
rs773332009 Chr.19: 5866570-5866571 on Build GRCh38
rs147659571 Chr.19: 5866956-5866956 on Build GRCh38
rs776273064 Chr.19: 5866723-5866723 on Build GRCh38
rs771727730 Chr.19: 5866708-5866708 on Build GRCh38
rs752826737 Chr.19: 5866820-5866820 on Build GRCh38
rs145269810 Chr.19: 5866776-5866776 on Build GRCh38
rs143372058 Chr.19: 5866977-5866977 on Build GRCh38
rs774878594 Chr.19: 5866870-5866870 on Build GRCh38
rs778482426 Chr.19: 5866567-5866567 on Build GRCh38
rs767556619 Chr.19: 5866970-5866970 on Build GRCh38
rs758641430 Chr.19: 5866624-5866624 on Build GRCh38
rs776852707 Chr.19: 5866628-5866630 on Build GRCh38
rs778982 Chr.19: 5866574-5866574 on Build GRCh38
rs755710824 Chr.19: 5866898-5866898 on Build GRCh38
rs150801033 Chr.19: 5866700-5866700 on Build GRCh38
rs780198147 Chr.19: 5866626-5866626 on Build GRCh38
rs926921600 Chr.19: 5866883-5866883 on Build GRCh38
rs145337594 Chr.19: 5866830-5866830 on Build GRCh38
rs771077566 Chr.19: 5866619-5866620 on Build GRCh38
rs199621061 Chr.19: 5866933-5866933 on Build GRCh38
rs753846498 Chr.19: 5866882-5866882 on Build GRCh38
rs754987658 Chr.19: 5866894-5866894 on Build GRCh38
rs528879578 Chr.19: 5866811-5866811 on Build GRCh38
rs773180104 Chr.19: 5866594-5866594 on Build GRCh38
rs4807054 Chr.19: 5866713-5866713 on Build GRCh38
rs374296055 Chr.19: 5866961-5866961 on Build GRCh38
rs778983 Chr.19: 5866643-5866643 on Build GRCh38
rs202015279 Chr.19: 5866591-5866591 on Build GRCh38
rs542830434 Chr.19: 5866631-5866631 on Build GRCh38
rs747158240 Chr.19: 5866589-5866589 on Build GRCh38
rs770716529 Chr.19: 5866596-5866596 on Build GRCh38
rs767146265 Chr.19: 5866615-5866615 on Build GRCh38
rs917216356 Chr.19: 5866854-5866854 on Build GRCh38
rs769701969 Chr.19: 5866592-5866592 on Build GRCh38
rs4044964 Chr.19: 5866955-5866955 on Build GRCh38
rs573940537 Chr.19: 5866641-5866641 on Build GRCh38
rs374202687 Chr.19: 5866743-5866743 on Build GRCh38
rs145203525 Chr.19: 5866588-5866588 on Build GRCh38
rs752156380 Chr.19: 5866564-5866564 on Build GRCh38
rs763650899 Chr.19: 5866652-5866652 on Build GRCh38
rs982770016 Chr.19: 5867010-5867010 on Build GRCh38
rs752539073 Chr.19: 5866897-5866897 on Build GRCh38
rs761284135 Chr.19: 5866724-5866724 on Build GRCh38
rs748887640 Chr.19: 5866856-5866856 on Build GRCh38
rs747064405 Chr.19: 5866629-5866629 on Build GRCh38
rs759818955 Chr.19: 5866777-5866777 on Build GRCh38
rs200309409 Chr.19: 5866680-5866680 on Build GRCh38
rs780790781 Chr.19: 5866896-5866896 on Build GRCh38
rs200111389 Chr.19: 5866945-5866945 on Build GRCh38
rs375796472 Chr.19: 5866595-5866595 on Build GRCh38
rs751927906 Chr.19: 5866732-5866732 on Build GRCh38
rs702065 Chr.19: 5866829-5866829 on Build GRCh38
rs368343456 Chr.19: 5866915-5866915 on Build GRCh38
rs771734106 Chr.19: 5866644-5866644 on Build GRCh38
rs770388340 Chr.19: 5866916-5866916 on Build GRCh38
rs769145980 Chr.19: 5866780-5866780 on Build GRCh38
rs756689655 Chr.19: 5866566-5866566 on Build GRCh38
rs557931883 Chr.19: 5866838-5866838 on Build GRCh38
rs149142282 Chr.19: 5866968-5866968 on Build GRCh38
rs754359097 Chr.19: 5866620-5866620 on Build GRCh38
rs368467966 Chr.19: 5866632-5866632 on Build GRCh38
rs201392917 Chr.19: 5866744-5866744 on Build GRCh38
rs759944233 Chr.19: 5866810-5866810 on Build GRCh38
rs200864352 Chr.19: 5866731-5866731 on Build GRCh38
rs201093757 Chr.19: 5866587-5866587 on Build GRCh38
rs533066806 Chr.19: 5866748-5866748 on Build GRCh38
rs898937147 Chr.19: 5866687-5866687 on Build GRCh38
rs145948674 Chr.19: 5866627-5866627 on Build GRCh38
rs199731954 Chr.19: 5866720-5866720 on Build GRCh38
rs549680928 Chr.19: 5866827-5866827 on Build GRCh38
rs28541500 Chr.19: 5866805-5866805 on Build GRCh38
rs949738999 Chr.19: 5866710-5866710 on Build GRCh38
rs760071180 Chr.19: 5866873-5866873 on Build GRCh38
rs746051810 Chr.19: 5866582-5866582 on Build GRCh38
rs147900234 Chr.19: 5866777-5866777 on Build GRCh38
rs61730514 Chr.19: 5866996-5866996 on Build GRCh38
rs370489070 Chr.19: 5866729-5866729 on Build GRCh38
rs778984 Chr.19: 5866725-5866725 on Build GRCh38
rs760438160 Chr.19: 5866651-5866651 on Build GRCh38
rs761157042 Chr.19: 5866946-5866946 on Build GRCh38
rs1016005845 Chr.19: 5866543-5866543 on Build GRCh38
rs778421572 Chr.19: 5866924-5866924 on Build GRCh38
rs760716657 Chr.19: 5866577-5866578 on Build GRCh38
rs751739700 Chr.19: 5866692-5866692 on Build GRCh38
rs755213521 Chr.19: 5866698-5866698 on Build GRCh38
rs759164256 Chr.19: 5866611-5866611 on Build GRCh38
rs760983276 Chr.19: 5866822-5866822 on Build GRCh38
rs377049009 Chr.19: 5866616-5866616 on Build GRCh38
rs769499062 Chr.19: 5866844-5866844 on Build GRCh38
rs763308831 Chr.19: 5866796-5866796 on Build GRCh38
rs61730515 Chr.19: 5866991-5866991 on Build GRCh38
rs759478889 Chr.19: 5866969-5866969 on Build GRCh38
rs780658085 Chr.19: 5867001-5867001 on Build GRCh38
rs781164561 Chr.19: 5866843-5866843 on Build GRCh38
rs777532476 Chr.19: 5866904-5866904 on Build GRCh38

View Full Product Details