Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6147380-6147895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSBG2
Entrez Gene ID
81616
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
BGR, BRGL, PRTD-NY3, PRTDNY3
UniGene
Hs.465720
Gene Chromosome Location
Chr.19: 6135633-6193103 on Build GRCh38
Transcripts
NM_001289177.1 NM_001289178.1 NM_001289179.1 NM_001289180.1 NM_001321384.1 NM_030924.4 XM_005259653.3 XM_017027332.1 XM_017027333.1 XM_017027334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139112 c.173G>A p.R58Q Chr.19: 6147551-6147551 on Build GRCh38
6592531 c.230A>T p.K77M Chr.19: 6147608-6147608 on Build GRCh38
3736208 c.158T>C p.I53T Chr.19: 6147536-6147536 on Build GRCh38
4658969 c.263A>C p.E88A Chr.19: 6147641-6147641 on Build GRCh38
4469621 c.160C>T p.P54S Chr.19: 6147538-6147538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780212769 Chr.19: 6147614-6147614 on Build GRCh38
rs761813244 Chr.19: 6147537-6147537 on Build GRCh38
rs146288460 Chr.19: 6147629-6147629 on Build GRCh38
rs945285605 Chr.19: 6147853-6147853 on Build GRCh38
rs761057372 Chr.19: 6147671-6147671 on Build GRCh38
rs267605691 Chr.19: 6147487-6147487 on Build GRCh38
rs533225894 Chr.19: 6147454-6147454 on Build GRCh38
rs149614793 Chr.19: 6147568-6147568 on Build GRCh38
rs577031271 Chr.19: 6147588-6147588 on Build GRCh38
rs936039891 Chr.19: 6147483-6147483 on Build GRCh38
rs758667944 Chr.19: 6147722-6147722 on Build GRCh38
rs771978866 Chr.19: 6147600-6147600 on Build GRCh38
rs757637910 Chr.19: 6147678-6147678 on Build GRCh38
rs767294404 Chr.19: 6147637-6147637 on Build GRCh38
rs141830378 Chr.19: 6147471-6147471 on Build GRCh38
rs200700863 Chr.19: 6147591-6147591 on Build GRCh38
rs746012119 Chr.19: 6147512-6147512 on Build GRCh38
rs74980839 Chr.19: 6147665-6147665 on Build GRCh38
rs1023602457 Chr.19: 6147777-6147777 on Build GRCh38
rs1054502817 Chr.19: 6147846-6147846 on Build GRCh38
rs971431255 Chr.19: 6147739-6147739 on Build GRCh38
rs763503021 Chr.19: 6147686-6147686 on Build GRCh38
rs143949111 Chr.19: 6147638-6147638 on Build GRCh38
rs186319373 Chr.19: 6147536-6147536 on Build GRCh38
rs958589732 Chr.19: 6147723-6147723 on Build GRCh38
rs749549988 Chr.19: 6147423-6147423 on Build GRCh38
rs754295937 Chr.19: 6147589-6147589 on Build GRCh38
rs775460618 Chr.19: 6147523-6147524 on Build GRCh38
rs139328780 Chr.19: 6147459-6147459 on Build GRCh38
rs893760566 Chr.19: 6147518-6147518 on Build GRCh38
rs935994859 Chr.19: 6147792-6147792 on Build GRCh38
rs771329990 Chr.19: 6147508-6147508 on Build GRCh38
rs940861063 Chr.19: 6147531-6147531 on Build GRCh38
rs766056617 Chr.19: 6147565-6147565 on Build GRCh38
rs527351973 Chr.19: 6147746-6147746 on Build GRCh38
rs534347073 Chr.19: 6147551-6147551 on Build GRCh38
rs774583743 Chr.19: 6147450-6147450 on Build GRCh38
rs773930895 Chr.19: 6147650-6147650 on Build GRCh38
rs751364914 Chr.19: 6147473-6147473 on Build GRCh38
rs767062226 Chr.19: 6147572-6147572 on Build GRCh38
rs982175083 Chr.19: 6147864-6147864 on Build GRCh38
rs905878459 Chr.19: 6147481-6147481 on Build GRCh38
rs570663483 Chr.19: 6147550-6147550 on Build GRCh38
rs779973277 Chr.19: 6147726-6147726 on Build GRCh38
rs907951441 Chr.19: 6147866-6147866 on Build GRCh38
rs530669676 Chr.19: 6147791-6147791 on Build GRCh38
rs142290585 Chr.19: 6147472-6147472 on Build GRCh38
rs758076555 Chr.19: 6147470-6147470 on Build GRCh38
rs80235752 Chr.19: 6147841-6147841 on Build GRCh38
rs890123713 Chr.19: 6147610-6147610 on Build GRCh38
rs374169522 Chr.19: 6147721-6147721 on Build GRCh38
rs779423915 Chr.19: 6147694-6147694 on Build GRCh38
rs972840521 Chr.19: 6147693-6147693 on Build GRCh38
rs764765337 Chr.19: 6147672-6147672 on Build GRCh38
rs200855627 Chr.19: 6147527-6147527 on Build GRCh38
rs751060741 Chr.19: 6147566-6147566 on Build GRCh38
rs755720445 Chr.19: 6147676-6147676 on Build GRCh38
rs111748149 Chr.19: 6147587-6147587 on Build GRCh38
rs774542751 Chr.19: 6147510-6147510 on Build GRCh38
rs759267056 Chr.19: 6147660-6147660 on Build GRCh38
rs181467954 Chr.19: 6147760-6147760 on Build GRCh38
rs781611507 Chr.19: 6147476-6147476 on Build GRCh38
rs747985013 Chr.19: 6147626-6147626 on Build GRCh38
rs138620159 Chr.19: 6147649-6147649 on Build GRCh38
rs748707371 Chr.19: 6147495-6147495 on Build GRCh38
rs761562256 Chr.19: 6147461-6147461 on Build GRCh38
rs371939042 Chr.19: 6147541-6147541 on Build GRCh38
rs1032870024 Chr.19: 6147688-6147688 on Build GRCh38
rs796070807 Chr.19: 6147759-6147759 on Build GRCh38
rs756668627 Chr.19: 6147585-6147585 on Build GRCh38
rs752303762 Chr.19: 6147669-6147669 on Build GRCh38
rs368884285 Chr.19: 6147528-6147528 on Build GRCh38
rs549380931 Chr.19: 6147847-6147847 on Build GRCh38
rs949469351 Chr.19: 6147500-6147500 on Build GRCh38
rs778234519 Chr.19: 6147499-6147499 on Build GRCh38
rs372826000 Chr.19: 6147468-6147468 on Build GRCh38
rs746101885 Chr.19: 6147599-6147599 on Build GRCh38
rs138248184 Chr.19: 6147532-6147532 on Build GRCh38
rs9304929 Chr.19: 6147732-6147732 on Build GRCh38
rs756370137 Chr.19: 6147498-6147498 on Build GRCh38
rs760675119 Chr.19: 6147455-6147455 on Build GRCh38
rs16993408 Chr.19: 6147442-6147442 on Build GRCh38
rs765103941 Chr.19: 6147467-6147467 on Build GRCh38
rs189150426 Chr.19: 6147639-6147639 on Build GRCh38
rs753108543 Chr.19: 6147581-6147581 on Build GRCh38
rs749474710 Chr.19: 6147507-6147507 on Build GRCh38

View Full Product Details