Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6182669-6183172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSBG2
Entrez Gene ID
81616
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
BGR, BRGL, PRTD-NY3, PRTDNY3
UniGene
Hs.465720
Gene Chromosome Location
Chr.19: 6135633-6193103 on Build GRCh38
Transcripts
NM_001289177.1 NM_001289178.1 NM_001289179.1 NM_001289180.1 NM_001321384.1 NM_030924.4 XM_005259653.3 XM_011528329.1 XM_017027332.1 XM_017027333.1 XM_017027334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2727150 c.1043G>A p.W348* Chr.19: 6182887-6182887 on Build GRCh38
5941408 c.1210G>A p.E404K Chr.19: 6183160-6183160 on Build GRCh38
6592529 c.1039G>A p.V347M Chr.19: 6182883-6182883 on Build GRCh38
2727148 c.948C>A p.V316V Chr.19: 6182792-6182792 on Build GRCh38
4658970 c.1190G>T p.G397V Chr.19: 6183140-6183140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938240240 Chr.19: 6182834-6182834 on Build GRCh38
rs778081008 Chr.19: 6182905-6182905 on Build GRCh38
rs563799248 Chr.19: 6183016-6183016 on Build GRCh38
rs755541739 Chr.19: 6183007-6183007 on Build GRCh38
rs775398697 Chr.19: 6183115-6183115 on Build GRCh38
rs1017473254 Chr.19: 6182709-6182709 on Build GRCh38
rs768241359 Chr.19: 6182777-6182777 on Build GRCh38
rs769638958 Chr.19: 6183073-6183073 on Build GRCh38
rs780488549 Chr.19: 6183075-6183075 on Build GRCh38
rs750935112 Chr.19: 6183068-6183068 on Build GRCh38
rs551610771 Chr.19: 6182983-6182983 on Build GRCh38
rs768569446 Chr.19: 6183038-6183038 on Build GRCh38
rs770004413 Chr.19: 6183110-6183110 on Build GRCh38
rs757195089 Chr.19: 6182746-6182746 on Build GRCh38
rs988211332 Chr.19: 6182758-6182758 on Build GRCh38
rs375380589 Chr.19: 6183113-6183113 on Build GRCh38
rs777367724 Chr.19: 6183013-6183013 on Build GRCh38
rs773494730 Chr.19: 6182718-6182718 on Build GRCh38
rs577521043 Chr.19: 6182741-6182741 on Build GRCh38
rs377656710 Chr.19: 6182850-6182850 on Build GRCh38
rs749827875 Chr.19: 6182924-6182924 on Build GRCh38
rs772500319 Chr.19: 6183024-6183024 on Build GRCh38
rs199630235 Chr.19: 6182939-6182939 on Build GRCh38
rs1005762261 Chr.19: 6183117-6183117 on Build GRCh38
rs1057118397 Chr.19: 6182845-6182845 on Build GRCh38
rs113123404 Chr.19: 6182745-6182745 on Build GRCh38
rs746290076 Chr.19: 6183022-6183022 on Build GRCh38
rs748292175 Chr.19: 6183109-6183109 on Build GRCh38
rs73923464 Chr.19: 6183014-6183014 on Build GRCh38
rs771291867 Chr.19: 6182839-6182839 on Build GRCh38
rs891895988 Chr.19: 6182733-6182733 on Build GRCh38
rs927474988 Chr.19: 6182692-6182692 on Build GRCh38
rs746062729 Chr.19: 6182931-6182931 on Build GRCh38
rs774271618 Chr.19: 6182874-6182874 on Build GRCh38
rs763283203 Chr.19: 6182833-6182833 on Build GRCh38
rs377036111 Chr.19: 6182980-6182980 on Build GRCh38
rs752245997 Chr.19: 6183055-6183055 on Build GRCh38
rs1043857966 Chr.19: 6182800-6182800 on Build GRCh38
rs541714967 Chr.19: 6182726-6182726 on Build GRCh38
rs748662574 Chr.19: 6182805-6182805 on Build GRCh38
rs766652304 Chr.19: 6182731-6182731 on Build GRCh38
rs371165248 Chr.19: 6182852-6182852 on Build GRCh38
rs776604962 Chr.19: 6182947-6182948 on Build GRCh38
rs780871830 Chr.19: 6182802-6182802 on Build GRCh38
rs759434629 Chr.19: 6182875-6182875 on Build GRCh38
rs539844379 Chr.19: 6182928-6182928 on Build GRCh38
rs746663021 Chr.19: 6182775-6182775 on Build GRCh38
rs374130121 Chr.19: 6182826-6182826 on Build GRCh38
rs201845150 Chr.19: 6183054-6183054 on Build GRCh38
rs764236474 Chr.19: 6182954-6182954 on Build GRCh38
rs149469974 Chr.19: 6182882-6182882 on Build GRCh38
rs760244436 Chr.19: 6183119-6183119 on Build GRCh38
rs778739485 Chr.19: 6182750-6182750 on Build GRCh38
rs759542744 Chr.19: 6183112-6183112 on Build GRCh38
rs754726964 Chr.19: 6182888-6182888 on Build GRCh38
rs745633287 Chr.19: 6183052-6183052 on Build GRCh38
rs144132436 Chr.19: 6182885-6182885 on Build GRCh38
rs775627451 Chr.19: 6182940-6182940 on Build GRCh38
rs776819900 Chr.19: 6183039-6183039 on Build GRCh38
rs766486498 Chr.19: 6183046-6183046 on Build GRCh38
rs879642988 Chr.19: 6182723-6182723 on Build GRCh38
rs781413421 Chr.19: 6183104-6183104 on Build GRCh38
rs775509673 Chr.19: 6183033-6183033 on Build GRCh38
rs756627200 Chr.19: 6182900-6182900 on Build GRCh38
rs913969106 Chr.19: 6182767-6182767 on Build GRCh38
rs548746467 Chr.19: 6182899-6182899 on Build GRCh38
rs759344024 Chr.19: 6183053-6183053 on Build GRCh38
rs752406063 Chr.19: 6183004-6183004 on Build GRCh38
rs779245840 Chr.19: 6183021-6183021 on Build GRCh38
rs747175659 Chr.19: 6183081-6183081 on Build GRCh38
rs866017688 Chr.19: 6182856-6182856 on Build GRCh38
rs763005478 Chr.19: 6182961-6182961 on Build GRCh38
rs139677949 Chr.19: 6183146-6183146 on Build GRCh38
rs147009605 Chr.19: 6183044-6183044 on Build GRCh38
rs758652190 Chr.19: 6183070-6183070 on Build GRCh38
rs759549699 Chr.19: 6182740-6182740 on Build GRCh38
rs773619641 Chr.19: 6182828-6182828 on Build GRCh38
rs865778112 Chr.19: 6182725-6182725 on Build GRCh38
rs762259007 Chr.19: 6182711-6182711 on Build GRCh38
rs767130162 Chr.19: 6182989-6182989 on Build GRCh38
rs760427454 Chr.19: 6183058-6183058 on Build GRCh38
rs758112150 Chr.19: 6182754-6182754 on Build GRCh38
rs779794020 Chr.19: 6182771-6182771 on Build GRCh38
rs938189062 Chr.19: 6183094-6183094 on Build GRCh38
rs3810456 Chr.19: 6183017-6183017 on Build GRCh38
rs766254316 Chr.19: 6182977-6182977 on Build GRCh38
rs564637392 Chr.19: 6182739-6182739 on Build GRCh38
rs772981239 Chr.19: 6182955-6182955 on Build GRCh38
rs867340883 Chr.19: 6183057-6183057 on Build GRCh38
rs770681838 Chr.19: 6182876-6182887 on Build GRCh38
rs777576026 Chr.19: 6182789-6182789 on Build GRCh38
rs746730255 Chr.19: 6182867-6182869 on Build GRCh38
rs369153834 Chr.19: 6182874-6182876 on Build GRCh38
rs774360873 Chr.19: 6183111-6183111 on Build GRCh38
rs758197998 Chr.19: 6182883-6182883 on Build GRCh38
rs36111004 Chr.19: 6183151-6183152 on Build GRCh38
rs112361030 Chr.19: 6183037-6183037 on Build GRCh38
rs768963353 Chr.19: 6183084-6183084 on Build GRCh38
rs372123644 Chr.19: 6183065-6183065 on Build GRCh38
rs956019039 Chr.19: 6182958-6182958 on Build GRCh38
rs867826032 Chr.19: 6183114-6183114 on Build GRCh38
rs142334228 Chr.19: 6183147-6183147 on Build GRCh38
rs369436164 Chr.19: 6182751-6182751 on Build GRCh38
rs760614538 Chr.19: 6182944-6182944 on Build GRCh38
rs779335865 Chr.19: 6183067-6183067 on Build GRCh38
rs763484624 Chr.19: 6183064-6183064 on Build GRCh38
rs1050085481 Chr.19: 6183066-6183066 on Build GRCh38
rs768292877 Chr.19: 6182710-6182710 on Build GRCh38
rs147652076 Chr.19: 6183124-6183124 on Build GRCh38
rs955561375 Chr.19: 6182753-6182753 on Build GRCh38
rs572752210 Chr.19: 6182714-6182714 on Build GRCh38
rs746577188 Chr.19: 6182706-6182706 on Build GRCh38
rs780717732 Chr.19: 6182889-6182889 on Build GRCh38
rs998083695 Chr.19: 6182843-6182843 on Build GRCh38

View Full Product Details