Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRP19
Entrez Gene ID
6728
Gene Name
signal recognition particle 19
Gene Aliases
-
Chromosome Location
Chr.5: 112861188 - 112893079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204193.1 NP_001191122.1 1 - 2 231 118
NM_001204194.1 NP_001191123.1 1 - 2 231 118
NM_001204199.1 NP_001191128.1 1 - 2 231 118
NM_003135.2 NP_003126.1 1 - 2 231 118
GenBank mRNA AK311803.1 - 1 - 2 139 118
BC000186.1 - 1 - 2 123 118
BC010947.1 - 1 - 2 115 118
BC017830.1 - 1 - 2 114 118
BY798924.2 - 1 - 2 243 118
DA930867.1 - 1 - 2 114 118
DN915735.1 - 1 - 2 96 118
X12791.1 - 1 - 2 123 118

View Full Product Details