Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11576631-11577134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP5
Entrez Gene ID
54
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
UniGene
Hs.1211
Gene Chromosome Location
Chr.19: 11574660-11579008 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Gene Symbol
ZNF627
Entrez Gene ID
199692
Gene Name
zinc finger protein 627
Gene Aliases
-
UniGene
Hs.526665
Gene Chromosome Location
Chr.19: 11575255-11619159 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4811356 c.353C>T p.S118F Chr.19: 11576752-11576752 on Build GRCh38
6890667 c.267C>T p.T89T Chr.19: 11576838-11576838 on Build GRCh38
6869645 c.294C>T p.S98S Chr.19: 11576811-11576811 on Build GRCh38
3796611 c.353C>T p.S118F Chr.19: 11576752-11576752 on Build GRCh38
4811879 c.254G>C p.R85T Chr.19: 11577064-11577064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923146284 Chr.19: 11576926-11576926 on Build GRCh38
rs752275809 Chr.19: 11576781-11576781 on Build GRCh38
rs773553601 Chr.19: 11576938-11576938 on Build GRCh38
rs113937776 Chr.19: 11576652-11576652 on Build GRCh38
rs562918956 Chr.19: 11576701-11576701 on Build GRCh38
rs758032450 Chr.19: 11576999-11576999 on Build GRCh38
rs148219285 Chr.19: 11576789-11576789 on Build GRCh38
rs773606741 Chr.19: 11576836-11576836 on Build GRCh38
rs777140546 Chr.19: 11576815-11576815 on Build GRCh38
rs577018427 Chr.19: 11576939-11576939 on Build GRCh38
rs756487730 Chr.19: 11577003-11577003 on Build GRCh38
rs745927602 Chr.19: 11577028-11577028 on Build GRCh38
rs756938825 Chr.19: 11576982-11576982 on Build GRCh38
rs781050795 Chr.19: 11576780-11576780 on Build GRCh38
rs574317748 Chr.19: 11576979-11576979 on Build GRCh38
rs566300441 Chr.19: 11576894-11576894 on Build GRCh38
rs762413067 Chr.19: 11576757-11576757 on Build GRCh38
rs774986467 Chr.19: 11576705-11576705 on Build GRCh38
rs759339133 Chr.19: 11576950-11576950 on Build GRCh38
rs1007317128 Chr.19: 11577089-11577089 on Build GRCh38
rs62638747 Chr.19: 11577093-11577093 on Build GRCh38
rs775396112 Chr.19: 11577051-11577051 on Build GRCh38
rs201094805 Chr.19: 11577035-11577035 on Build GRCh38
rs755949462 Chr.19: 11576882-11576882 on Build GRCh38
rs767582437 Chr.19: 11576863-11576864 on Build GRCh38
rs370365286 Chr.19: 11576678-11576678 on Build GRCh38
rs539880925 Chr.19: 11576910-11576910 on Build GRCh38
rs200885075 Chr.19: 11576863-11576863 on Build GRCh38
rs568576678 Chr.19: 11576828-11576828 on Build GRCh38
rs978287680 Chr.19: 11576878-11576878 on Build GRCh38
rs555490099 Chr.19: 11576699-11576699 on Build GRCh38
rs558569018 Chr.19: 11576917-11576917 on Build GRCh38
rs541543818 Chr.19: 11576689-11576689 on Build GRCh38
rs747619825 Chr.19: 11576736-11576736 on Build GRCh38
rs763938381 Chr.19: 11576975-11576975 on Build GRCh38
rs774482395 Chr.19: 11577026-11577026 on Build GRCh38
rs911315743 Chr.19: 11576897-11576897 on Build GRCh38
rs2229530 Chr.19: 11576790-11576790 on Build GRCh38
rs144703899 Chr.19: 11577009-11577010 on Build GRCh38
rs764612996 Chr.19: 11576983-11576983 on Build GRCh38
rs770348727 Chr.19: 11576936-11576936 on Build GRCh38
rs781762903 Chr.19: 11577001-11577001 on Build GRCh38
rs774517154 Chr.19: 11576942-11576942 on Build GRCh38
rs749972758 Chr.19: 11576994-11576994 on Build GRCh38
rs756448158 Chr.19: 11576732-11576733 on Build GRCh38
rs752508731 Chr.19: 11576880-11576880 on Build GRCh38
rs965741395 Chr.19: 11577096-11577096 on Build GRCh38
rs766538784 Chr.19: 11576855-11576855 on Build GRCh38
rs528748445 Chr.19: 11577080-11577080 on Build GRCh38
rs563929774 Chr.19: 11577069-11577069 on Build GRCh38
rs774097590 Chr.19: 11576861-11576861 on Build GRCh38
rs202233676 Chr.19: 11577073-11577073 on Build GRCh38
rs534619992 Chr.19: 11576941-11576941 on Build GRCh38
rs1046178477 Chr.19: 11577008-11577008 on Build GRCh38
rs748490504 Chr.19: 11576931-11576931 on Build GRCh38
rs746331116 Chr.19: 11576690-11576690 on Build GRCh38
rs768845671 Chr.19: 11576814-11576814 on Build GRCh38
rs73496603 Chr.19: 11577002-11577002 on Build GRCh38
rs768373843 Chr.19: 11577066-11577066 on Build GRCh38
rs563079303 Chr.19: 11577020-11577020 on Build GRCh38
rs767255534 Chr.19: 11576864-11576864 on Build GRCh38
rs966848108 Chr.19: 11576874-11576874 on Build GRCh38
rs748777525 Chr.19: 11576800-11576800 on Build GRCh38
rs769135184 Chr.19: 11576707-11576707 on Build GRCh38
rs781650240 Chr.19: 11576930-11576930 on Build GRCh38
rs148415735 Chr.19: 11577021-11577021 on Build GRCh38
rs750563102 Chr.19: 11576722-11576722 on Build GRCh38
rs75921937 Chr.19: 11577102-11577102 on Build GRCh38
rs923204953 Chr.19: 11576697-11576697 on Build GRCh38
rs776116121 Chr.19: 11576703-11576703 on Build GRCh38
rs74705563 Chr.19: 11577104-11577104 on Build GRCh38
rs749753832 Chr.19: 11576744-11576744 on Build GRCh38
rs772661435 Chr.19: 11576909-11576909 on Build GRCh38
rs34375794 Chr.19: 11576829-11576829 on Build GRCh38
rs760623185 Chr.19: 11577052-11577052 on Build GRCh38
rs1025173451 Chr.19: 11576873-11576873 on Build GRCh38
rs763172932 Chr.19: 11576765-11576765 on Build GRCh38
rs387906668 Chr.19: 11576839-11576839 on Build GRCh38
rs768647004 Chr.19: 11576678-11576679 on Build GRCh38
rs141651325 Chr.19: 11576806-11576806 on Build GRCh38
rs767418596 Chr.19: 11576954-11576954 on Build GRCh38
rs553100380 Chr.19: 11576956-11576956 on Build GRCh38
rs753558356 Chr.19: 11576895-11576895 on Build GRCh38
rs776981075 Chr.19: 11576713-11576713 on Build GRCh38
rs755541324 Chr.19: 11576928-11576928 on Build GRCh38
rs200250079 Chr.19: 11576858-11576858 on Build GRCh38
rs899103554 Chr.19: 11576700-11576700 on Build GRCh38
rs764923415 Chr.19: 11577083-11577083 on Build GRCh38
rs762935882 Chr.19: 11576846-11576846 on Build GRCh38
rs765343526 Chr.19: 11576720-11576720 on Build GRCh38
rs113122216 Chr.19: 11576788-11576788 on Build GRCh38
rs199580546 Chr.19: 11576719-11576719 on Build GRCh38
rs760461677 Chr.19: 11576968-11576968 on Build GRCh38
rs762537777 Chr.19: 11577107-11577107 on Build GRCh38
rs751580952 Chr.19: 11576772-11576772 on Build GRCh38
rs144419140 Chr.19: 11576812-11576812 on Build GRCh38
rs754714567 Chr.19: 11576773-11576773 on Build GRCh38

View Full Product Details