Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11576973-11577473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP5
Entrez Gene ID
54
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
UniGene
Hs.1211
Gene Chromosome Location
Chr.19: 11574660-11579008 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Gene Symbol
ZNF627
Entrez Gene ID
199692
Gene Name
zinc finger protein 627
Gene Aliases
-
UniGene
Hs.526665
Gene Chromosome Location
Chr.19: 11575255-11619159 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4811879 c.254G>C p.R85T Chr.19: 11577064-11577064 on Build GRCh38
5460790 c.79C>T p.R27C Chr.19: 11577239-11577239 on Build GRCh38
6884992 c.68C>A p.T23N Chr.19: 11577250-11577250 on Build GRCh38
39798 c.102G>A p.W34* Chr.19: 11577216-11577216 on Build GRCh38
3528589 c.133G>T p.A45S Chr.19: 11577185-11577185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765600744 Chr.19: 11577327-11577327 on Build GRCh38
rs202233676 Chr.19: 11577073-11577073 on Build GRCh38
rs963539484 Chr.19: 11577446-11577446 on Build GRCh38
rs556637113 Chr.19: 11577304-11577304 on Build GRCh38
rs772862057 Chr.19: 11577199-11577199 on Build GRCh38
rs371591030 Chr.19: 11577152-11577152 on Build GRCh38
rs142372538 Chr.19: 11577170-11577170 on Build GRCh38
rs1046178477 Chr.19: 11577008-11577008 on Build GRCh38
rs369579418 Chr.19: 11577239-11577239 on Build GRCh38
rs758032450 Chr.19: 11576999-11576999 on Build GRCh38
rs200284614 Chr.19: 11577310-11577310 on Build GRCh38
rs755025237 Chr.19: 11577363-11577363 on Build GRCh38
rs1016120863 Chr.19: 11577422-11577422 on Build GRCh38
rs769534166 Chr.19: 11577196-11577196 on Build GRCh38
rs768659695 Chr.19: 11577280-11577280 on Build GRCh38
rs73496603 Chr.19: 11577002-11577002 on Build GRCh38
rs768373843 Chr.19: 11577066-11577066 on Build GRCh38
rs756487730 Chr.19: 11577003-11577003 on Build GRCh38
rs746565633 Chr.19: 11577403-11577403 on Build GRCh38
rs747070328 Chr.19: 11577279-11577279 on Build GRCh38
rs563079303 Chr.19: 11577020-11577020 on Build GRCh38
rs765947561 Chr.19: 11577227-11577227 on Build GRCh38
rs745927602 Chr.19: 11577028-11577028 on Build GRCh38
rs564079812 Chr.19: 11577348-11577348 on Build GRCh38
rs777141156 Chr.19: 11577315-11577315 on Build GRCh38
rs746137618 Chr.19: 11577172-11577172 on Build GRCh38
rs754367027 Chr.19: 11577254-11577254 on Build GRCh38
rs376043348 Chr.19: 11577238-11577238 on Build GRCh38
rs949066797 Chr.19: 11577276-11577276 on Build GRCh38
rs935506217 Chr.19: 11577300-11577300 on Build GRCh38
rs2071485 Chr.19: 11577385-11577385 on Build GRCh38
rs929091337 Chr.19: 11577204-11577204 on Build GRCh38
rs148415735 Chr.19: 11577021-11577021 on Build GRCh38
rs1007317128 Chr.19: 11577089-11577089 on Build GRCh38
rs939236666 Chr.19: 11577250-11577250 on Build GRCh38
rs35621512 Chr.19: 11577212-11577213 on Build GRCh38
rs75921937 Chr.19: 11577102-11577102 on Build GRCh38
rs762234725 Chr.19: 11577321-11577321 on Build GRCh38
rs201047202 Chr.19: 11577271-11577271 on Build GRCh38
rs62638747 Chr.19: 11577093-11577093 on Build GRCh38
rs775396112 Chr.19: 11577051-11577051 on Build GRCh38
rs780061234 Chr.19: 11577278-11577278 on Build GRCh38
rs74705563 Chr.19: 11577104-11577104 on Build GRCh38
rs758812133 Chr.19: 11577341-11577341 on Build GRCh38
rs201094805 Chr.19: 11577035-11577035 on Build GRCh38
rs766000656 Chr.19: 11577126-11577126 on Build GRCh38
rs189825826 Chr.19: 11577169-11577169 on Build GRCh38
rs746909423 Chr.19: 11577184-11577184 on Build GRCh38
rs998103110 Chr.19: 11577316-11577316 on Build GRCh38
rs941984729 Chr.19: 11577218-11577218 on Build GRCh38
rs978320219 Chr.19: 11577133-11577133 on Build GRCh38
rs774184401 Chr.19: 11577307-11577307 on Build GRCh38
rs760623185 Chr.19: 11577052-11577052 on Build GRCh38
rs368967598 Chr.19: 11577364-11577364 on Build GRCh38
rs150582430 Chr.19: 11577186-11577186 on Build GRCh38
rs774482395 Chr.19: 11577026-11577026 on Build GRCh38
rs766997062 Chr.19: 11577312-11577312 on Build GRCh38
rs748048119 Chr.19: 11577296-11577296 on Build GRCh38
rs529545521 Chr.19: 11577155-11577155 on Build GRCh38
rs369804864 Chr.19: 11577187-11577187 on Build GRCh38
rs922892618 Chr.19: 11577151-11577151 on Build GRCh38
rs144703899 Chr.19: 11577009-11577010 on Build GRCh38
rs765370739 Chr.19: 11577127-11577127 on Build GRCh38
rs200380116 Chr.19: 11577228-11577228 on Build GRCh38
rs757203906 Chr.19: 11577202-11577202 on Build GRCh38
rs139654624 Chr.19: 11577288-11577288 on Build GRCh38
rs781762903 Chr.19: 11577001-11577001 on Build GRCh38
rs202217114 Chr.19: 11577258-11577258 on Build GRCh38
rs182776939 Chr.19: 11577301-11577301 on Build GRCh38
rs780116316 Chr.19: 11577182-11577182 on Build GRCh38
rs757630659 Chr.19: 11577257-11577257 on Build GRCh38
rs764923415 Chr.19: 11577083-11577083 on Build GRCh38
rs886043536 Chr.19: 11577191-11577191 on Build GRCh38
rs545590300 Chr.19: 11577334-11577334 on Build GRCh38
rs779737292 Chr.19: 11577432-11577432 on Build GRCh38
rs201184983 Chr.19: 11577181-11577181 on Build GRCh38
rs777947402 Chr.19: 11577135-11577135 on Build GRCh38
rs955652311 Chr.19: 11577410-11577410 on Build GRCh38
rs762537777 Chr.19: 11577107-11577107 on Build GRCh38
rs772911511 Chr.19: 11577303-11577303 on Build GRCh38
rs1029768838 Chr.19: 11577409-11577409 on Build GRCh38
rs965741395 Chr.19: 11577096-11577096 on Build GRCh38
rs528748445 Chr.19: 11577080-11577080 on Build GRCh38
rs563929774 Chr.19: 11577069-11577069 on Build GRCh38

View Full Product Details