The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000826.3 | NP_000817.2 | 3 - 4 | 932 | 71 |
NM_001083619.1 | NP_001077088.1 | 3 - 4 | 932 | 71 | |
NM_001083620.1 | NP_001077089.1 | 3 - 4 | 443 | 71 | |
XM_017008113.1 | XP_016863602.1 | 3 - 4 | 683 | 71 | |
XM_017008114.1 | XP_016863603.1 | 3 - 4 | 841 | 71 | |
XM_017008115.1 | XP_016863604.1 | 3 - 4 | 404 | 71 | |
XM_017008116.1 | XP_016863605.1 | 3 - 4 | 628 | 71 | |
XM_017008117.1 | XP_016863606.1 | 3 - 4 | 686 | 71 | |
GenBank mRNA | AB209567.1 | - | 3 - 4 | 761 | 71 |
AK294458.1 | - | 3 - 4 | 609 | 71 | |
AK308775.1 | - | 3 - 4 | 686 | 71 | |
AK309067.1 | - | 3 - 4 | 456 | 71 | |
AL832605.3 | - | 3 - 4 | 792 | 71 | |
BC010574.1 | - | 3 - 4 | 752 | 71 | |
BC028736.1 | - | 3 - 4 | 421 | 71 | |
BP202686.1 | - | 3 - 4 | 444 | 71 | |
L20814.1 | - | 3 - 4 | 633 | 71 |