Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15379607-15380094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
UniGene
Hs.594496
Gene Chromosome Location
Chr.19: 15353521-15379815 on Build GRCh38
Transcripts
NM_005858.3 NM_001291478.1 NM_014371.3 NR_111971.1 XM_005259854.3

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
UniGene
Hs.399800
Gene Chromosome Location
Chr.19: 15380048-15419121 on Build GRCh38
Transcripts
NM_005858.3 NM_001291478.1 NM_014371.3 NR_111971.1 XM_005259854.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3959645 c.10G>T p.G4C Chr.19: 15379722-15379722 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750215358 Chr.19: 15379687-15379687 on Build GRCh38
rs567963490 Chr.19: 15379836-15379836 on Build GRCh38
rs1046402927 Chr.19: 15379807-15379807 on Build GRCh38
rs748934580 Chr.19: 15379770-15379770 on Build GRCh38
rs553187478 Chr.19: 15380006-15380006 on Build GRCh38
rs748639941 Chr.19: 15379717-15379717 on Build GRCh38
rs902297640 Chr.19: 15379824-15379824 on Build GRCh38
rs11879108 Chr.19: 15380012-15380012 on Build GRCh38
rs1017594858 Chr.19: 15379852-15379852 on Build GRCh38
rs556743067 Chr.19: 15379875-15379875 on Build GRCh38
rs533328820 Chr.19: 15379697-15379697 on Build GRCh38
rs928076681 Chr.19: 15379988-15379988 on Build GRCh38
rs919545150 Chr.19: 15379988-15380004 on Build GRCh38
rs945346460 Chr.19: 15379635-15379635 on Build GRCh38
rs761452803 Chr.19: 15379741-15379741 on Build GRCh38
rs199553362 Chr.19: 15379740-15379740 on Build GRCh38
rs997566289 Chr.19: 15379886-15379886 on Build GRCh38
rs375106546 Chr.19: 15379790-15379790 on Build GRCh38
rs754029347 Chr.19: 15379757-15379757 on Build GRCh38
rs997512326 Chr.19: 15380072-15380072 on Build GRCh38
rs767384506 Chr.19: 15379685-15379685 on Build GRCh38
rs373595393 Chr.19: 15379946-15379946 on Build GRCh38
rs911077899 Chr.19: 15379968-15379968 on Build GRCh38
rs774105303 Chr.19: 15379771-15379771 on Build GRCh38
rs1016871278 Chr.19: 15379912-15379912 on Build GRCh38
rs910116086 Chr.19: 15380007-15380011 on Build GRCh38
rs374460362 Chr.19: 15379758-15379758 on Build GRCh38
rs778250051 Chr.19: 15379766-15379766 on Build GRCh38
rs897334178 Chr.19: 15380057-15380057 on Build GRCh38
rs773213686 Chr.19: 15379780-15379780 on Build GRCh38
rs901845628 Chr.19: 15379864-15379864 on Build GRCh38
rs767542180 Chr.19: 15379743-15379743 on Build GRCh38
rs778290852 Chr.19: 15379726-15379726 on Build GRCh38
rs573112236 Chr.19: 15380055-15380055 on Build GRCh38
rs936449332 Chr.19: 15379945-15379945 on Build GRCh38
rs998351048 Chr.19: 15380063-15380064 on Build GRCh38
rs751455889 Chr.19: 15379680-15379680 on Build GRCh38
rs537055942 Chr.19: 15379851-15379851 on Build GRCh38
rs776280853 Chr.19: 15379722-15379722 on Build GRCh38
rs368699019 Chr.19: 15379763-15379763 on Build GRCh38
rs1002471893 Chr.19: 15379668-15379668 on Build GRCh38
rs929523358 Chr.19: 15379634-15379634 on Build GRCh38
rs1026390733 Chr.19: 15379891-15379891 on Build GRCh38
rs370021454 Chr.19: 15379660-15379660 on Build GRCh38
rs370823822 Chr.19: 15379783-15379783 on Build GRCh38
rs747650349 Chr.19: 15379776-15379776 on Build GRCh38
rs775142597 Chr.19: 15379733-15379733 on Build GRCh38
rs949101434 Chr.19: 15379819-15379819 on Build GRCh38
rs959586441 Chr.19: 15379915-15379915 on Build GRCh38
rs770535269 Chr.19: 15379670-15379671 on Build GRCh38
rs57295859 Chr.19: 15379674-15379674 on Build GRCh38
rs938018105 Chr.19: 15379995-15379995 on Build GRCh38
rs780407833 Chr.19: 15379692-15379692 on Build GRCh38
rs974052934 Chr.19: 15379984-15380000 on Build GRCh38
rs1053870935 Chr.19: 15379629-15379629 on Build GRCh38
rs1042173068 Chr.19: 15379706-15379706 on Build GRCh38
rs889697439 Chr.19: 15379834-15379834 on Build GRCh38
rs1007003126 Chr.19: 15379926-15379926 on Build GRCh38
rs941632806 Chr.19: 15380045-15380045 on Build GRCh38
rs778867868 Chr.19: 15379704-15379704 on Build GRCh38
rs555680417 Chr.19: 15379804-15379804 on Build GRCh38
rs954066857 Chr.19: 15379761-15379761 on Build GRCh38
rs774737081 Chr.19: 15379659-15379659 on Build GRCh38
rs919439609 Chr.19: 15379626-15379626 on Build GRCh38
rs771353196 Chr.19: 15379732-15379732 on Build GRCh38
rs923696509 Chr.19: 15379941-15379941 on Build GRCh38
rs781433397 Chr.19: 15379769-15379769 on Build GRCh38
rs537820331 Chr.19: 15380006-15380007 on Build GRCh38
rs771366615 Chr.19: 15379657-15379657 on Build GRCh38
rs767748821 Chr.19: 15379876-15379876 on Build GRCh38
rs747362693 Chr.19: 15379728-15379728 on Build GRCh38
rs1040988962 Chr.19: 15379987-15379987 on Build GRCh38
rs754072342 Chr.19: 15379693-15379693 on Build GRCh38
rs927993748 Chr.19: 15379817-15379817 on Build GRCh38
rs760793998 Chr.19: 15379977-15379977 on Build GRCh38
rs888729552 Chr.19: 15379837-15379837 on Build GRCh38
rs925903016 Chr.19: 15379774-15379774 on Build GRCh38
rs1037183267 Chr.19: 15379870-15379870 on Build GRCh38
rs1046457773 Chr.19: 15379641-15379641 on Build GRCh38
rs748906847 Chr.19: 15379736-15379736 on Build GRCh38
rs535428626 Chr.19: 15379678-15379678 on Build GRCh38
rs917549910 Chr.19: 15379920-15379920 on Build GRCh38
rs1006826557 Chr.19: 15379846-15379846 on Build GRCh38
rs913971063 Chr.19: 15379802-15379802 on Build GRCh38
rs896884495 Chr.19: 15379890-15379890 on Build GRCh38
rs1042130291 Chr.19: 15379822-15379822 on Build GRCh38
rs879061518 Chr.19: 15379745-15379745 on Build GRCh38
rs768168269 Chr.19: 15379737-15379737 on Build GRCh38
rs1050858618 Chr.19: 15380066-15380066 on Build GRCh38
rs766088287 Chr.19: 15379751-15379751 on Build GRCh38
rs978201589 Chr.19: 15379938-15379938 on Build GRCh38
rs1054052725 Chr.19: 15379828-15379828 on Build GRCh38
rs990431105 Chr.19: 15379796-15379796 on Build GRCh38
rs888622982 Chr.19: 15379679-15379679 on Build GRCh38
rs957298576 Chr.19: 15379951-15379951 on Build GRCh38
rs758520628 Chr.19: 15379765-15379765 on Build GRCh38
rs771773322 Chr.19: 15379779-15379779 on Build GRCh38
rs576890106 Chr.19: 15379897-15379897 on Build GRCh38
rs545940271 Chr.19: 15379959-15379959 on Build GRCh38
rs866372438 Chr.19: 15380053-15380053 on Build GRCh38
rs747520628 Chr.19: 15379768-15379768 on Build GRCh38
rs566754006 Chr.19: 15379760-15379760 on Build GRCh38
rs945462455 Chr.19: 15379805-15379805 on Build GRCh38
rs760540082 Chr.19: 15379747-15379747 on Build GRCh38
rs974150009 Chr.19: 15379989-15379989 on Build GRCh38
rs550750742 Chr.19: 15379631-15379631 on Build GRCh38
rs1037131029 Chr.19: 15380048-15380048 on Build GRCh38
rs936517298 Chr.19: 15379827-15379827 on Build GRCh38
rs970360592 Chr.19: 15379929-15379929 on Build GRCh38
rs1006368706 Chr.19: 15379900-15379900 on Build GRCh38
rs938287712 Chr.19: 15379795-15379795 on Build GRCh38
rs1026858514 Chr.19: 15379778-15379778 on Build GRCh38
rs1053598536 Chr.19: 15379955-15379955 on Build GRCh38
rs991049147 Chr.19: 15379917-15379917 on Build GRCh38
rs761682850 Chr.19: 15379684-15379684 on Build GRCh38

View Full Product Details