Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17320473-17320965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDA1
Entrez Gene ID
79016
Gene Name
DET1 and DDB1 associated 1
Gene Aliases
C19orf58, PCIA1
UniGene
Hs.466154
Gene Chromosome Location
Chr.19: 17309528-17323301 on Build GRCh38
Transcripts
NM_024050.5

SNP Detail

SNP ID Chromosome Location
rs894419732 Chr.19: 17320943-17320943 on Build GRCh38
rs1023816750 Chr.19: 17320915-17320915 on Build GRCh38
rs1012795518 Chr.19: 17320912-17320912 on Build GRCh38
rs565901529 Chr.19: 17320881-17320881 on Build GRCh38
rs567151965 Chr.19: 17320789-17320789 on Build GRCh38
rs902210356 Chr.19: 17320879-17320879 on Build GRCh38
rs777136235 Chr.19: 17320829-17320829 on Build GRCh38
rs572053740 Chr.19: 17320860-17320860 on Build GRCh38
rs1047252795 Chr.19: 17320861-17320861 on Build GRCh38
rs761564042 Chr.19: 17320550-17320550 on Build GRCh38
rs1005427781 Chr.19: 17320661-17320661 on Build GRCh38
rs918636413 Chr.19: 17320534-17320534 on Build GRCh38
rs971448546 Chr.19: 17320730-17320730 on Build GRCh38
rs936237341 Chr.19: 17320577-17320577 on Build GRCh38
rs991085457 Chr.19: 17320687-17320687 on Build GRCh38
rs924623506 Chr.19: 17320555-17320555 on Build GRCh38
rs962940138 Chr.19: 17320721-17320721 on Build GRCh38
rs1024376290 Chr.19: 17320698-17320698 on Build GRCh38
rs10403469 Chr.19: 17320629-17320629 on Build GRCh38
rs971373588 Chr.19: 17320551-17320551 on Build GRCh38
rs540915845 Chr.19: 17320884-17320884 on Build GRCh38
rs930073923 Chr.19: 17320537-17320537 on Build GRCh38
rs113607938 Chr.19: 17320640-17320640 on Build GRCh38
rs1043396211 Chr.19: 17320869-17320869 on Build GRCh38
rs11666036 Chr.19: 17320686-17320686 on Build GRCh38
rs1046705253 Chr.19: 17320631-17320631 on Build GRCh38
rs1031637076 Chr.19: 17320786-17320786 on Build GRCh38
rs866782311 Chr.19: 17320817-17320817 on Build GRCh38
rs900391113 Chr.19: 17320827-17320827 on Build GRCh38
rs529649260 Chr.19: 17320790-17320790 on Build GRCh38
rs1012783278 Chr.19: 17320697-17320697 on Build GRCh38
rs777952752 Chr.19: 17320812-17320812 on Build GRCh38
rs974015370 Chr.19: 17320780-17320780 on Build GRCh38
rs941055087 Chr.19: 17320885-17320885 on Build GRCh38
rs977515246 Chr.19: 17320740-17320740 on Build GRCh38
rs547640103 Chr.19: 17320758-17320758 on Build GRCh38
rs543555762 Chr.19: 17320591-17320591 on Build GRCh38
rs552339273 Chr.19: 17320808-17320808 on Build GRCh38
rs527597771 Chr.19: 17320714-17320714 on Build GRCh38
rs910830162 Chr.19: 17320800-17320800 on Build GRCh38
rs1039394349 Chr.19: 17320608-17320608 on Build GRCh38
rs563793988 Chr.19: 17320619-17320619 on Build GRCh38
rs1035183144 Chr.19: 17320671-17320671 on Build GRCh38
rs71338626 Chr.19: 17320623-17320623 on Build GRCh38
rs890308239 Chr.19: 17320567-17320567 on Build GRCh38
rs374548804 Chr.19: 17320916-17320916 on Build GRCh38
rs555199157 Chr.19: 17320939-17320939 on Build GRCh38
rs910682448 Chr.19: 17320592-17320592 on Build GRCh38
rs923462020 Chr.19: 17320822-17320822 on Build GRCh38
rs1038274947 Chr.19: 17320941-17320941 on Build GRCh38
rs943580029 Chr.19: 17320614-17320614 on Build GRCh38
rs991566480 Chr.19: 17320526-17320526 on Build GRCh38
rs896240403 Chr.19: 17320901-17320901 on Build GRCh38
rs944575143 Chr.19: 17320803-17320803 on Build GRCh38
rs76045424 Chr.19: 17320553-17320553 on Build GRCh38
rs71338627 Chr.19: 17320791-17320791 on Build GRCh38
rs999300395 Chr.19: 17320645-17320645 on Build GRCh38
rs751624486 Chr.19: 17320813-17320813 on Build GRCh38
rs750040338 Chr.19: 17320576-17320576 on Build GRCh38
rs541132048 Chr.19: 17320792-17320792 on Build GRCh38
rs879410257 Chr.19: 17320766-17320766 on Build GRCh38
rs538876720 Chr.19: 17320794-17320794 on Build GRCh38
rs1024428338 Chr.19: 17320541-17320541 on Build GRCh38
rs1017302585 Chr.19: 17320666-17320666 on Build GRCh38

View Full Product Details