Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17323504-17323998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO8
Entrez Gene ID
57719
Gene Name
anoctamin 8
Gene Aliases
KIAA1623, TMEM16H
UniGene
Hs.590990
Gene Chromosome Location
Chr.19: 17323223-17334834 on Build GRCh38
Transcripts
NM_020959.2 XM_017027048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601180 c.3638C>T p.A1213V Chr.19: 17323578-17323578 on Build GRCh38
5957748 c.3446G>T p.W1149L Chr.19: 17323770-17323770 on Build GRCh38
3530336 c.3624C>T p.P1208P Chr.19: 17323592-17323592 on Build GRCh38
5680063 c.3567G>A p.P1189P Chr.19: 17323649-17323649 on Build GRCh38
5792462 c.3694C>T p.H1232Y Chr.19: 17323522-17323522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759635422 Chr.19: 17323549-17323573 on Build GRCh38
rs937475858 Chr.19: 17323842-17323842 on Build GRCh38
rs866221063 Chr.19: 17323678-17323678 on Build GRCh38
rs1034523765 Chr.19: 17323558-17323558 on Build GRCh38
rs912927157 Chr.19: 17323781-17323781 on Build GRCh38
rs763084675 Chr.19: 17323576-17323577 on Build GRCh38
rs751292346 Chr.19: 17323664-17323664 on Build GRCh38
rs1006449694 Chr.19: 17323928-17323928 on Build GRCh38
rs759542955 Chr.19: 17323619-17323619 on Build GRCh38
rs764216956 Chr.19: 17323604-17323618 on Build GRCh38
rs765516826 Chr.19: 17323549-17323550 on Build GRCh38
rs1030384216 Chr.19: 17323949-17323949 on Build GRCh38
rs763723821 Chr.19: 17323642-17323642 on Build GRCh38
rs781546048 Chr.19: 17323531-17323531 on Build GRCh38
rs770763087 Chr.19: 17323598-17323598 on Build GRCh38
rs768976940 Chr.19: 17323589-17323589 on Build GRCh38
rs955154374 Chr.19: 17323687-17323687 on Build GRCh38
rs756097878 Chr.19: 17323560-17323560 on Build GRCh38
rs552570603 Chr.19: 17323923-17323923 on Build GRCh38
rs997930166 Chr.19: 17323937-17323937 on Build GRCh38
rs12977101 Chr.19: 17323778-17323778 on Build GRCh38
rs951619673 Chr.19: 17323846-17323846 on Build GRCh38
rs746772687 Chr.19: 17323597-17323597 on Build GRCh38
rs945780172 Chr.19: 17323793-17323793 on Build GRCh38
rs985300412 Chr.19: 17323730-17323730 on Build GRCh38
rs746391889 Chr.19: 17323547-17323547 on Build GRCh38
rs774954101 Chr.19: 17323622-17323622 on Build GRCh38
rs911115787 Chr.19: 17323734-17323734 on Build GRCh38
rs577685770 Chr.19: 17323921-17323921 on Build GRCh38
rs776371558 Chr.19: 17323607-17323607 on Build GRCh38
rs767939515 Chr.19: 17323693-17323693 on Build GRCh38
rs900117032 Chr.19: 17323666-17323666 on Build GRCh38
rs961029451 Chr.19: 17323956-17323956 on Build GRCh38
rs780309990 Chr.19: 17323569-17323569 on Build GRCh38
rs1029909070 Chr.19: 17323679-17323679 on Build GRCh38
rs757498816 Chr.19: 17323670-17323670 on Build GRCh38
rs1004650682 Chr.19: 17323909-17323909 on Build GRCh38
rs1015324297 Chr.19: 17323929-17323929 on Build GRCh38
rs775368243 Chr.19: 17323550-17323555 on Build GRCh38
rs906948330 Chr.19: 17323819-17323819 on Build GRCh38
rs764690354 Chr.19: 17323621-17323621 on Build GRCh38
rs988402490 Chr.19: 17323970-17323970 on Build GRCh38
rs962417322 Chr.19: 17323608-17323608 on Build GRCh38
rs1055901822 Chr.19: 17323916-17323916 on Build GRCh38
rs1045523086 Chr.19: 17323802-17323802 on Build GRCh38
rs757700130 Chr.19: 17323530-17323530 on Build GRCh38
rs762502115 Chr.19: 17323637-17323637 on Build GRCh38
rs895801714 Chr.19: 17323781-17323791 on Build GRCh38
rs1001737667 Chr.19: 17323540-17323540 on Build GRCh38
rs749419427 Chr.19: 17323579-17323579 on Build GRCh38
rs572701711 Chr.19: 17323936-17323936 on Build GRCh38

View Full Product Details