Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17327221-17327718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO8
Entrez Gene ID
57719
Gene Name
anoctamin 8
Gene Aliases
KIAA1623, TMEM16H
UniGene
Hs.590990
Gene Chromosome Location
Chr.19: 17323223-17334834 on Build GRCh38
Transcripts
NM_020959.2 XM_017027048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970795 c.2393G>A p.R798H Chr.19: 17327714-17327714 on Build GRCh38
5589672 c.2502C>T p.F834F Chr.19: 17327486-17327486 on Build GRCh38
5614615 c.2418G>A p.Q806Q Chr.19: 17327689-17327689 on Build GRCh38
7284300 c.2418+8delG p.? Chr.19: 17327681-17327681 on Build GRCh38
1750715 c.2418+2T>G p.? Chr.19: 17327687-17327687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527269675 Chr.19: 17327334-17327334 on Build GRCh38
rs780997885 Chr.19: 17327549-17327549 on Build GRCh38
rs745686709 Chr.19: 17327680-17327681 on Build GRCh38
rs899931873 Chr.19: 17327300-17327300 on Build GRCh38
rs956332627 Chr.19: 17327411-17327411 on Build GRCh38
rs543057123 Chr.19: 17327608-17327608 on Build GRCh38
rs768014396 Chr.19: 17327441-17327441 on Build GRCh38
rs990248775 Chr.19: 17327588-17327588 on Build GRCh38
rs761229392 Chr.19: 17327446-17327446 on Build GRCh38
rs995988694 Chr.19: 17327327-17327327 on Build GRCh38
rs549976404 Chr.19: 17327427-17327427 on Build GRCh38
rs1047043926 Chr.19: 17327263-17327263 on Build GRCh38
rs567330096 Chr.19: 17327393-17327393 on Build GRCh38
rs1001222376 Chr.19: 17327371-17327371 on Build GRCh38
rs753940125 Chr.19: 17327458-17327458 on Build GRCh38
rs773811139 Chr.19: 17327443-17327443 on Build GRCh38
rs774003160 Chr.19: 17327609-17327609 on Build GRCh38
rs755079048 Chr.19: 17327493-17327493 on Build GRCh38
rs1024693683 Chr.19: 17327424-17327424 on Build GRCh38
rs750210667 Chr.19: 17327686-17327686 on Build GRCh38
rs1017745198 Chr.19: 17327595-17327595 on Build GRCh38
rs141211641 Chr.19: 17327279-17327279 on Build GRCh38
rs779978623 Chr.19: 17327695-17327695 on Build GRCh38
rs558788957 Chr.19: 17327605-17327605 on Build GRCh38
rs756027211 Chr.19: 17327578-17327578 on Build GRCh38
rs751604601 Chr.19: 17327486-17327486 on Build GRCh38
rs199668891 Chr.19: 17327477-17327477 on Build GRCh38
rs372002710 Chr.19: 17327401-17327401 on Build GRCh38
rs773944466 Chr.19: 17327429-17327429 on Build GRCh38
rs771888009 Chr.19: 17327644-17327644 on Build GRCh38
rs771023313 Chr.19: 17327413-17327415 on Build GRCh38
rs763978088 Chr.19: 17327383-17327383 on Build GRCh38
rs762849091 Chr.19: 17327341-17327341 on Build GRCh38
rs967980362 Chr.19: 17327352-17327352 on Build GRCh38
rs564881471 Chr.19: 17327262-17327262 on Build GRCh38
rs774950009 Chr.19: 17327417-17327417 on Build GRCh38
rs757241050 Chr.19: 17327698-17327698 on Build GRCh38
rs1041086763 Chr.19: 17327658-17327658 on Build GRCh38
rs758589134 Chr.19: 17327681-17327681 on Build GRCh38
rs777469846 Chr.19: 17327348-17327352 on Build GRCh38
rs765432336 Chr.19: 17327521-17327521 on Build GRCh38
rs748802558 Chr.19: 17327418-17327418 on Build GRCh38
rs569885331 Chr.19: 17327430-17327430 on Build GRCh38
rs886936537 Chr.19: 17327280-17327280 on Build GRCh38
rs957521726 Chr.19: 17327471-17327471 on Build GRCh38
rs920116771 Chr.19: 17327474-17327474 on Build GRCh38
rs757030485 Chr.19: 17327542-17327542 on Build GRCh38
rs1030661026 Chr.19: 17327376-17327376 on Build GRCh38
rs114597852 Chr.19: 17327618-17327618 on Build GRCh38
rs75478510 Chr.19: 17327687-17327687 on Build GRCh38
rs201314015 Chr.19: 17327671-17327671 on Build GRCh38
rs752877827 Chr.19: 17327531-17327531 on Build GRCh38
rs764349656 Chr.19: 17327685-17327685 on Build GRCh38
rs780307970 Chr.19: 17327319-17327319 on Build GRCh38
rs529255891 Chr.19: 17327606-17327606 on Build GRCh38
rs201862681 Chr.19: 17327666-17327666 on Build GRCh38
rs111696480 Chr.19: 17327255-17327255 on Build GRCh38
rs766988121 Chr.19: 17327655-17327655 on Build GRCh38
rs777007317 Chr.19: 17327657-17327657 on Build GRCh38
rs894528980 Chr.19: 17327390-17327390 on Build GRCh38
rs750978772 Chr.19: 17327299-17327299 on Build GRCh38
rs545935904 Chr.19: 17327384-17327384 on Build GRCh38
rs756211334 Chr.19: 17327310-17327310 on Build GRCh38
rs376028722 Chr.19: 17327454-17327454 on Build GRCh38
rs781051384 Chr.19: 17327400-17327400 on Build GRCh38
rs983399802 Chr.19: 17327597-17327597 on Build GRCh38
rs773686108 Chr.19: 17327648-17327648 on Build GRCh38
rs547221815 Chr.19: 17327379-17327379 on Build GRCh38
rs746912281 Chr.19: 17327412-17327413 on Build GRCh38
rs776517161 Chr.19: 17327423-17327423 on Build GRCh38
rs761178747 Chr.19: 17327650-17327650 on Build GRCh38
rs745703687 Chr.19: 17327563-17327563 on Build GRCh38
rs758498116 Chr.19: 17327359-17327359 on Build GRCh38
rs752925514 Chr.19: 17327680-17327680 on Build GRCh38

View Full Product Details