Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17328056-17328551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO8
Entrez Gene ID
57719
Gene Name
anoctamin 8
Gene Aliases
KIAA1623, TMEM16H
UniGene
Hs.590990
Gene Chromosome Location
Chr.19: 17323223-17334834 on Build GRCh38
Transcripts
NM_020959.2 XM_017027048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756463 c.1877G>A p.R626Q Chr.19: 17328511-17328511 on Build GRCh38
3764708 c.1952C>T p.P651L Chr.19: 17328436-17328436 on Build GRCh38
4452837 c.2218A>T p.K740* Chr.19: 17328170-17328170 on Build GRCh38
7212314 c.2226+5G>T p.? Chr.19: 17328157-17328157 on Build GRCh38
7221506 c.2163G>A p.P721P Chr.19: 17328225-17328225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771946384 Chr.19: 17328342-17328342 on Build GRCh38
rs761093570 Chr.19: 17328211-17328212 on Build GRCh38
rs76655932 Chr.19: 17328511-17328511 on Build GRCh38
rs3745190 Chr.19: 17328129-17328129 on Build GRCh38
rs749819667 Chr.19: 17328112-17328112 on Build GRCh38
rs377396526 Chr.19: 17328316-17328316 on Build GRCh38
rs528803838 Chr.19: 17328341-17328341 on Build GRCh38
rs913770792 Chr.19: 17328154-17328155 on Build GRCh38
rs747905823 Chr.19: 17328513-17328513 on Build GRCh38
rs1001884575 Chr.19: 17328082-17328082 on Build GRCh38
rs566108107 Chr.19: 17328335-17328335 on Build GRCh38
rs753268979 Chr.19: 17328304-17328304 on Build GRCh38
rs200209917 Chr.19: 17328183-17328183 on Build GRCh38
rs3745191 Chr.19: 17328225-17328225 on Build GRCh38
rs776226696 Chr.19: 17328429-17328429 on Build GRCh38
rs373194725 Chr.19: 17328209-17328209 on Build GRCh38
rs762262865 Chr.19: 17328149-17328150 on Build GRCh38
rs952513209 Chr.19: 17328100-17328100 on Build GRCh38
rs748105742 Chr.19: 17328208-17328208 on Build GRCh38
rs561827919 Chr.19: 17328328-17328328 on Build GRCh38
rs750006508 Chr.19: 17328268-17328268 on Build GRCh38
rs200489783 Chr.19: 17328373-17328373 on Build GRCh38
rs756385506 Chr.19: 17328509-17328509 on Build GRCh38
rs750689373 Chr.19: 17328168-17328170 on Build GRCh38
rs761174039 Chr.19: 17328259-17328259 on Build GRCh38
rs772038959 Chr.19: 17328411-17328411 on Build GRCh38
rs750575220 Chr.19: 17328506-17328506 on Build GRCh38
rs112357946 Chr.19: 17328291-17328291 on Build GRCh38
rs766709864 Chr.19: 17328316-17328328 on Build GRCh38
rs982422874 Chr.19: 17328089-17328089 on Build GRCh38
rs781509842 Chr.19: 17328387-17328387 on Build GRCh38
rs113207944 Chr.19: 17328114-17328114 on Build GRCh38
rs974021168 Chr.19: 17328106-17328106 on Build GRCh38
rs201481629 Chr.19: 17328297-17328297 on Build GRCh38
rs759155514 Chr.19: 17328430-17328430 on Build GRCh38
rs760166919 Chr.19: 17328343-17328343 on Build GRCh38
rs201801028 Chr.19: 17328243-17328243 on Build GRCh38
rs772177029 Chr.19: 17328121-17328121 on Build GRCh38
rs767013840 Chr.19: 17328165-17328165 on Build GRCh38
rs774449784 Chr.19: 17328122-17328138 on Build GRCh38
rs915860543 Chr.19: 17328307-17328307 on Build GRCh38
rs878873291 Chr.19: 17328116-17328116 on Build GRCh38
rs760116254 Chr.19: 17328423-17328423 on Build GRCh38
rs962588789 Chr.19: 17328102-17328102 on Build GRCh38
rs746118960 Chr.19: 17328389-17328389 on Build GRCh38
rs572318281 Chr.19: 17328175-17328175 on Build GRCh38
rs188358285 Chr.19: 17328087-17328087 on Build GRCh38
rs888181588 Chr.19: 17328274-17328274 on Build GRCh38
rs768588684 Chr.19: 17328338-17328338 on Build GRCh38
rs764596282 Chr.19: 17328483-17328483 on Build GRCh38
rs955069997 Chr.19: 17328249-17328249 on Build GRCh38
rs748280945 Chr.19: 17328115-17328115 on Build GRCh38
rs139815862 Chr.19: 17328224-17328224 on Build GRCh38
rs773486993 Chr.19: 17328126-17328126 on Build GRCh38
rs747561299 Chr.19: 17328207-17328207 on Build GRCh38
rs777714339 Chr.19: 17328518-17328518 on Build GRCh38
rs1015800564 Chr.19: 17328103-17328103 on Build GRCh38
rs762954564 Chr.19: 17328156-17328156 on Build GRCh38
rs781628863 Chr.19: 17328323-17328323 on Build GRCh38
rs766075066 Chr.19: 17328286-17328286 on Build GRCh38
rs199879006 Chr.19: 17328408-17328408 on Build GRCh38
rs371069110 Chr.19: 17328145-17328145 on Build GRCh38
rs765618746 Chr.19: 17328363-17328363 on Build GRCh38
rs1003975556 Chr.19: 17328266-17328266 on Build GRCh38
rs141292857 Chr.19: 17328186-17328186 on Build GRCh38
rs768028524 Chr.19: 17328155-17328156 on Build GRCh38
rs757365626 Chr.19: 17328491-17328491 on Build GRCh38
rs759844321 Chr.19: 17328359-17328359 on Build GRCh38
rs1004773095 Chr.19: 17328101-17328101 on Build GRCh38
rs202118586 Chr.19: 17328400-17328400 on Build GRCh38
rs867074097 Chr.19: 17328398-17328399 on Build GRCh38
rs773023923 Chr.19: 17328419-17328419 on Build GRCh38
rs775911653 Chr.19: 17328435-17328435 on Build GRCh38
rs765945952 Chr.19: 17328347-17328347 on Build GRCh38
rs868509535 Chr.19: 17328409-17328409 on Build GRCh38
rs576350956 Chr.19: 17328123-17328123 on Build GRCh38
rs770449557 Chr.19: 17328425-17328425 on Build GRCh38
rs763134634 Chr.19: 17328117-17328133 on Build GRCh38
rs760349683 Chr.19: 17328279-17328279 on Build GRCh38
rs747948159 Chr.19: 17328407-17328407 on Build GRCh38
rs758368013 Chr.19: 17328311-17328311 on Build GRCh38
rs762899533 Chr.19: 17328238-17328238 on Build GRCh38
rs377730643 Chr.19: 17328138-17328138 on Build GRCh38
rs749025870 Chr.19: 17328337-17328337 on Build GRCh38
rs560940837 Chr.19: 17328188-17328188 on Build GRCh38
rs11667523 Chr.19: 17328109-17328109 on Build GRCh38
rs774438428 Chr.19: 17328255-17328255 on Build GRCh38
rs767740023 Chr.19: 17328500-17328500 on Build GRCh38
rs755462757 Chr.19: 17328199-17328199 on Build GRCh38
rs917340438 Chr.19: 17328399-17328399 on Build GRCh38
rs1036746006 Chr.19: 17328487-17328487 on Build GRCh38
rs200795418 Chr.19: 17328434-17328434 on Build GRCh38
rs753041457 Chr.19: 17328369-17328369 on Build GRCh38
rs566987257 Chr.19: 17328121-17328122 on Build GRCh38
rs933572176 Chr.19: 17328503-17328503 on Build GRCh38
rs548557928 Chr.19: 17328349-17328349 on Build GRCh38
rs150766407 Chr.19: 17328216-17328216 on Build GRCh38
rs978797205 Chr.19: 17328125-17328125 on Build GRCh38
rs998360049 Chr.19: 17328339-17328339 on Build GRCh38
rs908212997 Chr.19: 17328097-17328097 on Build GRCh38
rs537439476 Chr.19: 17328474-17328474 on Build GRCh38
rs773256997 Chr.19: 17328215-17328215 on Build GRCh38
rs756859073 Chr.19: 17328158-17328158 on Build GRCh38
rs374120885 Chr.19: 17328318-17328318 on Build GRCh38
rs775768790 Chr.19: 17328111-17328112 on Build GRCh38
rs764178395 Chr.19: 17328157-17328157 on Build GRCh38
rs370432348 Chr.19: 17328317-17328317 on Build GRCh38
rs758138981 Chr.19: 17328516-17328516 on Build GRCh38
rs779459801 Chr.19: 17328203-17328203 on Build GRCh38
rs778741457 Chr.19: 17328405-17328405 on Build GRCh38
rs900934423 Chr.19: 17328493-17328493 on Build GRCh38
rs767078644 Chr.19: 17328261-17328261 on Build GRCh38
rs928460128 Chr.19: 17328354-17328354 on Build GRCh38
rs552722166 Chr.19: 17328298-17328298 on Build GRCh38
rs758917864 Chr.19: 17328372-17328372 on Build GRCh38
rs757418390 Chr.19: 17328374-17328374 on Build GRCh38
rs888160751 Chr.19: 17328470-17328470 on Build GRCh38
rs752021536 Chr.19: 17328489-17328489 on Build GRCh38

View Full Product Details