Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNG12
Entrez Gene ID
55970
Gene Name
G protein subunit gamma 12
Gene Aliases
-
Chromosome Location
Chr.1: 67701466 - 67833779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018841.5 NP_061329.3 3 - 4 299 112
XM_017001809.1 XP_016857298.1 3 - 4 466 112
XM_017001810.1 XP_016857299.1 4 - 5 707 112
XM_017001811.1 XP_016857300.1 4 - 5 636 112
GenBank mRNA AF119663.1 - 3 - 4 283 112
AF123766.1 - 3 - 4 290 112
AF365871.1 - 3 - 4 226 112
AF493879.1 - 1 - 2 94 112
AK293101.1 - 4 - 5 240 112
AL832431.1 - 3 - 4 252 112
BC005940.1 - 3 - 4 112
BP236225.1 - 3 - 4 286 112

View Full Product Details