The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021138.3 | NP_066961.2 | 6 - 7 | 662 | 72 |
XM_011518973.2 | XP_011517275.1 | 7 - 8 | 966 | 72 | |
XM_011518974.2 | XP_011517276.1 | 6 - 7 | 1567 | 72 | |
XM_011518975.2 | XP_011517277.1 | 7 - 8 | 894 | 72 | |
XM_011518976.2 | XP_011517278.1 | 7 - 8 | 935 | 72 | |
XM_011518977.2 | XP_011517279.1 | 6 - 7 | 655 | 72 | |
XM_011518978.2 | XP_011517280.1 | 6 - 7 | 2076 | 72 | |
XM_017015094.1 | XP_016870583.1 | 6 - 7 | 710 | 72 | |
XM_017015095.1 | XP_016870584.1 | 6 - 7 | 1420 | 72 | |
GenBank mRNA | AK289722.1 | - | 7 - 8 | 778 | 72 |
AK294607.1 | - | 6 - 7 | 521 | 72 | |
AK298370.1 | - | 6 - 7 | 626 | 72 | |
BC032410.1 | - | 5 - 6 | 648 | 72 | |
BC033810.1 | - | 6 - 7 | 650 | 72 | |
BC043492.1 | - | 6 - 7 | 662 | 72 | |
BC064662.1 | - | 5 - 6 | 657 | 72 | |
HM991672.1 | - | 5 - 6 | 605 | 72 |