Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40231961-40232459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT2
Entrez Gene ID
208
Gene Name
AKT serine/threonine kinase 2
Gene Aliases
HIHGHH, PKBB, PKBBETA, PRKBB, RAC-BETA
UniGene
Hs.631535
Gene Chromosome Location
Chr.19: 40230317-40285531 on Build GRCh38
Transcripts
NM_001243027.2 NM_001243028.2 NM_001626.5 XM_011526614.1 XM_011526615.1 XM_011526616.1 XM_011526618.1 XM_011526619.1 XM_011526620.1 XM_017026470.1 XM_017026471.1

SNP Detail

SNP ID Chromosome Location
rs982754069 Chr.19: 40232232-40232232 on Build GRCh38
rs944606379 Chr.19: 40232122-40232122 on Build GRCh38
rs1048878008 Chr.19: 40232402-40232402 on Build GRCh38
rs570168496 Chr.19: 40232222-40232222 on Build GRCh38
rs959821983 Chr.19: 40232247-40232247 on Build GRCh38
rs548384201 Chr.19: 40232220-40232220 on Build GRCh38
rs149821698 Chr.19: 40232424-40232424 on Build GRCh38
rs906882635 Chr.19: 40232433-40232433 on Build GRCh38
rs915332931 Chr.19: 40232278-40232278 on Build GRCh38
rs766687564 Chr.19: 40232290-40232290 on Build GRCh38
rs1004543369 Chr.19: 40232189-40232189 on Build GRCh38
rs1012496808 Chr.19: 40232006-40232006 on Build GRCh38
rs144053442 Chr.19: 40232103-40232103 on Build GRCh38
rs1015020094 Chr.19: 40232050-40232051 on Build GRCh38
rs1014925250 Chr.19: 40232236-40232236 on Build GRCh38
rs900350017 Chr.19: 40232139-40232139 on Build GRCh38
rs557611072 Chr.19: 40232114-40232114 on Build GRCh38
rs779462254 Chr.19: 40232415-40232415 on Build GRCh38
rs574929065 Chr.19: 40232401-40232401 on Build GRCh38
rs923607856 Chr.19: 40232386-40232386 on Build GRCh38
rs530467989 Chr.19: 40232092-40232092 on Build GRCh38
rs1027367281 Chr.19: 40232160-40232160 on Build GRCh38
rs762055206 Chr.19: 40232112-40232112 on Build GRCh38
rs983428151 Chr.19: 40232024-40232024 on Build GRCh38
rs1035255258 Chr.19: 40232238-40232238 on Build GRCh38
rs755489698 Chr.19: 40232397-40232397 on Build GRCh38
rs952013192 Chr.19: 40232021-40232021 on Build GRCh38
rs887989486 Chr.19: 40232207-40232207 on Build GRCh38
rs528362334 Chr.19: 40232121-40232121 on Build GRCh38
rs556763599 Chr.19: 40232436-40232436 on Build GRCh38
rs758179603 Chr.19: 40232425-40232425 on Build GRCh38
rs923917955 Chr.19: 40232318-40232318 on Build GRCh38
rs549882888 Chr.19: 40232132-40232132 on Build GRCh38
rs41275750 Chr.19: 40232188-40232188 on Build GRCh38
rs563443135 Chr.19: 40232107-40232107 on Build GRCh38
rs537029863 Chr.19: 40232249-40232249 on Build GRCh38
rs188407538 Chr.19: 40232364-40232364 on Build GRCh38
rs982337076 Chr.19: 40232094-40232094 on Build GRCh38
rs953913715 Chr.19: 40232346-40232346 on Build GRCh38
rs946604261 Chr.19: 40232356-40232356 on Build GRCh38
rs534945416 Chr.19: 40232349-40232349 on Build GRCh38
rs375232480 Chr.19: 40232203-40232203 on Build GRCh38
rs184160447 Chr.19: 40232038-40232038 on Build GRCh38
rs529133488 Chr.19: 40232218-40232218 on Build GRCh38
rs978113874 Chr.19: 40232312-40232312 on Build GRCh38
rs140298279 Chr.19: 40232080-40232080 on Build GRCh38
rs1012942447 Chr.19: 40232165-40232165 on Build GRCh38
rs1028093380 Chr.19: 40232019-40232019 on Build GRCh38
rs555931423 Chr.19: 40232311-40232311 on Build GRCh38
rs895298949 Chr.19: 40232150-40232150 on Build GRCh38
rs375216036 Chr.19: 40232135-40232135 on Build GRCh38
rs900382192 Chr.19: 40232384-40232384 on Build GRCh38
rs966155699 Chr.19: 40232237-40232237 on Build GRCh38
rs868724745 Chr.19: 40232300-40232300 on Build GRCh38
rs1004577585 Chr.19: 40232423-40232423 on Build GRCh38
rs1042374100 Chr.19: 40232118-40232118 on Build GRCh38
rs147329443 Chr.19: 40232108-40232108 on Build GRCh38
rs931810053 Chr.19: 40232396-40232396 on Build GRCh38
rs570526739 Chr.19: 40232313-40232313 on Build GRCh38
rs985384093 Chr.19: 40232102-40232102 on Build GRCh38
rs966085133 Chr.19: 40232072-40232072 on Build GRCh38
rs1056204374 Chr.19: 40232411-40232411 on Build GRCh38
rs41275748 Chr.19: 40232143-40232143 on Build GRCh38
rs990972544 Chr.19: 40232267-40232267 on Build GRCh38
rs552182357 Chr.19: 40232113-40232113 on Build GRCh38
rs1029936884 Chr.19: 40232345-40232345 on Build GRCh38
rs915163098 Chr.19: 40232355-40232355 on Build GRCh38
rs765587103 Chr.19: 40232417-40232419 on Build GRCh38
rs895420165 Chr.19: 40232002-40232002 on Build GRCh38
rs865820250 Chr.19: 40232380-40232380 on Build GRCh38

View Full Product Details