Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40395078-40395587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
UniGene
Hs.205457
Gene Chromosome Location
Chr.19: 40393762-40414718 on Build GRCh38
Transcripts
NM_020956.2 NM_181882.2 XM_011527171.2 XM_017027046.1 XM_017027047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6757326 c.3156G>A p.W1052* Chr.19: 40395196-40395196 on Build GRCh38
1641162 c.3160G>A p.G1054R Chr.19: 40395192-40395192 on Build GRCh38
4753501 c.3106G>T p.A1036S Chr.19: 40395246-40395246 on Build GRCh38
4140646 c.2775C>G p.V925V Chr.19: 40395577-40395577 on Build GRCh38
6237027 c.2852C>T p.A951V Chr.19: 40395500-40395500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746300226 Chr.19: 40395256-40395256 on Build GRCh38
rs1008150085 Chr.19: 40395287-40395287 on Build GRCh38
rs548086012 Chr.19: 40395315-40395315 on Build GRCh38
rs762456536 Chr.19: 40395404-40395404 on Build GRCh38
rs766119564 Chr.19: 40395311-40395311 on Build GRCh38
rs968818827 Chr.19: 40395542-40395542 on Build GRCh38
rs562422065 Chr.19: 40395414-40395414 on Build GRCh38
rs758668778 Chr.19: 40395460-40395460 on Build GRCh38
rs996865233 Chr.19: 40395341-40395341 on Build GRCh38
rs770520776 Chr.19: 40395502-40395502 on Build GRCh38
rs200045096 Chr.19: 40395114-40395114 on Build GRCh38
rs772591261 Chr.19: 40395400-40395400 on Build GRCh38
rs767995607 Chr.19: 40395116-40395116 on Build GRCh38
rs112544351 Chr.19: 40395535-40395535 on Build GRCh38
rs748106303 Chr.19: 40395234-40395234 on Build GRCh38
rs937051533 Chr.19: 40395162-40395162 on Build GRCh38
rs762213719 Chr.19: 40395113-40395113 on Build GRCh38
rs773246710 Chr.19: 40395111-40395111 on Build GRCh38
rs376423497 Chr.19: 40395453-40395453 on Build GRCh38
rs767386430 Chr.19: 40395419-40395419 on Build GRCh38
rs774666043 Chr.19: 40395105-40395105 on Build GRCh38
rs374750300 Chr.19: 40395441-40395441 on Build GRCh38
rs781205858 Chr.19: 40395230-40395230 on Build GRCh38
rs747451387 Chr.19: 40395396-40395396 on Build GRCh38
rs769263827 Chr.19: 40395399-40395399 on Build GRCh38
rs767439555 Chr.19: 40395316-40395316 on Build GRCh38
rs868592190 Chr.19: 40395377-40395377 on Build GRCh38
rs748965299 Chr.19: 40395411-40395431 on Build GRCh38
rs536876712 Chr.19: 40395384-40395384 on Build GRCh38
rs868460197 Chr.19: 40395459-40395459 on Build GRCh38
rs928322159 Chr.19: 40395159-40395159 on Build GRCh38
rs104894708 Chr.19: 40395144-40395144 on Build GRCh38
rs769156028 Chr.19: 40395299-40395299 on Build GRCh38
rs903097017 Chr.19: 40395260-40395260 on Build GRCh38
rs145226687 Chr.19: 40395520-40395520 on Build GRCh38
rs372715275 Chr.19: 40395102-40395102 on Build GRCh38
rs935218572 Chr.19: 40395393-40395393 on Build GRCh38
rs771237505 Chr.19: 40395245-40395245 on Build GRCh38
rs571311308 Chr.19: 40395243-40395243 on Build GRCh38
rs34766564 Chr.19: 40395338-40395338 on Build GRCh38
rs772380056 Chr.19: 40395390-40395390 on Build GRCh38
rs143873932 Chr.19: 40395239-40395239 on Build GRCh38
rs566114248 Chr.19: 40395356-40395356 on Build GRCh38
rs371192524 Chr.19: 40395405-40395405 on Build GRCh38
rs538922683 Chr.19: 40395302-40395302 on Build GRCh38
rs752104110 Chr.19: 40395286-40395286 on Build GRCh38
rs537040907 Chr.19: 40395507-40395507 on Build GRCh38
rs139188673 Chr.19: 40395166-40395166 on Build GRCh38
rs765924621 Chr.19: 40395353-40395353 on Build GRCh38
rs764547883 Chr.19: 40395205-40395210 on Build GRCh38
rs779042943 Chr.19: 40395361-40395361 on Build GRCh38
rs961710252 Chr.19: 40395466-40395466 on Build GRCh38
rs775919265 Chr.19: 40395284-40395284 on Build GRCh38
rs561080969 Chr.19: 40395188-40395188 on Build GRCh38
rs199734816 Chr.19: 40395521-40395521 on Build GRCh38
rs748622767 Chr.19: 40395501-40395501 on Build GRCh38
rs3745202 Chr.19: 40395104-40395104 on Build GRCh38
rs148600818 Chr.19: 40395242-40395242 on Build GRCh38
rs62109486 Chr.19: 40395277-40395277 on Build GRCh38
rs139700059 Chr.19: 40395515-40395515 on Build GRCh38
rs761267361 Chr.19: 40395292-40395292 on Build GRCh38
rs375773532 Chr.19: 40395148-40395148 on Build GRCh38
rs142228732 Chr.19: 40395409-40395409 on Build GRCh38
rs104894714 Chr.19: 40395495-40395495 on Build GRCh38
rs751456785 Chr.19: 40395206-40395206 on Build GRCh38
rs778987729 Chr.19: 40395450-40395450 on Build GRCh38
rs758109040 Chr.19: 40395176-40395176 on Build GRCh38
rs759290682 Chr.19: 40395417-40395417 on Build GRCh38
rs186086914 Chr.19: 40395207-40395207 on Build GRCh38
rs1027083300 Chr.19: 40395123-40395123 on Build GRCh38
rs779929777 Chr.19: 40395101-40395101 on Build GRCh38
rs764817403 Chr.19: 40395152-40395152 on Build GRCh38
rs954851586 Chr.19: 40395285-40395285 on Build GRCh38
rs764162630 Chr.19: 40395443-40395443 on Build GRCh38
rs986274653 Chr.19: 40395294-40395294 on Build GRCh38
rs1043055387 Chr.19: 40395379-40395379 on Build GRCh38
rs200424895 Chr.19: 40395395-40395395 on Build GRCh38
rs750559680 Chr.19: 40395559-40395559 on Build GRCh38
rs947190221 Chr.19: 40395228-40395228 on Build GRCh38
rs752590323 Chr.19: 40395429-40395429 on Build GRCh38
rs765270572 Chr.19: 40395545-40395545 on Build GRCh38
rs758500214 Chr.19: 40395099-40395099 on Build GRCh38
rs752638036 Chr.19: 40395339-40395339 on Build GRCh38
rs147842738 Chr.19: 40395504-40395504 on Build GRCh38
rs747471167 Chr.19: 40395492-40395492 on Build GRCh38
rs886054436 Chr.19: 40395132-40395132 on Build GRCh38
rs777695738 Chr.19: 40395355-40395355 on Build GRCh38
rs774823527 Chr.19: 40395115-40395115 on Build GRCh38
rs372961087 Chr.19: 40395447-40395447 on Build GRCh38
rs981510213 Chr.19: 40395158-40395158 on Build GRCh38
rs971346863 Chr.19: 40395432-40395432 on Build GRCh38
rs761769929 Chr.19: 40395536-40395536 on Build GRCh38
rs554688365 Chr.19: 40395394-40395394 on Build GRCh38
rs879940695 Chr.19: 40395424-40395424 on Build GRCh38
rs1027011237 Chr.19: 40395349-40395349 on Build GRCh38
rs1005848599 Chr.19: 40395132-40395132 on Build GRCh38
rs746910596 Chr.19: 40395103-40395103 on Build GRCh38
rs776607887 Chr.19: 40395344-40395344 on Build GRCh38
rs146222815 Chr.19: 40395143-40395143 on Build GRCh38
rs1035734860 Chr.19: 40395477-40395477 on Build GRCh38
rs369688519 Chr.19: 40395493-40395493 on Build GRCh38
rs61733451 Chr.19: 40395134-40395134 on Build GRCh38
rs757620514 Chr.19: 40395350-40395350 on Build GRCh38
rs780382855 Chr.19: 40395483-40395483 on Build GRCh38
rs369224308 Chr.19: 40395444-40395444 on Build GRCh38
rs772827175 Chr.19: 40395300-40395300 on Build GRCh38
rs774688060 Chr.19: 40395250-40395250 on Build GRCh38
rs758614032 Chr.19: 40395562-40395562 on Build GRCh38
rs757419019 Chr.19: 40395360-40395360 on Build GRCh38
rs781470128 Chr.19: 40395393-40395394 on Build GRCh38
rs772431799 Chr.19: 40395258-40395258 on Build GRCh38
rs781679882 Chr.19: 40395496-40395496 on Build GRCh38
rs771912595 Chr.19: 40395511-40395511 on Build GRCh38

View Full Product Details