The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017418.2 | NP_059114.1 | 1 - 2 | 51 | 80 |
GenBank mRNA | AB022761.1 | - | 1 - 2 | 51 | 80 |