The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013229.2 | NP_037361.1 | 18 - 19 | 3139 | 63 |
NM_181861.1 | NP_863651.1 | 18 - 19 | 3172 | 63 | |
GenBank mRNA | AB007873.2 | - | 17 - 18 | 2562 | 63 |
AF134397.1 | - | 17 - 18 | 2562 | 63 | |
AF149794.1 | - | 17 - 18 | 2595 | 63 | |
AJ243009.1 | - | 17 - 18 | 2595 | 63 | |
AJ243048.1 | - | 17 - 18 | 2595 | 63 | |
BC136531.1 | - | 18 - 19 | 2743 | 63 | |
BC136532.1 | - | 18 - 19 | 2743 | 63 | |
BC150255.1 | - | 17 - 18 | 2591 | 63 |